Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 22585190-22585693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA2
Entrez Gene ID
3170
Gene Name
forkhead box A2
Gene Aliases
HNF3B, TCF3B
UniGene
Hs.155651
Gene Chromosome Location
Chr.20: 22581004-22585463 on Build GRCh38
Transcripts
NM_153675.2

SNP Detail

SNP ID Chromosome Location
rs912454128 Chr.20: 22585360-22585360 on Build GRCh38
rs72470552 Chr.20: 22585399-22585399 on Build GRCh38
rs948796196 Chr.20: 22585354-22585354 on Build GRCh38
rs919957356 Chr.20: 22585542-22585542 on Build GRCh38
rs972681822 Chr.20: 22585541-22585541 on Build GRCh38
rs2277764 Chr.20: 22585486-22585486 on Build GRCh38
rs1044240751 Chr.20: 22585545-22585545 on Build GRCh38
rs966442849 Chr.20: 22585331-22585331 on Build GRCh38
rs113235858 Chr.20: 22585396-22585396 on Build GRCh38
rs970026473 Chr.20: 22585252-22585252 on Build GRCh38
rs868040510 Chr.20: 22585437-22585437 on Build GRCh38
rs953902507 Chr.20: 22585304-22585304 on Build GRCh38
rs72470553 Chr.20: 22585336-22585336 on Build GRCh38
rs970621127 Chr.20: 22585502-22585502 on Build GRCh38
rs41279040 Chr.20: 22585376-22585376 on Build GRCh38
rs868358540 Chr.20: 22585452-22585452 on Build GRCh38
rs72470551 Chr.20: 22585427-22585427 on Build GRCh38
rs766527225 Chr.20: 22585465-22585465 on Build GRCh38
rs537681032 Chr.20: 22585539-22585539 on Build GRCh38
rs761716156 Chr.20: 22585382-22585382 on Build GRCh38
rs939989176 Chr.20: 22585366-22585366 on Build GRCh38
rs574332953 Chr.20: 22585337-22585337 on Build GRCh38
rs928617631 Chr.20: 22585365-22585365 on Build GRCh38
rs1050051962 Chr.20: 22585592-22585592 on Build GRCh38
rs568698697 Chr.20: 22585533-22585533 on Build GRCh38
rs1035708071 Chr.20: 22585412-22585412 on Build GRCh38
rs776985975 Chr.20: 22585217-22585217 on Build GRCh38
rs1005325484 Chr.20: 22585637-22585637 on Build GRCh38
rs1050806399 Chr.20: 22585344-22585344 on Build GRCh38
rs559960427 Chr.20: 22585359-22585359 on Build GRCh38
rs755198160 Chr.20: 22585512-22585512 on Build GRCh38
rs931548847 Chr.20: 22585550-22585550 on Build GRCh38
rs990623145 Chr.20: 22585342-22585342 on Build GRCh38
rs983997704 Chr.20: 22585307-22585307 on Build GRCh38
rs546374465 Chr.20: 22585362-22585362 on Build GRCh38
rs947205254 Chr.20: 22585543-22585543 on Build GRCh38
rs563291785 Chr.20: 22585241-22585241 on Build GRCh38
rs1002917265 Chr.20: 22585258-22585258 on Build GRCh38
rs547785059 Chr.20: 22585471-22585471 on Build GRCh38
rs751462631 Chr.20: 22585313-22585313 on Build GRCh38
rs1008007814 Chr.20: 22585310-22585310 on Build GRCh38
rs951010068 Chr.20: 22585414-22585414 on Build GRCh38
rs554541466 Chr.20: 22585664-22585664 on Build GRCh38
rs889986054 Chr.20: 22585433-22585433 on Build GRCh38
rs7353110 Chr.20: 22585394-22585394 on Build GRCh38
rs370688536 Chr.20: 22585361-22585361 on Build GRCh38
rs532824108 Chr.20: 22585384-22585384 on Build GRCh38
rs1028468883 Chr.20: 22585286-22585286 on Build GRCh38
rs905982921 Chr.20: 22585407-22585407 on Build GRCh38
rs150761586 Chr.20: 22585268-22585268 on Build GRCh38
rs371100879 Chr.20: 22585340-22585340 on Build GRCh38
rs796342500 Chr.20: 22585467-22585467 on Build GRCh38
rs957943971 Chr.20: 22585341-22585341 on Build GRCh38
rs928695068 Chr.20: 22585529-22585529 on Build GRCh38
rs995730488 Chr.20: 22585413-22585413 on Build GRCh38
rs1003275318 Chr.20: 22585409-22585409 on Build GRCh38
rs889944100 Chr.20: 22585626-22585626 on Build GRCh38
rs938798956 Chr.20: 22585589-22585589 on Build GRCh38
rs1019395174 Chr.20: 22585445-22585445 on Build GRCh38
rs944186336 Chr.20: 22585666-22585666 on Build GRCh38
rs1019681370 Chr.20: 22585321-22585321 on Build GRCh38
rs916446234 Chr.20: 22585353-22585353 on Build GRCh38
rs539547448 Chr.20: 22585484-22585484 on Build GRCh38
rs886450592 Chr.20: 22585620-22585620 on Build GRCh38
rs951026743 Chr.20: 22585271-22585271 on Build GRCh38
rs934531659 Chr.20: 22585486-22585487 on Build GRCh38
rs141207172 Chr.20: 22585243-22585243 on Build GRCh38
rs957927371 Chr.20: 22585466-22585466 on Build GRCh38
rs986841256 Chr.20: 22585477-22585477 on Build GRCh38
rs531983388 Chr.20: 22585392-22585392 on Build GRCh38
rs974946283 Chr.20: 22585448-22585448 on Build GRCh38
rs72470550 Chr.20: 22585459-22585459 on Build GRCh38
rs1022879672 Chr.20: 22585244-22585244 on Build GRCh38
rs570815274 Chr.20: 22585481-22585481 on Build GRCh38
rs1053338158 Chr.20: 22585363-22585363 on Build GRCh38

View Full Product Details