Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23491049-23491555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST8
Entrez Gene ID
10047
Gene Name
cystatin 8
Gene Aliases
CRES, CTES5
UniGene
Hs.121602
Gene Chromosome Location
Chr.20: 23491077-23507124 on Build GRCh38
Transcripts
NM_001281730.1 NM_005492.3

SNP Detail

SNP ID Chromosome Location
rs979829554 Chr.20: 23491128-23491128 on Build GRCh38
rs542039053 Chr.20: 23491202-23491202 on Build GRCh38
rs573193142 Chr.20: 23491478-23491478 on Build GRCh38
rs992382071 Chr.20: 23491147-23491147 on Build GRCh38
rs1034029957 Chr.20: 23491350-23491350 on Build GRCh38
rs564320680 Chr.20: 23491256-23491256 on Build GRCh38
rs780450647 Chr.20: 23491332-23491332 on Build GRCh38
rs760778456 Chr.20: 23491274-23491274 on Build GRCh38
rs1012001837 Chr.20: 23491155-23491155 on Build GRCh38
rs1039084582 Chr.20: 23491205-23491205 on Build GRCh38
rs774432895 Chr.20: 23491078-23491078 on Build GRCh38
rs570071668 Chr.20: 23491447-23491447 on Build GRCh38
rs902346668 Chr.20: 23491109-23491109 on Build GRCh38
rs922590656 Chr.20: 23491095-23491095 on Build GRCh38
rs143039479 Chr.20: 23491117-23491117 on Build GRCh38
rs938314090 Chr.20: 23491138-23491138 on Build GRCh38
rs539060420 Chr.20: 23491534-23491534 on Build GRCh38
rs1010102678 Chr.20: 23491297-23491297 on Build GRCh38
rs527390189 Chr.20: 23491424-23491424 on Build GRCh38
rs185071900 Chr.20: 23491075-23491075 on Build GRCh38
rs955877004 Chr.20: 23491087-23491087 on Build GRCh38
rs112494567 Chr.20: 23491312-23491312 on Build GRCh38
rs562788814 Chr.20: 23491307-23491307 on Build GRCh38
rs929499584 Chr.20: 23491191-23491191 on Build GRCh38
rs546133278 Chr.20: 23491233-23491233 on Build GRCh38
rs555956022 Chr.20: 23491203-23491203 on Build GRCh38
rs767647189 Chr.20: 23491100-23491100 on Build GRCh38
rs1021872357 Chr.20: 23491302-23491302 on Build GRCh38
rs541555679 Chr.20: 23491243-23491243 on Build GRCh38
rs2073299 Chr.20: 23491358-23491358 on Build GRCh38
rs900517631 Chr.20: 23491232-23491232 on Build GRCh38
rs1053641327 Chr.20: 23491403-23491403 on Build GRCh38
rs575639098 Chr.20: 23491088-23491088 on Build GRCh38
rs942184002 Chr.20: 23491485-23491485 on Build GRCh38
rs1048349085 Chr.20: 23491201-23491201 on Build GRCh38
rs577770724 Chr.20: 23491348-23491348 on Build GRCh38
rs964134495 Chr.20: 23491299-23491299 on Build GRCh38
rs553477603 Chr.20: 23491073-23491073 on Build GRCh38
rs893587745 Chr.20: 23491145-23491145 on Build GRCh38
rs149448894 Chr.20: 23491086-23491086 on Build GRCh38
rs547469180 Chr.20: 23491429-23491429 on Build GRCh38
rs6132632 Chr.20: 23491525-23491525 on Build GRCh38
rs918197977 Chr.20: 23491385-23491385 on Build GRCh38
rs568283031 Chr.20: 23491418-23491418 on Build GRCh38
rs138573680 Chr.20: 23491304-23491304 on Build GRCh38
rs914359116 Chr.20: 23491102-23491102 on Build GRCh38
rs572432744 Chr.20: 23491230-23491230 on Build GRCh38
rs992434586 Chr.20: 23491365-23491365 on Build GRCh38
rs909380038 Chr.20: 23491479-23491479 on Build GRCh38
rs997495988 Chr.20: 23491251-23491251 on Build GRCh38
rs918144216 Chr.20: 23491172-23491172 on Build GRCh38
rs188557431 Chr.20: 23491134-23491134 on Build GRCh38
rs76662912 Chr.20: 23491267-23491267 on Build GRCh38
rs906219367 Chr.20: 23491518-23491518 on Build GRCh38

View Full Product Details