Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23564643-23565145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST9L
Entrez Gene ID
128821
Gene Name
cystatin 9-like
Gene Aliases
CTES7B, bA218C14.1
UniGene
Hs.121554
Gene Chromosome Location
Chr.20: 23564732-23568749 on Build GRCh38
Transcripts
NM_080610.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4097162 c.390C>A p.P130P Chr.20: 23565002-23565002 on Build GRCh38
6159603 c.441C>A p.H147Q Chr.20: 23564951-23564951 on Build GRCh38
4844015 c.386G>C p.R129T Chr.20: 23565006-23565006 on Build GRCh38
3740061 c.435A>G p.G145G Chr.20: 23564957-23564957 on Build GRCh38
3963401 c.372C>T p.F124F Chr.20: 23565020-23565020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894028924 Chr.20: 23565050-23565050 on Build GRCh38
rs376354808 Chr.20: 23565087-23565087 on Build GRCh38
rs760544889 Chr.20: 23564971-23564971 on Build GRCh38
rs941050763 Chr.20: 23565101-23565101 on Build GRCh38
rs759250255 Chr.20: 23565004-23565004 on Build GRCh38
rs886805950 Chr.20: 23564801-23564801 on Build GRCh38
rs750882629 Chr.20: 23565026-23565026 on Build GRCh38
rs528187585 Chr.20: 23564671-23564671 on Build GRCh38
rs989767490 Chr.20: 23564815-23564815 on Build GRCh38
rs146390169 Chr.20: 23565021-23565021 on Build GRCh38
rs367975014 Chr.20: 23564985-23564985 on Build GRCh38
rs143008910 Chr.20: 23564917-23564917 on Build GRCh38
rs768974494 Chr.20: 23564799-23564799 on Build GRCh38
rs749423942 Chr.20: 23565053-23565053 on Build GRCh38
rs370309011 Chr.20: 23565042-23565042 on Build GRCh38
rs757402883 Chr.20: 23564686-23564686 on Build GRCh38
rs1038019410 Chr.20: 23564832-23564832 on Build GRCh38
rs924637147 Chr.20: 23564670-23564670 on Build GRCh38
rs558947955 Chr.20: 23564973-23564973 on Build GRCh38
rs776347598 Chr.20: 23565000-23565000 on Build GRCh38
rs939156099 Chr.20: 23564770-23564770 on Build GRCh38
rs370091230 Chr.20: 23565068-23565068 on Build GRCh38
rs939170298 Chr.20: 23565046-23565046 on Build GRCh38
rs780118220 Chr.20: 23564934-23564934 on Build GRCh38
rs550933093 Chr.20: 23564731-23564731 on Build GRCh38
rs760270167 Chr.20: 23564898-23564898 on Build GRCh38
rs1046830732 Chr.20: 23564791-23564791 on Build GRCh38
rs997474257 Chr.20: 23564841-23564841 on Build GRCh38
rs779145789 Chr.20: 23565065-23565065 on Build GRCh38
rs545929857 Chr.20: 23564704-23564704 on Build GRCh38
rs929346541 Chr.20: 23564972-23564972 on Build GRCh38
rs763125567 Chr.20: 23565079-23565079 on Build GRCh38
rs755170627 Chr.20: 23565055-23565055 on Build GRCh38
rs374521793 Chr.20: 23565005-23565005 on Build GRCh38
rs998563949 Chr.20: 23565096-23565096 on Build GRCh38
rs199758673 Chr.20: 23564968-23564968 on Build GRCh38
rs749442325 Chr.20: 23564937-23564937 on Build GRCh38
rs138984571 Chr.20: 23564903-23564903 on Build GRCh38
rs993581490 Chr.20: 23564717-23564717 on Build GRCh38
rs148541767 Chr.20: 23565118-23565118 on Build GRCh38
rs866228123 Chr.20: 23564958-23564958 on Build GRCh38
rs1025116910 Chr.20: 23564727-23564727 on Build GRCh38
rs1046904039 Chr.20: 23565012-23565012 on Build GRCh38
rs942005721 Chr.20: 23564688-23564688 on Build GRCh38
rs575561554 Chr.20: 23564989-23564989 on Build GRCh38
rs539160143 Chr.20: 23564969-23564969 on Build GRCh38
rs183694586 Chr.20: 23564902-23564902 on Build GRCh38
rs967062492 Chr.20: 23564873-23564873 on Build GRCh38
rs957354250 Chr.20: 23564677-23564677 on Build GRCh38
rs191147047 Chr.20: 23564831-23564831 on Build GRCh38
rs781744354 Chr.20: 23564921-23564921 on Build GRCh38
rs771848866 Chr.20: 23565078-23565078 on Build GRCh38
rs958355727 Chr.20: 23564794-23564794 on Build GRCh38
rs757687248 Chr.20: 23564920-23564920 on Build GRCh38
rs927777362 Chr.20: 23564767-23564767 on Build GRCh38
rs1054613 Chr.20: 23564922-23564922 on Build GRCh38
rs969364561 Chr.20: 23564976-23564976 on Build GRCh38
rs1030477268 Chr.20: 23565117-23565117 on Build GRCh38
rs990079937 Chr.20: 23564702-23564702 on Build GRCh38
rs780099822 Chr.20: 23565036-23565036 on Build GRCh38
rs1034328650 Chr.20: 23564784-23564784 on Build GRCh38
rs947923765 Chr.20: 23564712-23564712 on Build GRCh38
rs375087614 Chr.20: 23565080-23565080 on Build GRCh38
rs373620373 Chr.20: 23564939-23564939 on Build GRCh38
rs1029866047 Chr.20: 23564845-23564845 on Build GRCh38
rs761253291 Chr.20: 23564719-23564719 on Build GRCh38
rs376798152 Chr.20: 23564959-23564959 on Build GRCh38
rs1037663734 Chr.20: 23564954-23564954 on Build GRCh38
rs900084482 Chr.20: 23564835-23564835 on Build GRCh38
rs955708499 Chr.20: 23564870-23564870 on Build GRCh38
rs1002779811 Chr.20: 23565029-23565029 on Build GRCh38
rs1011115999 Chr.20: 23565051-23565051 on Build GRCh38
rs767796704 Chr.20: 23565025-23565025 on Build GRCh38
rs981091313 Chr.20: 23565020-23565020 on Build GRCh38
rs751945216 Chr.20: 23565017-23565017 on Build GRCh38
rs778278324 Chr.20: 23564914-23564914 on Build GRCh38
rs1033886747 Chr.20: 23565035-23565035 on Build GRCh38
rs773945906 Chr.20: 23564711-23564711 on Build GRCh38
rs773159392 Chr.20: 23564970-23564970 on Build GRCh38
rs967032897 Chr.20: 23565086-23565086 on Build GRCh38
rs1002403191 Chr.20: 23564763-23564763 on Build GRCh38
rs756580593 Chr.20: 23565033-23565033 on Build GRCh38

View Full Product Details