Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23568548-23569047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST9L
Entrez Gene ID
128821
Gene Name
cystatin 9-like
Gene Aliases
CTES7B, bA218C14.1
UniGene
Hs.121554
Gene Chromosome Location
Chr.20: 23564732-23568749 on Build GRCh38
Transcripts
NM_080610.2

SNP Detail

SNP ID Chromosome Location
rs985058801 Chr.20: 23568988-23568988 on Build GRCh38
rs45552736 Chr.20: 23568819-23568819 on Build GRCh38
rs45563834 Chr.20: 23568707-23568707 on Build GRCh38
rs539340608 Chr.20: 23568598-23568598 on Build GRCh38
rs897158608 Chr.20: 23568734-23568734 on Build GRCh38
rs554475499 Chr.20: 23568625-23568625 on Build GRCh38
rs1032984412 Chr.20: 23568710-23568710 on Build GRCh38
rs957450866 Chr.20: 23568714-23568714 on Build GRCh38
rs1010268060 Chr.20: 23568886-23568886 on Build GRCh38
rs557200653 Chr.20: 23568768-23568768 on Build GRCh38
rs148161347 Chr.20: 23568861-23568862 on Build GRCh38
rs146385935 Chr.20: 23568928-23568928 on Build GRCh38
rs1017821226 Chr.20: 23568796-23568796 on Build GRCh38
rs1029978505 Chr.20: 23568824-23568824 on Build GRCh38
rs45455304 Chr.20: 23568931-23568931 on Build GRCh38
rs6083173 Chr.20: 23568579-23568579 on Build GRCh38
rs1048833123 Chr.20: 23568974-23568974 on Build GRCh38
rs528886226 Chr.20: 23568659-23568659 on Build GRCh38
rs953580333 Chr.20: 23568978-23568978 on Build GRCh38
rs569633144 Chr.20: 23568616-23568616 on Build GRCh38
rs866387469 Chr.20: 23568613-23568613 on Build GRCh38
rs747200680 Chr.20: 23568620-23568620 on Build GRCh38
rs978350259 Chr.20: 23568677-23568677 on Build GRCh38
rs759138912 Chr.20: 23569023-23569023 on Build GRCh38
rs543483918 Chr.20: 23568885-23568885 on Build GRCh38
rs577809286 Chr.20: 23568880-23568880 on Build GRCh38
rs553422300 Chr.20: 23568818-23568818 on Build GRCh38
rs761859217 Chr.20: 23568719-23568719 on Build GRCh38
rs537549265 Chr.20: 23568766-23568766 on Build GRCh38
rs896997124 Chr.20: 23568647-23568647 on Build GRCh38
rs916743716 Chr.20: 23568894-23568894 on Build GRCh38
rs1036836767 Chr.20: 23568641-23568641 on Build GRCh38
rs138928344 Chr.20: 23568729-23568729 on Build GRCh38
rs1467503 Chr.20: 23568904-23568904 on Build GRCh38
rs571826759 Chr.20: 23568590-23568590 on Build GRCh38
rs929235292 Chr.20: 23568747-23568747 on Build GRCh38
rs182940532 Chr.20: 23568841-23568841 on Build GRCh38
rs1467502 Chr.20: 23568694-23568694 on Build GRCh38
rs1045578113 Chr.20: 23568831-23568831 on Build GRCh38
rs1029247133 Chr.20: 23568732-23568732 on Build GRCh38
rs916671762 Chr.20: 23568718-23568718 on Build GRCh38
rs563081342 Chr.20: 23568889-23568889 on Build GRCh38
rs918644448 Chr.20: 23568929-23568929 on Build GRCh38
rs373286592 Chr.20: 23568864-23568865 on Build GRCh38
rs901966728 Chr.20: 23568925-23568925 on Build GRCh38
rs762865671 Chr.20: 23568847-23568847 on Build GRCh38
rs77287699 Chr.20: 23568709-23568709 on Build GRCh38
rs941115390 Chr.20: 23568633-23568633 on Build GRCh38
rs964389852 Chr.20: 23568638-23568638 on Build GRCh38
rs953613374 Chr.20: 23568752-23568752 on Build GRCh38
rs149600856 Chr.20: 23568667-23568667 on Build GRCh38
rs191428073 Chr.20: 23568793-23568793 on Build GRCh38
rs112325854 Chr.20: 23568829-23568829 on Build GRCh38
rs142161095 Chr.20: 23568781-23568781 on Build GRCh38
rs966599074 Chr.20: 23568728-23568728 on Build GRCh38
rs772307929 Chr.20: 23568597-23568597 on Build GRCh38
rs1054363646 Chr.20: 23568863-23568863 on Build GRCh38
rs1020392458 Chr.20: 23568725-23568725 on Build GRCh38
rs985090125 Chr.20: 23568757-23568757 on Build GRCh38

View Full Product Details