Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23875851-23876360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST5
Entrez Gene ID
1473
Gene Name
cystatin D
Gene Aliases
-
UniGene
Hs.121489
Gene Chromosome Location
Chr.20: 23875935-23879743 on Build GRCh38
Transcripts
NM_001900.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1410820 c.419G>A p.R140Q Chr.20: 23876198-23876198 on Build GRCh38
7161011 c.369C>T p.I123I Chr.20: 23876248-23876248 on Build GRCh38
7206636 c.428A>G p.*143W Chr.20: 23876189-23876189 on Build GRCh38
5857513 c.346G>A p.E116K Chr.20: 23876271-23876271 on Build GRCh38
4929925 c.419G>T p.R140L Chr.20: 23876198-23876198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776894389 Chr.20: 23876142-23876142 on Build GRCh38
rs915139876 Chr.20: 23876123-23876140 on Build GRCh38
rs773622440 Chr.20: 23876056-23876056 on Build GRCh38
rs777922085 Chr.20: 23876173-23876174 on Build GRCh38
rs748691032 Chr.20: 23875945-23875945 on Build GRCh38
rs1027786762 Chr.20: 23876024-23876024 on Build GRCh38
rs202236710 Chr.20: 23876305-23876305 on Build GRCh38
rs375383971 Chr.20: 23875932-23875932 on Build GRCh38
rs981868740 Chr.20: 23875993-23875993 on Build GRCh38
rs970773587 Chr.20: 23875976-23875976 on Build GRCh38
rs557446943 Chr.20: 23876069-23876069 on Build GRCh38
rs571756149 Chr.20: 23876014-23876014 on Build GRCh38
rs1014790294 Chr.20: 23876253-23876253 on Build GRCh38
rs1043827278 Chr.20: 23876087-23876087 on Build GRCh38
rs755351907 Chr.20: 23876177-23876177 on Build GRCh38
rs752654917 Chr.20: 23876302-23876302 on Build GRCh38
rs747138531 Chr.20: 23876184-23876185 on Build GRCh38
rs149451063 Chr.20: 23876216-23876216 on Build GRCh38
rs564973350 Chr.20: 23876009-23876009 on Build GRCh38
rs997560308 Chr.20: 23875899-23875899 on Build GRCh38
rs759371053 Chr.20: 23876143-23876143 on Build GRCh38
rs902486732 Chr.20: 23876334-23876334 on Build GRCh38
rs767188498 Chr.20: 23876258-23876258 on Build GRCh38
rs771219227 Chr.20: 23876206-23876206 on Build GRCh38
rs532052611 Chr.20: 23876010-23876010 on Build GRCh38
rs745986649 Chr.20: 23876306-23876306 on Build GRCh38
rs551680830 Chr.20: 23875965-23875965 on Build GRCh38
rs999616591 Chr.20: 23875907-23875907 on Build GRCh38
rs780990330 Chr.20: 23876173-23876173 on Build GRCh38
rs970408975 Chr.20: 23876151-23876151 on Build GRCh38
rs372937441 Chr.20: 23876139-23876139 on Build GRCh38
rs933294870 Chr.20: 23876089-23876089 on Build GRCh38
rs1030350379 Chr.20: 23875902-23875902 on Build GRCh38
rs779098813 Chr.20: 23876178-23876178 on Build GRCh38
rs528811528 Chr.20: 23875896-23875896 on Build GRCh38
rs755682280 Chr.20: 23876267-23876267 on Build GRCh38
rs752653730 Chr.20: 23876157-23876157 on Build GRCh38
rs909825220 Chr.20: 23876046-23876046 on Build GRCh38
rs942643592 Chr.20: 23876052-23876052 on Build GRCh38
rs549285474 Chr.20: 23875912-23875912 on Build GRCh38
rs762840649 Chr.20: 23876319-23876319 on Build GRCh38
rs758177825 Chr.20: 23876144-23876144 on Build GRCh38
rs958306305 Chr.20: 23875941-23875941 on Build GRCh38
rs758819974 Chr.20: 23876290-23876290 on Build GRCh38
rs951866828 Chr.20: 23876025-23876025 on Build GRCh38
rs992466517 Chr.20: 23876049-23876049 on Build GRCh38
rs201737711 Chr.20: 23876199-23876199 on Build GRCh38
rs202039216 Chr.20: 23876261-23876261 on Build GRCh38
rs778246010 Chr.20: 23876291-23876291 on Build GRCh38
rs1039987784 Chr.20: 23876068-23876068 on Build GRCh38
rs141858159 Chr.20: 23876196-23876196 on Build GRCh38
rs768680581 Chr.20: 23876282-23876283 on Build GRCh38
rs1044178144 Chr.20: 23875889-23875889 on Build GRCh38
rs925321794 Chr.20: 23876104-23876104 on Build GRCh38
rs745551143 Chr.20: 23876175-23876175 on Build GRCh38
rs907706430 Chr.20: 23876028-23876028 on Build GRCh38
rs548552256 Chr.20: 23876332-23876332 on Build GRCh38
rs1012516673 Chr.20: 23876133-23876133 on Build GRCh38
rs951815028 Chr.20: 23876231-23876231 on Build GRCh38
rs756888210 Chr.20: 23876171-23876171 on Build GRCh38
rs772199049 Chr.20: 23876171-23876173 on Build GRCh38
rs4813509 Chr.20: 23876026-23876026 on Build GRCh38
rs775102122 Chr.20: 23876224-23876225 on Build GRCh38
rs535254884 Chr.20: 23875937-23875937 on Build GRCh38
rs146272783 Chr.20: 23876198-23876198 on Build GRCh38
rs113476538 Chr.20: 23876083-23876083 on Build GRCh38
rs1051826754 Chr.20: 23876107-23876107 on Build GRCh38
rs372319805 Chr.20: 23876329-23876329 on Build GRCh38
rs117440485 Chr.20: 23875914-23875914 on Build GRCh38
rs764156112 Chr.20: 23876167-23876167 on Build GRCh38
rs759724794 Chr.20: 23876203-23876203 on Build GRCh38
rs759629097 Chr.20: 23876125-23876125 on Build GRCh38
rs137939967 Chr.20: 23876204-23876204 on Build GRCh38
rs148371132 Chr.20: 23876263-23876263 on Build GRCh38
rs527503200 Chr.20: 23876082-23876082 on Build GRCh38
rs779763862 Chr.20: 23876268-23876268 on Build GRCh38
rs184494847 Chr.20: 23876116-23876116 on Build GRCh38
rs774588203 Chr.20: 23876123-23876123 on Build GRCh38
rs751043871 Chr.20: 23876169-23876169 on Build GRCh38
rs541266751 Chr.20: 23876200-23876200 on Build GRCh38
rs751616622 Chr.20: 23876250-23876250 on Build GRCh38
rs748576835 Chr.20: 23876185-23876185 on Build GRCh38
rs904034605 Chr.20: 23875904-23875904 on Build GRCh38
rs960642503 Chr.20: 23876043-23876043 on Build GRCh38
rs200427729 Chr.20: 23876252-23876252 on Build GRCh38
rs975372000 Chr.20: 23876240-23876240 on Build GRCh38
rs775104063 Chr.20: 23876214-23876214 on Build GRCh38
rs577725854 Chr.20: 23876241-23876241 on Build GRCh38
rs758216597 Chr.20: 23876166-23876166 on Build GRCh38
rs562953820 Chr.20: 23875890-23875890 on Build GRCh38
rs916604732 Chr.20: 23876064-23876064 on Build GRCh38
rs143411516 Chr.20: 23876107-23876107 on Build GRCh38
rs775791099 Chr.20: 23876307-23876307 on Build GRCh38
rs764980756 Chr.20: 23876153-23876153 on Build GRCh38
rs762893249 Chr.20: 23876278-23876281 on Build GRCh38
rs996210965 Chr.20: 23876206-23876206 on Build GRCh38
rs964710828 Chr.20: 23875957-23875957 on Build GRCh38
rs138639807 Chr.20: 23876192-23876192 on Build GRCh38

View Full Product Details