Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23986159-23986656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGTLC1
Entrez Gene ID
92086
Gene Name
gamma-glutamyltransferase light chain 1
Gene Aliases
GGTL6, GGTLA3, GGTLA4, dJ831C21.1, dJ831C21.2
UniGene
Hs.355394
Gene Chromosome Location
Chr.20: 23985053-23989134 on Build GRCh38
Transcripts
NM_178311.2 NM_178312.2 XM_005260863.4 XM_005260864.4 XM_005260865.4 XM_011529391.2 XM_011529392.2 XM_011529393.2 XM_017028126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
723307 c.156C>A p.A52A Chr.20: 23986456-23986456 on Build GRCh38
6670792 c.27G>A p.Q9Q Chr.20: 23986585-23986585 on Build GRCh38
6670788 c.189G>T p.K63N Chr.20: 23986191-23986191 on Build GRCh38
7229217 c.9C>T p.S3S Chr.20: 23986603-23986603 on Build GRCh38
5391714 c.208G>A p.G70R Chr.20: 23986172-23986172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765540830 Chr.20: 23986236-23986236 on Build GRCh38
rs747138352 Chr.20: 23986238-23986239 on Build GRCh38
rs774297579 Chr.20: 23986632-23986633 on Build GRCh38
rs544003351 Chr.20: 23986247-23986247 on Build GRCh38
rs770572069 Chr.20: 23986432-23986432 on Build GRCh38
rs373992644 Chr.20: 23986215-23986215 on Build GRCh38
rs746531430 Chr.20: 23986486-23986486 on Build GRCh38
rs368480074 Chr.20: 23986616-23986616 on Build GRCh38
rs758367107 Chr.20: 23986482-23986482 on Build GRCh38
rs778306866 Chr.20: 23986632-23986632 on Build GRCh38
rs553725844 Chr.20: 23986617-23986617 on Build GRCh38
rs960712514 Chr.20: 23986440-23986440 on Build GRCh38
rs749051701 Chr.20: 23986394-23986394 on Build GRCh38
rs759614807 Chr.20: 23986588-23986588 on Build GRCh38
rs956715274 Chr.20: 23986332-23986332 on Build GRCh38
rs757239635 Chr.20: 23986622-23986622 on Build GRCh38
rs200907068 Chr.20: 23986231-23986231 on Build GRCh38
rs754771816 Chr.20: 23986469-23986469 on Build GRCh38
rs768627348 Chr.20: 23986593-23986595 on Build GRCh38
rs565707134 Chr.20: 23986513-23986513 on Build GRCh38
rs374954016 Chr.20: 23986554-23986554 on Build GRCh38
rs369309954 Chr.20: 23986187-23986187 on Build GRCh38
rs1030933374 Chr.20: 23986312-23986312 on Build GRCh38
rs749088264 Chr.20: 23986528-23986529 on Build GRCh38
rs755708914 Chr.20: 23986628-23986628 on Build GRCh38
rs777865788 Chr.20: 23986194-23986194 on Build GRCh38
rs753087553 Chr.20: 23986237-23986237 on Build GRCh38
rs773898625 Chr.20: 23986575-23986575 on Build GRCh38
rs377399847 Chr.20: 23986197-23986197 on Build GRCh38
rs768151671 Chr.20: 23986512-23986512 on Build GRCh38
rs368958402 Chr.20: 23986451-23986451 on Build GRCh38
rs759963069 Chr.20: 23986186-23986186 on Build GRCh38
rs565009521 Chr.20: 23986408-23986408 on Build GRCh38
rs748582298 Chr.20: 23986564-23986564 on Build GRCh38
rs781520249 Chr.20: 23986505-23986505 on Build GRCh38
rs534711150 Chr.20: 23986530-23986530 on Build GRCh38
rs180925592 Chr.20: 23986208-23986208 on Build GRCh38
rs771039000 Chr.20: 23986458-23986458 on Build GRCh38
rs753665231 Chr.20: 23986464-23986464 on Build GRCh38
rs376069716 Chr.20: 23986233-23986233 on Build GRCh38
rs770553942 Chr.20: 23986498-23986498 on Build GRCh38
rs377552031 Chr.20: 23986577-23986577 on Build GRCh38
rs780873114 Chr.20: 23986550-23986550 on Build GRCh38
rs752472119 Chr.20: 23986614-23986614 on Build GRCh38
rs771710828 Chr.20: 23986222-23986222 on Build GRCh38
rs150881025 Chr.20: 23986580-23986580 on Build GRCh38
rs776211084 Chr.20: 23986501-23986501 on Build GRCh38
rs542769378 Chr.20: 23986199-23986199 on Build GRCh38
rs1009004265 Chr.20: 23986290-23986290 on Build GRCh38
rs199795480 Chr.20: 23986418-23986418 on Build GRCh38
rs778784113 Chr.20: 23986401-23986401 on Build GRCh38
rs753256821 Chr.20: 23986190-23986190 on Build GRCh38
rs773855088 Chr.20: 23986216-23986216 on Build GRCh38
rs780117483 Chr.20: 23986210-23986210 on Build GRCh38
rs7260970 Chr.20: 23986386-23986386 on Build GRCh38
rs543383346 Chr.20: 23986352-23986352 on Build GRCh38
rs369770828 Chr.20: 23986209-23986209 on Build GRCh38
rs867208162 Chr.20: 23986397-23986397 on Build GRCh38
rs138035106 Chr.20: 23986531-23986531 on Build GRCh38
rs141107815 Chr.20: 23986483-23986483 on Build GRCh38
rs772703971 Chr.20: 23986572-23986572 on Build GRCh38
rs779929104 Chr.20: 23986391-23986391 on Build GRCh38
rs749631518 Chr.20: 23986505-23986505 on Build GRCh38
rs763281453 Chr.20: 23986437-23986437 on Build GRCh38
rs372298499 Chr.20: 23986389-23986389 on Build GRCh38
rs768626540 Chr.20: 23986395-23986395 on Build GRCh38
rs143487091 Chr.20: 23986570-23986570 on Build GRCh38
rs757440132 Chr.20: 23986206-23986206 on Build GRCh38
rs775862580 Chr.20: 23986435-23986435 on Build GRCh38
rs190984751 Chr.20: 23986473-23986473 on Build GRCh38
rs1035568915 Chr.20: 23986590-23986590 on Build GRCh38
rs138247242 Chr.20: 23986603-23986603 on Build GRCh38
rs780526994 Chr.20: 23986625-23986625 on Build GRCh38
rs774521787 Chr.20: 23986563-23986563 on Build GRCh38
rs756797323 Chr.20: 23986549-23986549 on Build GRCh38
rs374913892 Chr.20: 23986180-23986180 on Build GRCh38
rs371694918 Chr.20: 23986458-23986458 on Build GRCh38
rs557323567 Chr.20: 23986280-23986280 on Build GRCh38
rs148565423 Chr.20: 23986615-23986615 on Build GRCh38
rs376723489 Chr.20: 23986571-23986571 on Build GRCh38
rs747666979 Chr.20: 23986406-23986406 on Build GRCh38
rs973758007 Chr.20: 23986619-23986619 on Build GRCh38
rs758854613 Chr.20: 23986191-23986191 on Build GRCh38
rs754058386 Chr.20: 23986521-23986521 on Build GRCh38
rs764655324 Chr.20: 23986447-23986447 on Build GRCh38
rs571715945 Chr.20: 23986540-23986540 on Build GRCh38
rs578083715 Chr.20: 23986232-23986232 on Build GRCh38
rs200608114 Chr.20: 23986631-23986631 on Build GRCh38
rs757346842 Chr.20: 23986249-23986249 on Build GRCh38
rs927717422 Chr.20: 23986438-23986438 on Build GRCh38
rs146964922 Chr.20: 23986510-23986510 on Build GRCh38
rs762369986 Chr.20: 23986511-23986511 on Build GRCh38
rs528122204 Chr.20: 23986371-23986371 on Build GRCh38
rs1011073293 Chr.20: 23986382-23986382 on Build GRCh38
rs761288477 Chr.20: 23986217-23986217 on Build GRCh38
rs148204497 Chr.20: 23986459-23986459 on Build GRCh38
rs572844063 Chr.20: 23986627-23986627 on Build GRCh38
rs750722593 Chr.20: 23986388-23986388 on Build GRCh38
rs755253895 Chr.20: 23986523-23986523 on Build GRCh38
rs1050521274 Chr.20: 23986235-23986235 on Build GRCh38
rs574272015 Chr.20: 23986347-23986347 on Build GRCh38
rs763447103 Chr.20: 23986241-23986241 on Build GRCh38
rs545033230 Chr.20: 23986629-23986629 on Build GRCh38
rs373114346 Chr.20: 23986195-23986195 on Build GRCh38
rs370253448 Chr.20: 23986474-23986474 on Build GRCh38
rs745460259 Chr.20: 23986551-23986551 on Build GRCh38
rs760906005 Chr.20: 23986517-23986517 on Build GRCh38
rs776807381 Chr.20: 23986581-23986581 on Build GRCh38
rs768949759 Chr.20: 23986509-23986509 on Build GRCh38
rs559376634 Chr.20: 23986361-23986361 on Build GRCh38

View Full Product Details