Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 24949139-24949640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST7
Entrez Gene ID
8530
Gene Name
cystatin F
Gene Aliases
CMAP
UniGene
Hs.143212
Gene Chromosome Location
Chr.20: 24949230-24959928 on Build GRCh38
Transcripts
NM_003650.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6636996 c.119C>T p.T40I Chr.20: 24949558-24949558 on Build GRCh38
4675221 c.130_131insC p.D46fs*8 Chr.20: 24949569-24949570 on Build GRCh38
443562 c.24C>T p.H8H Chr.20: 24949463-24949463 on Build GRCh38
1713292 c.128C>T p.P43L Chr.20: 24949567-24949567 on Build GRCh38
2151934 c.17C>A p.A6E Chr.20: 24949456-24949456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770389030 Chr.20: 24949610-24949610 on Build GRCh38
rs549598804 Chr.20: 24949344-24949344 on Build GRCh38
rs761066395 Chr.20: 24949536-24949536 on Build GRCh38
rs753347342 Chr.20: 24949559-24949559 on Build GRCh38
rs771941085 Chr.20: 24949438-24949438 on Build GRCh38
rs758618698 Chr.20: 24949557-24949557 on Build GRCh38
rs368801661 Chr.20: 24949584-24949584 on Build GRCh38
rs1015981013 Chr.20: 24949223-24949223 on Build GRCh38
rs764589940 Chr.20: 24949469-24949469 on Build GRCh38
rs762128527 Chr.20: 24949407-24949407 on Build GRCh38
rs894001550 Chr.20: 24949300-24949300 on Build GRCh38
rs970611969 Chr.20: 24949413-24949413 on Build GRCh38
rs760116553 Chr.20: 24949561-24949561 on Build GRCh38
rs386813541 Chr.20: 24949186-24949198 on Build GRCh38
rs746873725 Chr.20: 24949445-24949445 on Build GRCh38
rs529605946 Chr.20: 24949475-24949475 on Build GRCh38
rs549920012 Chr.20: 24949319-24949319 on Build GRCh38
rs386813542 Chr.20: 24949197-24949198 on Build GRCh38
rs227653 Chr.20: 24949449-24949449 on Build GRCh38
rs967005077 Chr.20: 24949340-24949342 on Build GRCh38
rs775685240 Chr.20: 24949360-24949360 on Build GRCh38
rs897220469 Chr.20: 24949513-24949513 on Build GRCh38
rs1002083702 Chr.20: 24949563-24949563 on Build GRCh38
rs746026113 Chr.20: 24949469-24949469 on Build GRCh38
rs45469992 Chr.20: 24949585-24949585 on Build GRCh38
rs776283035 Chr.20: 24949399-24949399 on Build GRCh38
rs537033491 Chr.20: 24949385-24949385 on Build GRCh38
rs372871405 Chr.20: 24949395-24949395 on Build GRCh38
rs757171720 Chr.20: 24949581-24949581 on Build GRCh38
rs372972699 Chr.20: 24949606-24949606 on Build GRCh38
rs776193957 Chr.20: 24949455-24949455 on Build GRCh38
rs527769166 Chr.20: 24949272-24949272 on Build GRCh38
rs777651450 Chr.20: 24949505-24949505 on Build GRCh38
rs769137909 Chr.20: 24949522-24949522 on Build GRCh38
rs374159769 Chr.20: 24949463-24949463 on Build GRCh38
rs45559431 Chr.20: 24949529-24949529 on Build GRCh38
rs768265509 Chr.20: 24949598-24949598 on Build GRCh38
rs773560833 Chr.20: 24949532-24949532 on Build GRCh38
rs191930787 Chr.20: 24949354-24949354 on Build GRCh38
rs1046433064 Chr.20: 24949241-24949241 on Build GRCh38
rs369375433 Chr.20: 24949607-24949607 on Build GRCh38
rs227652 Chr.20: 24949311-24949311 on Build GRCh38
rs779043652 Chr.20: 24949433-24949433 on Build GRCh38
rs544787291 Chr.20: 24949615-24949615 on Build GRCh38
rs112866181 Chr.20: 24949160-24949160 on Build GRCh38
rs573885996 Chr.20: 24949250-24949250 on Build GRCh38
rs745364309 Chr.20: 24949514-24949514 on Build GRCh38
rs35525649 Chr.20: 24949510-24949510 on Build GRCh38
rs777737633 Chr.20: 24949441-24949441 on Build GRCh38
rs551699172 Chr.20: 24949509-24949509 on Build GRCh38
rs563582293 Chr.20: 24949474-24949474 on Build GRCh38
rs147172253 Chr.20: 24949501-24949501 on Build GRCh38
rs556907580 Chr.20: 24949580-24949580 on Build GRCh38
rs921142048 Chr.20: 24949180-24949180 on Build GRCh38
rs371217620 Chr.20: 24949425-24949425 on Build GRCh38
rs184908378 Chr.20: 24949363-24949363 on Build GRCh38
rs755221453 Chr.20: 24949430-24949430 on Build GRCh38
rs375702917 Chr.20: 24949591-24949591 on Build GRCh38
rs111631119 Chr.20: 24949384-24949384 on Build GRCh38
rs11087488 Chr.20: 24949186-24949186 on Build GRCh38
rs369054305 Chr.20: 24949599-24949599 on Build GRCh38
rs374907780 Chr.20: 24949605-24949605 on Build GRCh38
rs187437242 Chr.20: 24949330-24949330 on Build GRCh38
rs201516661 Chr.20: 24949524-24949524 on Build GRCh38
rs747953071 Chr.20: 24949437-24949437 on Build GRCh38
rs756646597 Chr.20: 24949415-24949415 on Build GRCh38
rs11087489 Chr.20: 24949267-24949267 on Build GRCh38
rs756405817 Chr.20: 24949531-24949531 on Build GRCh38
rs556032750 Chr.20: 24949167-24949167 on Build GRCh38
rs227650 Chr.20: 24949171-24949171 on Build GRCh38
rs45470396 Chr.20: 24949527-24949527 on Build GRCh38
rs750880542 Chr.20: 24949411-24949411 on Build GRCh38
rs570315419 Chr.20: 24949329-24949329 on Build GRCh38
rs772877358 Chr.20: 24949315-24949315 on Build GRCh38
rs980614478 Chr.20: 24949416-24949416 on Build GRCh38
rs770968135 Chr.20: 24949353-24949353 on Build GRCh38
rs577525170 Chr.20: 24949197-24949197 on Build GRCh38
rs745439041 Chr.20: 24949460-24949460 on Build GRCh38
rs1010198424 Chr.20: 24949166-24949166 on Build GRCh38
rs970997439 Chr.20: 24949293-24949293 on Build GRCh38
rs199551132 Chr.20: 24949406-24949406 on Build GRCh38
rs227651 Chr.20: 24949198-24949198 on Build GRCh38
rs1005525452 Chr.20: 24949211-24949211 on Build GRCh38
rs774778228 Chr.20: 24949465-24949465 on Build GRCh38
rs766936580 Chr.20: 24949482-24949482 on Build GRCh38
rs572815945 Chr.20: 24949614-24949614 on Build GRCh38
rs374458409 Chr.20: 24949498-24949498 on Build GRCh38
rs73342985 Chr.20: 24949419-24949419 on Build GRCh38
rs141449868 Chr.20: 24949483-24949483 on Build GRCh38
rs368099459 Chr.20: 24949556-24949556 on Build GRCh38
rs936379209 Chr.20: 24949318-24949318 on Build GRCh38
rs1015423212 Chr.20: 24949593-24949594 on Build GRCh38
rs759174167 Chr.20: 24949400-24949400 on Build GRCh38
rs559169413 Chr.20: 24949271-24949271 on Build GRCh38
rs112245167 Chr.20: 24949204-24949204 on Build GRCh38
rs900861314 Chr.20: 24949517-24949517 on Build GRCh38
rs760706023 Chr.20: 24949347-24949347 on Build GRCh38
rs6138429 Chr.20: 24949512-24949512 on Build GRCh38
rs765158816 Chr.20: 24949564-24949564 on Build GRCh38
rs752833224 Chr.20: 24949574-24949574 on Build GRCh38
rs765379965 Chr.20: 24949497-24949497 on Build GRCh38
rs781255012 Chr.20: 24949461-24949462 on Build GRCh38
rs779375364 Chr.20: 24949589-24949589 on Build GRCh38
rs533301898 Chr.20: 24949464-24949464 on Build GRCh38
rs573876806 Chr.20: 24949401-24949401 on Build GRCh38
rs113405695 Chr.20: 24949178-24949178 on Build GRCh38
rs751843132 Chr.20: 24949472-24949472 on Build GRCh38
rs957774623 Chr.20: 24949423-24949423 on Build GRCh38
rs11087490 Chr.20: 24949277-24949277 on Build GRCh38
rs983975884 Chr.20: 24949365-24949365 on Build GRCh38
rs369490177 Chr.20: 24949410-24949410 on Build GRCh38
rs111568714 Chr.20: 24949275-24949275 on Build GRCh38
rs915335692 Chr.20: 24949608-24949608 on Build GRCh38
rs557840909 Chr.20: 24949364-24949364 on Build GRCh38
rs946827063 Chr.20: 24949617-24949617 on Build GRCh38
rs543780401 Chr.20: 24949468-24949468 on Build GRCh38
rs755061024 Chr.20: 24949491-24949491 on Build GRCh38
rs758677555 Chr.20: 24949576-24949576 on Build GRCh38
rs372312352 Chr.20: 24949582-24949582 on Build GRCh38
rs761113696 Chr.20: 24949481-24949481 on Build GRCh38

View Full Product Details