Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 24959320-24959829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST7
Entrez Gene ID
8530
Gene Name
cystatin F
Gene Aliases
CMAP
UniGene
Hs.143212
Gene Chromosome Location
Chr.20: 24949230-24959928 on Build GRCh38
Transcripts
NM_003650.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4545488 c.445G>A p.E149K Chr.20: 24959653-24959653 on Build GRCh38
5757877 c.437G>T p.C146F Chr.20: 24959645-24959645 on Build GRCh38
122397 c.478G>A p.E160K Chr.20: 24959686-24959686 on Build GRCh38
4406418 c.482T>G p.V161G Chr.20: 24959690-24959690 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs963044272 Chr.20: 24959430-24959430 on Build GRCh38
rs368920318 Chr.20: 24959685-24959685 on Build GRCh38
rs139792455 Chr.20: 24959667-24959667 on Build GRCh38
rs772190537 Chr.20: 24959653-24959653 on Build GRCh38
rs769494710 Chr.20: 24959735-24959735 on Build GRCh38
rs955741603 Chr.20: 24959709-24959709 on Build GRCh38
rs767948940 Chr.20: 24959479-24959479 on Build GRCh38
rs775523909 Chr.20: 24959371-24959371 on Build GRCh38
rs1037063788 Chr.20: 24959422-24959422 on Build GRCh38
rs745454606 Chr.20: 24959733-24959733 on Build GRCh38
rs746556910 Chr.20: 24959660-24959660 on Build GRCh38
rs113475360 Chr.20: 24959781-24959781 on Build GRCh38
rs764506777 Chr.20: 24959651-24959652 on Build GRCh38
rs937827710 Chr.20: 24959486-24959486 on Build GRCh38
rs1465 Chr.20: 24959789-24959789 on Build GRCh38
rs1007032853 Chr.20: 24959601-24959601 on Build GRCh38
rs754683459 Chr.20: 24959629-24959629 on Build GRCh38
rs374641152 Chr.20: 24959591-24959591 on Build GRCh38
rs932498272 Chr.20: 24959552-24959552 on Build GRCh38
rs1012875921 Chr.20: 24959439-24959439 on Build GRCh38
rs922388306 Chr.20: 24959541-24959541 on Build GRCh38
rs764300309 Chr.20: 24959676-24959676 on Build GRCh38
rs1034877347 Chr.20: 24959770-24959770 on Build GRCh38
rs11906996 Chr.20: 24959355-24959355 on Build GRCh38
rs897173361 Chr.20: 24959575-24959575 on Build GRCh38
rs927830813 Chr.20: 24959485-24959485 on Build GRCh38
rs762360951 Chr.20: 24959474-24959474 on Build GRCh38
rs150607778 Chr.20: 24959665-24959665 on Build GRCh38
rs546844788 Chr.20: 24959590-24959590 on Build GRCh38
rs1022502202 Chr.20: 24959440-24959440 on Build GRCh38
rs762295495 Chr.20: 24959682-24959682 on Build GRCh38
rs1035570521 Chr.20: 24959431-24959431 on Build GRCh38
rs76869474 Chr.20: 24959569-24959569 on Build GRCh38
rs773663272 Chr.20: 24959760-24959760 on Build GRCh38
rs774712833 Chr.20: 24959680-24959680 on Build GRCh38
rs113685329 Chr.20: 24959461-24959461 on Build GRCh38
rs140096851 Chr.20: 24959717-24959717 on Build GRCh38
rs147518800 Chr.20: 24959394-24959394 on Build GRCh38
rs369649161 Chr.20: 24959736-24959736 on Build GRCh38
rs545492905 Chr.20: 24959427-24959427 on Build GRCh38
rs766505972 Chr.20: 24959689-24959689 on Build GRCh38
rs982018722 Chr.20: 24959754-24959754 on Build GRCh38
rs761277730 Chr.20: 24959762-24959762 on Build GRCh38
rs1043046760 Chr.20: 24959354-24959354 on Build GRCh38
rs1042019511 Chr.20: 24959549-24959549 on Build GRCh38
rs187642533 Chr.20: 24959574-24959574 on Build GRCh38
rs1029877369 Chr.20: 24959379-24959379 on Build GRCh38
rs752162987 Chr.20: 24959699-24959705 on Build GRCh38
rs537584808 Chr.20: 24959719-24959719 on Build GRCh38
rs972647011 Chr.20: 24959783-24959783 on Build GRCh38
rs772840829 Chr.20: 24959659-24959659 on Build GRCh38
rs367899041 Chr.20: 24959718-24959718 on Build GRCh38
rs990548671 Chr.20: 24959497-24959497 on Build GRCh38
rs941334384 Chr.20: 24959404-24959404 on Build GRCh38
rs759262577 Chr.20: 24959668-24959668 on Build GRCh38
rs758378822 Chr.20: 24959712-24959712 on Build GRCh38
rs972724614 Chr.20: 24959436-24959436 on Build GRCh38
rs866550522 Chr.20: 24959390-24959390 on Build GRCh38
rs753938790 Chr.20: 24959691-24959691 on Build GRCh38
rs946384632 Chr.20: 24959508-24959508 on Build GRCh38
rs1012411538 Chr.20: 24959618-24959618 on Build GRCh38
rs755239440 Chr.20: 24959695-24959695 on Build GRCh38
rs200090392 Chr.20: 24959632-24959632 on Build GRCh38
rs139175095 Chr.20: 24959384-24959389 on Build GRCh38
rs748018376 Chr.20: 24959711-24959711 on Build GRCh38
rs941282036 Chr.20: 24959572-24959572 on Build GRCh38
rs756200346 Chr.20: 24959688-24959688 on Build GRCh38
rs955952055 Chr.20: 24959475-24959475 on Build GRCh38
rs774690204 Chr.20: 24959441-24959441 on Build GRCh38
rs753423813 Chr.20: 24959702-24959702 on Build GRCh38
rs373615005 Chr.20: 24959578-24959578 on Build GRCh38
rs775814830 Chr.20: 24959613-24959613 on Build GRCh38
rs750559979 Chr.20: 24959587-24959587 on Build GRCh38
rs914969919 Chr.20: 24959507-24959507 on Build GRCh38
rs998766092 Chr.20: 24959368-24959368 on Build GRCh38
rs778852668 Chr.20: 24959634-24959634 on Build GRCh38
rs959213261 Chr.20: 24959772-24959772 on Build GRCh38
rs902176805 Chr.20: 24959561-24959561 on Build GRCh38
rs772549635 Chr.20: 24959756-24959756 on Build GRCh38
rs777906860 Chr.20: 24959713-24959713 on Build GRCh38
rs748038423 Chr.20: 24959648-24959648 on Build GRCh38
rs6050210 Chr.20: 24959613-24959613 on Build GRCh38
rs1007000298 Chr.20: 24959400-24959400 on Build GRCh38
rs1031315113 Chr.20: 24959469-24959469 on Build GRCh38
rs763418462 Chr.20: 24959623-24959624 on Build GRCh38
rs762209764 Chr.20: 24959739-24959739 on Build GRCh38
rs149366656 Chr.20: 24959686-24959686 on Build GRCh38

View Full Product Details