Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31377081-31377554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB119
Entrez Gene ID
245932
Gene Name
defensin beta 119
Gene Aliases
DEFB-19, DEFB-20, DEFB120, DEFB20, ESC42-RELA, ESC42-RELB
UniGene
Hs.516998
Gene Chromosome Location
Chr.20: 31377163-31390649 on Build GRCh38
Transcripts
NM_153289.3 NR_073151.1 NR_073152.1 NR_073153.1 NR_126440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
396499 c.145T>A p.C49S Chr.20: 31377356-31377356 on Build GRCh38
244006 c.107C>T p.T36M Chr.20: 31377434-31377434 on Build GRCh38
274767 c.108G>A p.T36T Chr.20: 31377433-31377433 on Build GRCh38
2888352 c.127G>T p.G43W Chr.20: 31377414-31377414 on Build GRCh38
4097384 c.253T>C p.*85R Chr.20: 31377248-31377248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777189261 Chr.20: 31377205-31377205 on Build GRCh38
rs780519590 Chr.20: 31377323-31377323 on Build GRCh38
rs753444973 Chr.20: 31377299-31377299 on Build GRCh38
rs11558232 Chr.20: 31377314-31377314 on Build GRCh38
rs2272948 Chr.20: 31377132-31377132 on Build GRCh38
rs373413869 Chr.20: 31377364-31377364 on Build GRCh38
rs145112384 Chr.20: 31377421-31377421 on Build GRCh38
rs750803651 Chr.20: 31377382-31377382 on Build GRCh38
rs1005468491 Chr.20: 31377301-31377301 on Build GRCh38
rs768916240 Chr.20: 31377249-31377249 on Build GRCh38
rs112690953 Chr.20: 31377375-31377375 on Build GRCh38
rs141914877 Chr.20: 31377174-31377174 on Build GRCh38
rs752398180 Chr.20: 31377369-31377369 on Build GRCh38
rs1035844965 Chr.20: 31377168-31377168 on Build GRCh38
rs1042664303 Chr.20: 31377184-31377185 on Build GRCh38
rs987435855 Chr.20: 31377519-31377519 on Build GRCh38
rs775011756 Chr.20: 31377374-31377374 on Build GRCh38
rs776721266 Chr.20: 31377438-31377438 on Build GRCh38
rs144077858 Chr.20: 31377393-31377393 on Build GRCh38
rs991584134 Chr.20: 31377362-31377362 on Build GRCh38
rs766006707 Chr.20: 31377284-31377284 on Build GRCh38
rs763186753 Chr.20: 31377241-31377241 on Build GRCh38
rs748421133 Chr.20: 31377419-31377419 on Build GRCh38
rs777745627 Chr.20: 31377358-31377359 on Build GRCh38
rs774696849 Chr.20: 31377333-31377333 on Build GRCh38
rs764833777 Chr.20: 31377462-31377462 on Build GRCh38
rs746302593 Chr.20: 31377231-31377231 on Build GRCh38
rs772830343 Chr.20: 31377358-31377358 on Build GRCh38
rs183941727 Chr.20: 31377105-31377105 on Build GRCh38
rs141841649 Chr.20: 31377251-31377251 on Build GRCh38
rs201526799 Chr.20: 31377366-31377366 on Build GRCh38
rs138820976 Chr.20: 31377111-31377111 on Build GRCh38
rs763696202 Chr.20: 31377481-31377481 on Build GRCh38
rs760492914 Chr.20: 31377361-31377361 on Build GRCh38
rs745424687 Chr.20: 31377475-31377475 on Build GRCh38
rs709045 Chr.20: 31377211-31377211 on Build GRCh38
rs6120000 Chr.20: 31377203-31377203 on Build GRCh38
rs780746364 Chr.20: 31377450-31377450 on Build GRCh38
rs761887399 Chr.20: 31377334-31377334 on Build GRCh38
rs1037337620 Chr.20: 31377529-31377529 on Build GRCh38
rs749785591 Chr.20: 31377405-31377405 on Build GRCh38
rs186901050 Chr.20: 31377418-31377418 on Build GRCh38
rs867079819 Chr.20: 31377325-31377325 on Build GRCh38
rs950512580 Chr.20: 31377152-31377152 on Build GRCh38
rs376756000 Chr.20: 31377195-31377195 on Build GRCh38
rs753802251 Chr.20: 31377526-31377526 on Build GRCh38
rs750391797 Chr.20: 31377273-31377273 on Build GRCh38
rs139822193 Chr.20: 31377318-31377318 on Build GRCh38
rs972550060 Chr.20: 31377110-31377110 on Build GRCh38
rs762503511 Chr.20: 31377470-31377470 on Build GRCh38
rs143695145 Chr.20: 31377320-31377320 on Build GRCh38
rs781751467 Chr.20: 31377212-31377212 on Build GRCh38
rs982257707 Chr.20: 31377162-31377162 on Build GRCh38
rs768075616 Chr.20: 31377133-31377133 on Build GRCh38
rs111917092 Chr.20: 31377316-31377316 on Build GRCh38
rs550542493 Chr.20: 31377265-31377265 on Build GRCh38
rs769607917 Chr.20: 31377502-31377502 on Build GRCh38
rs924593833 Chr.20: 31377274-31377274 on Build GRCh38
rs749413522 Chr.20: 31377329-31377329 on Build GRCh38
rs115355226 Chr.20: 31377185-31377185 on Build GRCh38
rs372151570 Chr.20: 31377240-31377240 on Build GRCh38
rs781343809 Chr.20: 31377319-31377319 on Build GRCh38
rs759478957 Chr.20: 31377442-31377442 on Build GRCh38
rs530788920 Chr.20: 31377236-31377236 on Build GRCh38
rs934652087 Chr.20: 31377503-31377503 on Build GRCh38
rs773450020 Chr.20: 31377422-31377422 on Build GRCh38
rs888323487 Chr.20: 31377285-31377285 on Build GRCh38
rs146735308 Chr.20: 31377321-31377321 on Build GRCh38
rs916164489 Chr.20: 31377193-31377193 on Build GRCh38
rs780607634 Chr.20: 31377399-31377399 on Build GRCh38
rs150604485 Chr.20: 31377307-31377307 on Build GRCh38
rs760485723 Chr.20: 31377278-31377278 on Build GRCh38
rs3817876 Chr.20: 31377464-31377464 on Build GRCh38
rs759173432 Chr.20: 31377365-31377365 on Build GRCh38
rs757316158 Chr.20: 31377449-31377450 on Build GRCh38
rs768721823 Chr.20: 31377332-31377332 on Build GRCh38
rs374993061 Chr.20: 31377405-31377408 on Build GRCh38
rs755066623 Chr.20: 31377409-31377409 on Build GRCh38
rs947062821 Chr.20: 31377163-31377163 on Build GRCh38
rs767207708 Chr.20: 31377266-31377266 on Build GRCh38
rs150495779 Chr.20: 31377434-31377434 on Build GRCh38
rs567485409 Chr.20: 31377303-31377303 on Build GRCh38
rs771928483 Chr.20: 31377337-31377337 on Build GRCh38
rs991582360 Chr.20: 31377192-31377192 on Build GRCh38
rs746865254 Chr.20: 31377433-31377433 on Build GRCh38

View Full Product Details