Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31405986-31406529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB121
Entrez Gene ID
245934
Gene Name
defensin beta 121
Gene Aliases
DEFB21, ESC42RELC
UniGene
Hs.272294
Gene Chromosome Location
Chr.20: 31404845-31418522 on Build GRCh38
Transcripts
NM_001011878.2 NM_001171832.1 XM_005260383.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2888384 c.56C>T p.P19L Chr.20: 31406097-31406097 on Build GRCh38
5529665 c.40C>T p.L14L Chr.20: 31406113-31406113 on Build GRCh38
5544816 c.58G>A p.V20I Chr.20: 31406095-31406095 on Build GRCh38
2888387 c.10C>T p.L4F Chr.20: 31406143-31406143 on Build GRCh38
334321 c.55C>A p.P19T Chr.20: 31406098-31406098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772018861 Chr.20: 31406114-31406114 on Build GRCh38
rs894904071 Chr.20: 31406423-31406423 on Build GRCh38
rs556293260 Chr.20: 31406170-31406170 on Build GRCh38
rs542220494 Chr.20: 31406475-31406475 on Build GRCh38
rs767897758 Chr.20: 31406157-31406157 on Build GRCh38
rs576415213 Chr.20: 31406427-31406427 on Build GRCh38
rs554485515 Chr.20: 31406300-31406300 on Build GRCh38
rs956061271 Chr.20: 31406314-31406314 on Build GRCh38
rs144190719 Chr.20: 31406287-31406287 on Build GRCh38
rs146924056 Chr.20: 31406146-31406146 on Build GRCh38
rs755068078 Chr.20: 31406109-31406109 on Build GRCh38
rs562021533 Chr.20: 31406055-31406055 on Build GRCh38
rs761002992 Chr.20: 31406164-31406164 on Build GRCh38
rs767811447 Chr.20: 31406099-31406099 on Build GRCh38
rs899662631 Chr.20: 31406421-31406421 on Build GRCh38
rs546347831 Chr.20: 31406019-31406019 on Build GRCh38
rs979187634 Chr.20: 31406108-31406108 on Build GRCh38
rs1048119457 Chr.20: 31406473-31406473 on Build GRCh38
rs756572750 Chr.20: 31406101-31406101 on Build GRCh38
rs368566015 Chr.20: 31406194-31406194 on Build GRCh38
rs771930758 Chr.20: 31406047-31406047 on Build GRCh38
rs371119986 Chr.20: 31406389-31406389 on Build GRCh38
rs916741314 Chr.20: 31406279-31406279 on Build GRCh38
rs752896294 Chr.20: 31406190-31406190 on Build GRCh38
rs569647064 Chr.20: 31406163-31406163 on Build GRCh38
rs753970007 Chr.20: 31406136-31406138 on Build GRCh38
rs961141887 Chr.20: 31406222-31406222 on Build GRCh38
rs894859100 Chr.20: 31406283-31406283 on Build GRCh38
rs113870794 Chr.20: 31406370-31406370 on Build GRCh38
rs969419797 Chr.20: 31406286-31406286 on Build GRCh38
rs1003159251 Chr.20: 31406275-31406275 on Build GRCh38
rs776322166 Chr.20: 31406063-31406063 on Build GRCh38
rs773104983 Chr.20: 31406117-31406117 on Build GRCh38
rs546411134 Chr.20: 31406353-31406353 on Build GRCh38
rs755276011 Chr.20: 31406200-31406203 on Build GRCh38
rs1034713013 Chr.20: 31406034-31406034 on Build GRCh38
rs939027459 Chr.20: 31406416-31406416 on Build GRCh38
rs556668311 Chr.20: 31406359-31406359 on Build GRCh38
rs534468984 Chr.20: 31406271-31406271 on Build GRCh38
rs890928371 Chr.20: 31406031-31406031 on Build GRCh38
rs371955397 Chr.20: 31406156-31406156 on Build GRCh38
rs1039501349 Chr.20: 31406400-31406400 on Build GRCh38
rs531773351 Chr.20: 31406027-31406027 on Build GRCh38
rs781668247 Chr.20: 31406018-31406018 on Build GRCh38
rs756494994 Chr.20: 31406120-31406120 on Build GRCh38
rs1037232158 Chr.20: 31406228-31406228 on Build GRCh38
rs746773177 Chr.20: 31406121-31406121 on Build GRCh38
rs532398104 Chr.20: 31406358-31406358 on Build GRCh38
rs527553747 Chr.20: 31406078-31406078 on Build GRCh38
rs1003275810 Chr.20: 31406029-31406029 on Build GRCh38
rs912454789 Chr.20: 31406376-31406376 on Build GRCh38
rs149744379 Chr.20: 31406103-31406103 on Build GRCh38
rs868427094 Chr.20: 31406175-31406175 on Build GRCh38
rs368207317 Chr.20: 31406155-31406155 on Build GRCh38
rs74647926 Chr.20: 31406327-31406327 on Build GRCh38
rs903726590 Chr.20: 31406453-31406453 on Build GRCh38
rs1030921473 Chr.20: 31406302-31406302 on Build GRCh38
rs995334038 Chr.20: 31406435-31406435 on Build GRCh38
rs541858498 Chr.20: 31406051-31406051 on Build GRCh38
rs145474947 Chr.20: 31406110-31406110 on Build GRCh38
rs370680644 Chr.20: 31406132-31406132 on Build GRCh38
rs769282347 Chr.20: 31406152-31406152 on Build GRCh38
rs374410645 Chr.20: 31406096-31406096 on Build GRCh38
rs746532576 Chr.20: 31406402-31406402 on Build GRCh38
rs149044865 Chr.20: 31406195-31406195 on Build GRCh38
rs979507884 Chr.20: 31406289-31406289 on Build GRCh38
rs1037284434 Chr.20: 31406371-31406371 on Build GRCh38
rs986046231 Chr.20: 31406360-31406360 on Build GRCh38
rs543130516 Chr.20: 31406154-31406154 on Build GRCh38
rs144922209 Chr.20: 31406088-31406088 on Build GRCh38
rs750747672 Chr.20: 31406020-31406020 on Build GRCh38
rs754360458 Chr.20: 31406183-31406183 on Build GRCh38
rs570338026 Chr.20: 31406107-31406107 on Build GRCh38
rs764857461 Chr.20: 31406086-31406086 on Build GRCh38
rs965913920 Chr.20: 31406476-31406476 on Build GRCh38
rs779005447 Chr.20: 31406187-31406187 on Build GRCh38
rs758700043 Chr.20: 31406485-31406485 on Build GRCh38
rs978028827 Chr.20: 31406297-31406297 on Build GRCh38
rs568750645 Chr.20: 31406264-31406264 on Build GRCh38
rs532943201 Chr.20: 31406113-31406113 on Build GRCh38
rs1003811027 Chr.20: 31406481-31406481 on Build GRCh38
rs1029635479 Chr.20: 31406490-31406490 on Build GRCh38
rs766489590 Chr.20: 31406093-31406102 on Build GRCh38
rs896849283 Chr.20: 31406250-31406250 on Build GRCh38
rs992453511 Chr.20: 31406266-31406266 on Build GRCh38
rs562826540 Chr.20: 31406021-31406021 on Build GRCh38
rs1015111227 Chr.20: 31406197-31406197 on Build GRCh38
rs999352833 Chr.20: 31406456-31406456 on Build GRCh38
rs1043850638 Chr.20: 31406429-31406429 on Build GRCh38
rs141107623 Chr.20: 31406134-31406134 on Build GRCh38
rs755013847 Chr.20: 31406184-31406184 on Build GRCh38
rs778515730 Chr.20: 31406217-31406217 on Build GRCh38
rs111257128 Chr.20: 31406009-31406009 on Build GRCh38

View Full Product Details