Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31861357-31861848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP15
Entrez Gene ID
128853
Gene Name
dual specificity phosphatase 15
Gene Aliases
C20orf57, VHY
UniGene
Hs.434108
Gene Chromosome Location
Chr.20: 31845578-31870744 on Build GRCh38
Transcripts
NM_001012644.2 NM_001320478.1 NM_001320479.1 NM_080611.4 NM_177991.2 XM_017027654.1 XM_017027655.1 XM_017027656.1 XM_017027657.1 XM_017027658.1 XM_017027659.1 XM_017027660.1 XM_017027661.1 XM_017027662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6359679 c.658C>A p.Q220K Chr.20: 31861453-31861453 on Build GRCh38
4803616 c.467A>G p.E156G Chr.20: 31861644-31861644 on Build GRCh38
6388571 c.569C>G p.S190* Chr.20: 31861542-31861542 on Build GRCh38
4803537 c.466G>T p.E156* Chr.20: 31861645-31861645 on Build GRCh38
6433624 c.599T>C p.L200P Chr.20: 31861512-31861512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754536353 Chr.20: 31861519-31861519 on Build GRCh38
rs769971947 Chr.20: 31861548-31861548 on Build GRCh38
rs781549982 Chr.20: 31861540-31861540 on Build GRCh38
rs1014815851 Chr.20: 31861500-31861500 on Build GRCh38
rs947310 Chr.20: 31861522-31861522 on Build GRCh38
rs779252037 Chr.20: 31861382-31861382 on Build GRCh38
rs397866092 Chr.20: 31861714-31861715 on Build GRCh38
rs865791106 Chr.20: 31861494-31861494 on Build GRCh38
rs932516621 Chr.20: 31861590-31861590 on Build GRCh38
rs752601342 Chr.20: 31861414-31861414 on Build GRCh38
rs895851650 Chr.20: 31861599-31861599 on Build GRCh38
rs1033231092 Chr.20: 31861630-31861630 on Build GRCh38
rs749867046 Chr.20: 31861448-31861448 on Build GRCh38
rs763978815 Chr.20: 31861419-31861419 on Build GRCh38
rs751494522 Chr.20: 31861421-31861421 on Build GRCh38
rs760705192 Chr.20: 31861394-31861394 on Build GRCh38
rs927994419 Chr.20: 31861603-31861603 on Build GRCh38
rs749993547 Chr.20: 31861510-31861510 on Build GRCh38
rs762891229 Chr.20: 31861498-31861498 on Build GRCh38
rs749554204 Chr.20: 31861574-31861574 on Build GRCh38
rs774207455 Chr.20: 31861578-31861578 on Build GRCh38
rs376544682 Chr.20: 31861607-31861607 on Build GRCh38
rs754844364 Chr.20: 31861614-31861614 on Build GRCh38
rs780784853 Chr.20: 31861439-31861439 on Build GRCh38
rs755615825 Chr.20: 31861514-31861514 on Build GRCh38
rs773098660 Chr.20: 31861601-31861601 on Build GRCh38
rs921036871 Chr.20: 31861651-31861651 on Build GRCh38
rs772625177 Chr.20: 31861388-31861388 on Build GRCh38
rs917252379 Chr.20: 31861766-31861766 on Build GRCh38
rs535044751 Chr.20: 31861824-31861824 on Build GRCh38
rs772390782 Chr.20: 31861467-31861467 on Build GRCh38
rs565382099 Chr.20: 31861656-31861656 on Build GRCh38
rs979031205 Chr.20: 31861508-31861508 on Build GRCh38
rs762838662 Chr.20: 31861418-31861418 on Build GRCh38
rs950135938 Chr.20: 31861777-31861777 on Build GRCh38
rs761499083 Chr.20: 31861592-31861592 on Build GRCh38
rs939997256 Chr.20: 31861593-31861593 on Build GRCh38
rs764786243 Chr.20: 31861616-31861616 on Build GRCh38
rs766056551 Chr.20: 31861515-31861515 on Build GRCh38
rs756700246 Chr.20: 31861723-31861723 on Build GRCh38
rs759679396 Chr.20: 31861484-31861484 on Build GRCh38
rs894019363 Chr.20: 31861629-31861629 on Build GRCh38
rs983319085 Chr.20: 31861774-31861774 on Build GRCh38
rs570843290 Chr.20: 31861628-31861628 on Build GRCh38
rs1006075110 Chr.20: 31861704-31861704 on Build GRCh38
rs1002388762 Chr.20: 31861504-31861504 on Build GRCh38
rs767556843 Chr.20: 31861595-31861595 on Build GRCh38
rs776179652 Chr.20: 31861668-31861668 on Build GRCh38
rs748575175 Chr.20: 31861460-31861460 on Build GRCh38
rs758968419 Chr.20: 31861676-31861676 on Build GRCh38
rs1050849171 Chr.20: 31861568-31861568 on Build GRCh38
rs921098043 Chr.20: 31861566-31861566 on Build GRCh38
rs971238542 Chr.20: 31861801-31861801 on Build GRCh38
rs1036209452 Chr.20: 31861726-31861726 on Build GRCh38
rs572500091 Chr.20: 31861644-31861644 on Build GRCh38
rs897765407 Chr.20: 31861753-31861753 on Build GRCh38
rs983933223 Chr.20: 31861658-31861658 on Build GRCh38
rs566167449 Chr.20: 31861406-31861406 on Build GRCh38
rs752270638 Chr.20: 31861711-31861711 on Build GRCh38
rs768692357 Chr.20: 31861576-31861576 on Build GRCh38
rs1036960970 Chr.20: 31861748-31861748 on Build GRCh38
rs906808639 Chr.20: 31861499-31861499 on Build GRCh38
rs768026860 Chr.20: 31861721-31861721 on Build GRCh38
rs1033283121 Chr.20: 31861513-31861513 on Build GRCh38
rs779667073 Chr.20: 31861455-31861455 on Build GRCh38
rs745985502 Chr.20: 31861546-31861546 on Build GRCh38
rs987816979 Chr.20: 31861533-31861533 on Build GRCh38
rs775824817 Chr.20: 31861497-31861497 on Build GRCh38
rs764985414 Chr.20: 31861690-31861690 on Build GRCh38
rs765501755 Chr.20: 31861411-31861411 on Build GRCh38
rs750003657 Chr.20: 31861491-31861492 on Build GRCh38
rs748014717 Chr.20: 31861648-31861648 on Build GRCh38
rs11417441 Chr.20: 31861713-31861714 on Build GRCh38
rs974687854 Chr.20: 31861561-31861561 on Build GRCh38
rs755633909 Chr.20: 31861505-31861521 on Build GRCh38
rs373217307 Chr.20: 31861473-31861473 on Build GRCh38
rs778189334 Chr.20: 31861469-31861469 on Build GRCh38
rs930451220 Chr.20: 31861432-31861432 on Build GRCh38
rs181315795 Chr.20: 31861550-31861550 on Build GRCh38
rs1046413144 Chr.20: 31861597-31861597 on Build GRCh38
rs555017617 Chr.20: 31861505-31861505 on Build GRCh38
rs561330207 Chr.20: 31861645-31861645 on Build GRCh38
rs770211969 Chr.20: 31861496-31861496 on Build GRCh38
rs776788891 Chr.20: 31861483-31861483 on Build GRCh38
rs184251613 Chr.20: 31861617-31861617 on Build GRCh38
rs780559881 Chr.20: 31861491-31861496 on Build GRCh38
rs773978994 Chr.20: 31861506-31861506 on Build GRCh38
rs754676386 Chr.20: 31861613-31861613 on Build GRCh38
rs771018692 Chr.20: 31861400-31861400 on Build GRCh38
rs113505727 Chr.20: 31861380-31861380 on Build GRCh38
rs776536595 Chr.20: 31861404-31861404 on Build GRCh38
rs770784968 Chr.20: 31861657-31861657 on Build GRCh38
rs767297812 Chr.20: 31861507-31861507 on Build GRCh38
rs554191408 Chr.20: 31861637-31861637 on Build GRCh38
rs779613304 Chr.20: 31861619-31861621 on Build GRCh38
rs757254167 Chr.20: 31861476-31861477 on Build GRCh38
rs538005254 Chr.20: 31861531-31861531 on Build GRCh38
rs531718110 Chr.20: 31861646-31861646 on Build GRCh38
rs1027576628 Chr.20: 31861696-31861696 on Build GRCh38
rs373570132 Chr.20: 31861523-31861523 on Build GRCh38
rs12569 Chr.20: 31861378-31861378 on Build GRCh38
rs200474617 Chr.20: 31861431-31861431 on Build GRCh38
rs943803699 Chr.20: 31861555-31861555 on Build GRCh38
rs376427119 Chr.20: 31861495-31861495 on Build GRCh38
rs755777151 Chr.20: 31861452-31861452 on Build GRCh38
rs1048824993 Chr.20: 31861438-31861438 on Build GRCh38
rs747571045 Chr.20: 31861471-31861471 on Build GRCh38
rs1044470090 Chr.20: 31861786-31861786 on Build GRCh38
rs753462121 Chr.20: 31861516-31861516 on Build GRCh38
rs543995606 Chr.20: 31861653-31861653 on Build GRCh38
rs748454392 Chr.20: 31861385-31861385 on Build GRCh38
rs1048281149 Chr.20: 31861591-31861591 on Build GRCh38
rs139262048 Chr.20: 31861750-31861750 on Build GRCh38
rs762618150 Chr.20: 31861719-31861719 on Build GRCh38
rs760222579 Chr.20: 31861604-31861604 on Build GRCh38
rs547805138 Chr.20: 31861756-31861756 on Build GRCh38
rs976956046 Chr.20: 31861565-31861565 on Build GRCh38
rs774432581 Chr.20: 31861647-31861647 on Build GRCh38

View Full Product Details