Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32446125-32446618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL4L
Entrez Gene ID
140688
Gene Name
nucleolar protein 4 like
Gene Aliases
C20orf112, C20orf113, dJ1184F4.2, dJ1184F4.4
UniGene
Hs.516978
Gene Chromosome Location
Chr.20: 32443059-32585073 on Build GRCh38
Transcripts
NM_001256798.1 NM_080616.4

SNP Detail

SNP ID Chromosome Location
rs528723994 Chr.20: 32446306-32446306 on Build GRCh38
rs886626218 Chr.20: 32446217-32446217 on Build GRCh38
rs1024096745 Chr.20: 32446153-32446153 on Build GRCh38
rs992548502 Chr.20: 32446314-32446314 on Build GRCh38
rs1027789667 Chr.20: 32446173-32446173 on Build GRCh38
rs1001130979 Chr.20: 32446436-32446436 on Build GRCh38
rs930554423 Chr.20: 32446331-32446331 on Build GRCh38
rs762456397 Chr.20: 32446489-32446489 on Build GRCh38
rs574826666 Chr.20: 32446228-32446229 on Build GRCh38
rs552872258 Chr.20: 32446575-32446575 on Build GRCh38
rs1014412221 Chr.20: 32446151-32446152 on Build GRCh38
rs987959600 Chr.20: 32446253-32446253 on Build GRCh38
rs550828928 Chr.20: 32446424-32446424 on Build GRCh38
rs989571317 Chr.20: 32446232-32446233 on Build GRCh38
rs1001183333 Chr.20: 32446182-32446182 on Build GRCh38
rs1021550115 Chr.20: 32446236-32446236 on Build GRCh38
rs1033861424 Chr.20: 32446227-32446234 on Build GRCh38
rs912012712 Chr.20: 32446453-32446453 on Build GRCh38
rs372923363 Chr.20: 32446229-32446230 on Build GRCh38
rs762554983 Chr.20: 32446475-32446475 on Build GRCh38
rs775030746 Chr.20: 32446340-32446340 on Build GRCh38
rs1039013709 Chr.20: 32446479-32446479 on Build GRCh38
rs751461839 Chr.20: 32446537-32446537 on Build GRCh38
rs1054169643 Chr.20: 32446598-32446598 on Build GRCh38
rs775133526 Chr.20: 32446474-32446474 on Build GRCh38
rs562432063 Chr.20: 32446322-32446322 on Build GRCh38
rs964809328 Chr.20: 32446507-32446507 on Build GRCh38
rs974754950 Chr.20: 32446323-32446323 on Build GRCh38
rs907812511 Chr.20: 32446582-32446582 on Build GRCh38
rs368285426 Chr.20: 32446425-32446425 on Build GRCh38
rs967263654 Chr.20: 32446477-32446477 on Build GRCh38
rs564381076 Chr.20: 32446298-32446298 on Build GRCh38
rs907697762 Chr.20: 32446401-32446401 on Build GRCh38
rs937102786 Chr.20: 32446597-32446597 on Build GRCh38
rs536602056 Chr.20: 32446439-32446439 on Build GRCh38
rs1015372848 Chr.20: 32446261-32446261 on Build GRCh38
rs529526825 Chr.20: 32446400-32446400 on Build GRCh38
rs949892689 Chr.20: 32446591-32446592 on Build GRCh38
rs190077687 Chr.20: 32446321-32446321 on Build GRCh38
rs569334088 Chr.20: 32446431-32446431 on Build GRCh38
rs534867801 Chr.20: 32446547-32446547 on Build GRCh38
rs528120535 Chr.20: 32446415-32446415 on Build GRCh38
rs1004352602 Chr.20: 32446258-32446258 on Build GRCh38
rs71299231 Chr.20: 32446244-32446245 on Build GRCh38
rs764484705 Chr.20: 32446594-32446597 on Build GRCh38
rs763948335 Chr.20: 32446511-32446511 on Build GRCh38
rs1019419266 Chr.20: 32446495-32446495 on Build GRCh38
rs966939541 Chr.20: 32446237-32446238 on Build GRCh38
rs974707534 Chr.20: 32446520-32446520 on Build GRCh38
rs1005085682 Chr.20: 32446594-32446594 on Build GRCh38
rs886594032 Chr.20: 32446593-32446593 on Build GRCh38
rs570447140 Chr.20: 32446546-32446546 on Build GRCh38
rs67549403 Chr.20: 32446234-32446235 on Build GRCh38
rs995090357 Chr.20: 32446147-32446147 on Build GRCh38
rs920487306 Chr.20: 32446330-32446330 on Build GRCh38
rs969915521 Chr.20: 32446375-32446375 on Build GRCh38
rs949936958 Chr.20: 32446405-32446405 on Build GRCh38
rs1049366016 Chr.20: 32446576-32446576 on Build GRCh38
rs181772269 Chr.20: 32446449-32446449 on Build GRCh38
rs1045875712 Chr.20: 32446407-32446407 on Build GRCh38
rs1047598611 Chr.20: 32446146-32446146 on Build GRCh38

View Full Product Details