Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32447662-32448166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL4L
Entrez Gene ID
140688
Gene Name
nucleolar protein 4 like
Gene Aliases
C20orf112, C20orf113, dJ1184F4.2, dJ1184F4.4
UniGene
Hs.516978
Gene Chromosome Location
Chr.20: 32443059-32585073 on Build GRCh38
Transcripts
NM_001256798.1 NM_080616.4 XM_005260282.4 XM_005260283.4 XM_005260284.4 XM_005260285.4 XM_005260286.4 XM_005260288.2 XM_005260289.4 XM_006723702.3 XM_006723703.3 XM_011528562.2 XM_011528563.1 XM_017027669.1 XM_017027670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419002 c.1186A>C p.T396P Chr.20: 32447721-32447721 on Build GRCh38
1025748 c.1168A>G p.T390A Chr.20: 32447739-32447739 on Build GRCh38
6252279 c.1116C>A p.G372G Chr.20: 32447791-32447791 on Build GRCh38
6615503 c.1106G>A p.S369N Chr.20: 32447801-32447801 on Build GRCh38
4667456 c.1215C>T p.S405S Chr.20: 32447692-32447692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761283119 Chr.20: 32447777-32447777 on Build GRCh38
rs1024902516 Chr.20: 32447997-32447997 on Build GRCh38
rs758494565 Chr.20: 32447689-32447689 on Build GRCh38
rs920920562 Chr.20: 32448086-32448086 on Build GRCh38
rs1050359056 Chr.20: 32447904-32447904 on Build GRCh38
rs371317709 Chr.20: 32448077-32448077 on Build GRCh38
rs745812555 Chr.20: 32447762-32447762 on Build GRCh38
rs544434494 Chr.20: 32448045-32448045 on Build GRCh38
rs749152465 Chr.20: 32447768-32447768 on Build GRCh38
rs780026785 Chr.20: 32447854-32447854 on Build GRCh38
rs779721841 Chr.20: 32447767-32447767 on Build GRCh38
rs555481544 Chr.20: 32448001-32448001 on Build GRCh38
rs747061655 Chr.20: 32447756-32447756 on Build GRCh38
rs143292226 Chr.20: 32447692-32447692 on Build GRCh38
rs767361472 Chr.20: 32448065-32448065 on Build GRCh38
rs144443155 Chr.20: 32447923-32447924 on Build GRCh38
rs909382385 Chr.20: 32447836-32447836 on Build GRCh38
rs962060950 Chr.20: 32447723-32447723 on Build GRCh38
rs999106618 Chr.20: 32448041-32448041 on Build GRCh38
rs147561113 Chr.20: 32448132-32448132 on Build GRCh38
rs988677351 Chr.20: 32448066-32448066 on Build GRCh38
rs373397422 Chr.20: 32447797-32447797 on Build GRCh38
rs1011441046 Chr.20: 32447964-32447964 on Build GRCh38
rs777147266 Chr.20: 32447784-32447784 on Build GRCh38
rs185045046 Chr.20: 32447962-32447962 on Build GRCh38
rs183406769 Chr.20: 32447827-32447827 on Build GRCh38
rs896850940 Chr.20: 32448143-32448158 on Build GRCh38
rs779165706 Chr.20: 32447685-32447685 on Build GRCh38
rs766890241 Chr.20: 32447728-32447728 on Build GRCh38
rs922947668 Chr.20: 32447872-32447872 on Build GRCh38
rs778198218 Chr.20: 32447754-32447754 on Build GRCh38
rs758688029 Chr.20: 32447749-32447749 on Build GRCh38
rs774040982 Chr.20: 32447771-32447771 on Build GRCh38
rs756005579 Chr.20: 32447834-32447834 on Build GRCh38
rs368933653 Chr.20: 32447790-32447790 on Build GRCh38
rs887599037 Chr.20: 32447940-32447940 on Build GRCh38
rs769503846 Chr.20: 32447706-32447706 on Build GRCh38
rs34009028 Chr.20: 32447926-32447927 on Build GRCh38
rs975092133 Chr.20: 32447794-32447794 on Build GRCh38
rs748349050 Chr.20: 32447688-32447688 on Build GRCh38
rs763531949 Chr.20: 32447720-32447720 on Build GRCh38
rs569209037 Chr.20: 32447941-32447941 on Build GRCh38
rs773849504 Chr.20: 32447722-32447722 on Build GRCh38
rs894362823 Chr.20: 32447919-32447920 on Build GRCh38
rs189904792 Chr.20: 32448052-32448052 on Build GRCh38
rs755331445 Chr.20: 32447736-32447736 on Build GRCh38
rs78596202 Chr.20: 32447867-32447867 on Build GRCh38
rs776479346 Chr.20: 32447698-32447698 on Build GRCh38
rs907296438 Chr.20: 32447895-32447895 on Build GRCh38
rs71338906 Chr.20: 32447783-32447783 on Build GRCh38
rs952394507 Chr.20: 32448087-32448087 on Build GRCh38
rs746793599 Chr.20: 32448100-32448101 on Build GRCh38
rs141596773 Chr.20: 32447960-32447961 on Build GRCh38
rs771514773 Chr.20: 32447779-32447779 on Build GRCh38
rs774601152 Chr.20: 32447733-32447744 on Build GRCh38
rs749063678 Chr.20: 32447865-32447865 on Build GRCh38
rs745641509 Chr.20: 32447702-32447702 on Build GRCh38
rs753096212 Chr.20: 32447791-32447791 on Build GRCh38
rs969576664 Chr.20: 32447753-32447753 on Build GRCh38
rs144695451 Chr.20: 32447701-32447701 on Build GRCh38
rs1036444493 Chr.20: 32447980-32447980 on Build GRCh38
rs765543770 Chr.20: 32447738-32447738 on Build GRCh38
rs1036437754 Chr.20: 32447880-32447880 on Build GRCh38
rs752899897 Chr.20: 32447746-32447746 on Build GRCh38
rs897863694 Chr.20: 32447985-32447985 on Build GRCh38
rs762177818 Chr.20: 32447743-32447766 on Build GRCh38
rs748767627 Chr.20: 32447956-32447956 on Build GRCh38
rs770976546 Chr.20: 32447694-32447694 on Build GRCh38
rs578099578 Chr.20: 32448097-32448097 on Build GRCh38
rs914082097 Chr.20: 32448100-32448100 on Build GRCh38
rs148090404 Chr.20: 32447760-32447760 on Build GRCh38
rs774199753 Chr.20: 32448020-32448020 on Build GRCh38
rs373464965 Chr.20: 32447719-32447719 on Build GRCh38
rs1055725690 Chr.20: 32447903-32447903 on Build GRCh38
rs527497446 Chr.20: 32447900-32447900 on Build GRCh38
rs878994620 Chr.20: 32447936-32447936 on Build GRCh38
rs112169656 Chr.20: 32447950-32447950 on Build GRCh38
rs749342775 Chr.20: 32447732-32447733 on Build GRCh38
rs573793736 Chr.20: 32448032-32448032 on Build GRCh38
rs757451105 Chr.20: 32447758-32447758 on Build GRCh38
rs769570581 Chr.20: 32447763-32447763 on Build GRCh38
rs3746612 Chr.20: 32448133-32448133 on Build GRCh38
rs568149628 Chr.20: 32447761-32447761 on Build GRCh38
rs1006290048 Chr.20: 32447713-32447713 on Build GRCh38
rs775154968 Chr.20: 32447718-32447718 on Build GRCh38
rs759964502 Chr.20: 32447787-32447787 on Build GRCh38
rs115327812 Chr.20: 32447999-32447999 on Build GRCh38
rs201048795 Chr.20: 32447927-32447928 on Build GRCh38
rs368927465 Chr.20: 32447828-32447828 on Build GRCh38
rs985024782 Chr.20: 32448105-32448105 on Build GRCh38
rs745580736 Chr.20: 32447809-32447809 on Build GRCh38
rs1019336328 Chr.20: 32448044-32448044 on Build GRCh38
rs377542764 Chr.20: 32447799-32447799 on Build GRCh38
rs566756988 Chr.20: 32447996-32447996 on Build GRCh38
rs766264637 Chr.20: 32447762-32447770 on Build GRCh38
rs546385167 Chr.20: 32447691-32447691 on Build GRCh38
rs761373270 Chr.20: 32447727-32447727 on Build GRCh38
rs200048126 Chr.20: 32447921-32447921 on Build GRCh38
rs922138702 Chr.20: 32447810-32447810 on Build GRCh38
rs539366107 Chr.20: 32447959-32447959 on Build GRCh38
rs369968679 Chr.20: 32447683-32447683 on Build GRCh38
rs764377090 Chr.20: 32447798-32447798 on Build GRCh38
rs370815412 Chr.20: 32447819-32447819 on Build GRCh38
rs768651158 Chr.20: 32447733-32447738 on Build GRCh38

View Full Product Details