Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32452144-32452649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL4L
Entrez Gene ID
140688
Gene Name
nucleolar protein 4 like
Gene Aliases
C20orf112, C20orf113, dJ1184F4.2, dJ1184F4.4
UniGene
Hs.516978
Gene Chromosome Location
Chr.20: 32443059-32585073 on Build GRCh38
Transcripts
NM_001256798.1 NM_080616.4 XM_005260282.4 XM_005260283.4 XM_005260284.4 XM_005260285.4 XM_005260286.4 XM_005260288.2 XM_005260289.4 XM_006723702.3 XM_006723703.3 XM_011528562.2 XM_011528563.1 XM_017027669.1 XM_017027670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4752073 c.988G>A p.V330M Chr.20: 32452338-32452338 on Build GRCh38
2889329 c.971_973delCCT p.S324delS Chr.20: 32452353-32452355 on Build GRCh38
2889331 c.920C>T p.S307L Chr.20: 32452406-32452406 on Build GRCh38
4168790 c.1021C>T p.R341C Chr.20: 32452305-32452305 on Build GRCh38
6615505 c.975C>T p.Y325Y Chr.20: 32452351-32452351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753806738 Chr.20: 32452283-32452283 on Build GRCh38
rs754915933 Chr.20: 32452332-32452332 on Build GRCh38
rs895708009 Chr.20: 32452196-32452196 on Build GRCh38
rs763961209 Chr.20: 32452410-32452410 on Build GRCh38
rs71338907 Chr.20: 32452443-32452443 on Build GRCh38
rs569767483 Chr.20: 32452617-32452617 on Build GRCh38
rs767851391 Chr.20: 32452241-32452241 on Build GRCh38
rs564544713 Chr.20: 32452458-32452458 on Build GRCh38
rs1048647527 Chr.20: 32452170-32452170 on Build GRCh38
rs756767332 Chr.20: 32452364-32452364 on Build GRCh38
rs772591358 Chr.20: 32452202-32452202 on Build GRCh38
rs181509028 Chr.20: 32452524-32452524 on Build GRCh38
rs770503584 Chr.20: 32452303-32452303 on Build GRCh38
rs756961760 Chr.20: 32452423-32452423 on Build GRCh38
rs73906141 Chr.20: 32452264-32452264 on Build GRCh38
rs143421203 Chr.20: 32452297-32452297 on Build GRCh38
rs749822933 Chr.20: 32452315-32452315 on Build GRCh38
rs760632730 Chr.20: 32452328-32452328 on Build GRCh38
rs867811079 Chr.20: 32452205-32452205 on Build GRCh38
rs759407418 Chr.20: 32452451-32452452 on Build GRCh38
rs939753717 Chr.20: 32452620-32452620 on Build GRCh38
rs755727672 Chr.20: 32452446-32452446 on Build GRCh38
rs2236156 Chr.20: 32452228-32452228 on Build GRCh38
rs750459964 Chr.20: 32452253-32452253 on Build GRCh38
rs748432678 Chr.20: 32452393-32452393 on Build GRCh38
rs199635019 Chr.20: 32452476-32452476 on Build GRCh38
rs956080636 Chr.20: 32452545-32452545 on Build GRCh38
rs758618911 Chr.20: 32452428-32452428 on Build GRCh38
rs773543869 Chr.20: 32452166-32452166 on Build GRCh38
rs766380065 Chr.20: 32452403-32452403 on Build GRCh38
rs908068237 Chr.20: 32452608-32452608 on Build GRCh38
rs1010344742 Chr.20: 32452548-32452548 on Build GRCh38
rs771554192 Chr.20: 32452198-32452198 on Build GRCh38
rs776178605 Chr.20: 32452222-32452222 on Build GRCh38
rs370107804 Chr.20: 32452216-32452216 on Build GRCh38
rs756269249 Chr.20: 32452261-32452261 on Build GRCh38
rs374479145 Chr.20: 32452323-32452323 on Build GRCh38
rs766699175 Chr.20: 32452224-32452224 on Build GRCh38
rs752710529 Chr.20: 32452408-32452408 on Build GRCh38
rs745434199 Chr.20: 32452370-32452370 on Build GRCh38
rs975014798 Chr.20: 32452597-32452597 on Build GRCh38
rs966063965 Chr.20: 32452589-32452589 on Build GRCh38
rs1000399019 Chr.20: 32452489-32452489 on Build GRCh38
rs922217959 Chr.20: 32452598-32452598 on Build GRCh38
rs371649171 Chr.20: 32452340-32452340 on Build GRCh38
rs763366648 Chr.20: 32452226-32452226 on Build GRCh38
rs760290348 Chr.20: 32452210-32452210 on Build GRCh38
rs751102382 Chr.20: 32452412-32452412 on Build GRCh38
rs769277120 Chr.20: 32452376-32452376 on Build GRCh38
rs776088625 Chr.20: 32452304-32452304 on Build GRCh38
rs907646364 Chr.20: 32452599-32452600 on Build GRCh38
rs1010974412 Chr.20: 32452277-32452277 on Build GRCh38
rs547762682 Chr.20: 32452606-32452606 on Build GRCh38
rs575631370 Chr.20: 32452293-32452293 on Build GRCh38
rs186684995 Chr.20: 32452529-32452529 on Build GRCh38
rs760965780 Chr.20: 32452402-32452402 on Build GRCh38
rs776678981 Chr.20: 32452405-32452405 on Build GRCh38
rs767614583 Chr.20: 32452326-32452326 on Build GRCh38
rs752396462 Chr.20: 32452406-32452406 on Build GRCh38
rs1018938995 Chr.20: 32452554-32452554 on Build GRCh38
rs75184133 Chr.20: 32452288-32452288 on Build GRCh38
rs779250690 Chr.20: 32452452-32452452 on Build GRCh38
rs765215561 Chr.20: 32452333-32452333 on Build GRCh38
rs1003794966 Chr.20: 32452189-32452189 on Build GRCh38
rs952336967 Chr.20: 32452605-32452605 on Build GRCh38
rs956113393 Chr.20: 32452273-32452273 on Build GRCh38
rs749935362 Chr.20: 32452431-32452431 on Build GRCh38
rs747442157 Chr.20: 32452475-32452475 on Build GRCh38
rs773526028 Chr.20: 32452327-32452327 on Build GRCh38
rs753001958 Chr.20: 32452344-32452345 on Build GRCh38
rs753717012 Chr.20: 32452331-32452331 on Build GRCh38
rs367872242 Chr.20: 32452324-32452324 on Build GRCh38
rs777528566 Chr.20: 32452302-32452302 on Build GRCh38
rs189664799 Chr.20: 32452229-32452229 on Build GRCh38
rs778499322 Chr.20: 32452186-32452186 on Build GRCh38
rs376348600 Chr.20: 32452396-32452396 on Build GRCh38
rs778204145 Chr.20: 32452459-32452459 on Build GRCh38
rs777250272 Chr.20: 32452342-32452342 on Build GRCh38
rs748713489 Chr.20: 32452457-32452457 on Build GRCh38
rs774975220 Chr.20: 32452392-32452392 on Build GRCh38
rs772520747 Chr.20: 32452395-32452395 on Build GRCh38
rs780849512 Chr.20: 32452369-32452369 on Build GRCh38
rs546242617 Chr.20: 32452359-32452359 on Build GRCh38
rs757973555 Chr.20: 32452300-32452300 on Build GRCh38
rs752411871 Chr.20: 32452339-32452339 on Build GRCh38
rs922110659 Chr.20: 32452311-32452311 on Build GRCh38
rs540534054 Chr.20: 32452512-32452512 on Build GRCh38
rs62206935 Chr.20: 32452190-32452190 on Build GRCh38
rs758077558 Chr.20: 32452409-32452409 on Build GRCh38
rs781052029 Chr.20: 32452427-32452427 on Build GRCh38
rs988780263 Chr.20: 32452274-32452274 on Build GRCh38

View Full Product Details