Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32452803-32453310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOL4L
Entrez Gene ID
140688
Gene Name
nucleolar protein 4 like
Gene Aliases
C20orf112, C20orf113, dJ1184F4.2, dJ1184F4.4
UniGene
Hs.516978
Gene Chromosome Location
Chr.20: 32443059-32585073 on Build GRCh38
Transcripts
NM_001256798.1 NM_080616.4 XM_005260282.4 XM_005260283.4 XM_005260284.4 XM_005260285.4 XM_005260286.4 XM_005260288.2 XM_005260289.4 XM_006723702.3 XM_006723703.3 XM_011528562.2 XM_011528563.1 XM_017027669.1 XM_017027670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2889333 c.834C>T p.S278S Chr.20: 32452938-32452938 on Build GRCh38
6615494 c.795G>A p.S265S Chr.20: 32452977-32452977 on Build GRCh38
5857653 c.761A>G p.E254G Chr.20: 32453308-32453308 on Build GRCh38
4442901 c.863G>A p.R288H Chr.20: 32452909-32452909 on Build GRCh38
4884269 c.834C>A p.S278R Chr.20: 32452938-32452938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6058700 Chr.20: 32453080-32453080 on Build GRCh38
rs779630871 Chr.20: 32452874-32452874 on Build GRCh38
rs573141434 Chr.20: 32453089-32453089 on Build GRCh38
rs775348803 Chr.20: 32453175-32453175 on Build GRCh38
rs756073270 Chr.20: 32453239-32453239 on Build GRCh38
rs962269831 Chr.20: 32453103-32453103 on Build GRCh38
rs752083546 Chr.20: 32453044-32453044 on Build GRCh38
rs373913533 Chr.20: 32453020-32453020 on Build GRCh38
rs530480650 Chr.20: 32453136-32453136 on Build GRCh38
rs958376543 Chr.20: 32453125-32453125 on Build GRCh38
rs529528594 Chr.20: 32453102-32453102 on Build GRCh38
rs775631440 Chr.20: 32452992-32452992 on Build GRCh38
rs751353339 Chr.20: 32452856-32452856 on Build GRCh38
rs551997450 Chr.20: 32453197-32453197 on Build GRCh38
rs757559459 Chr.20: 32453047-32453047 on Build GRCh38
rs926960317 Chr.20: 32452863-32452863 on Build GRCh38
rs926941193 Chr.20: 32453134-32453134 on Build GRCh38
rs985082879 Chr.20: 32452875-32452875 on Build GRCh38
rs945958946 Chr.20: 32453237-32453237 on Build GRCh38
rs376606821 Chr.20: 32453269-32453269 on Build GRCh38
rs990226217 Chr.20: 32453204-32453204 on Build GRCh38
rs915497903 Chr.20: 32452946-32452946 on Build GRCh38
rs568363285 Chr.20: 32452845-32452845 on Build GRCh38
rs979901423 Chr.20: 32453120-32453120 on Build GRCh38
rs778191302 Chr.20: 32452914-32452914 on Build GRCh38
rs908035293 Chr.20: 32452927-32452927 on Build GRCh38
rs761752935 Chr.20: 32452859-32452859 on Build GRCh38
rs935715253 Chr.20: 32453206-32453206 on Build GRCh38
rs570519344 Chr.20: 32453216-32453216 on Build GRCh38
rs201640029 Chr.20: 32452851-32452851 on Build GRCh38
rs181863721 Chr.20: 32453270-32453270 on Build GRCh38
rs758939545 Chr.20: 32452898-32452898 on Build GRCh38
rs755982586 Chr.20: 32452886-32452886 on Build GRCh38
rs748112434 Chr.20: 32453281-32453281 on Build GRCh38
rs996293880 Chr.20: 32453069-32453069 on Build GRCh38
rs200806836 Chr.20: 32453043-32453043 on Build GRCh38
rs568224578 Chr.20: 32453288-32453288 on Build GRCh38
rs924089718 Chr.20: 32453252-32453252 on Build GRCh38
rs189099027 Chr.20: 32453116-32453116 on Build GRCh38
rs540423385 Chr.20: 32453093-32453093 on Build GRCh38
rs557090276 Chr.20: 32452854-32452854 on Build GRCh38
rs774385500 Chr.20: 32453272-32453272 on Build GRCh38
rs747342316 Chr.20: 32452938-32452938 on Build GRCh38
rs371968925 Chr.20: 32453050-32453050 on Build GRCh38
rs1029389011 Chr.20: 32453077-32453077 on Build GRCh38
rs1027531207 Chr.20: 32453056-32453056 on Build GRCh38
rs373854304 Chr.20: 32452995-32452995 on Build GRCh38
rs971533437 Chr.20: 32453118-32453118 on Build GRCh38
rs927049569 Chr.20: 32453124-32453124 on Build GRCh38
rs760409272 Chr.20: 32453027-32453027 on Build GRCh38
rs767446058 Chr.20: 32453019-32453019 on Build GRCh38
rs754485404 Chr.20: 32453041-32453041 on Build GRCh38
rs746294185 Chr.20: 32453051-32453051 on Build GRCh38
rs753377000 Chr.20: 32453036-32453036 on Build GRCh38
rs41310799 Chr.20: 32452980-32452980 on Build GRCh38
rs376588401 Chr.20: 32452895-32452895 on Build GRCh38
rs369056301 Chr.20: 32452985-32452985 on Build GRCh38
rs560876416 Chr.20: 32453223-32453223 on Build GRCh38
rs544469432 Chr.20: 32453110-32453110 on Build GRCh38
rs768619974 Chr.20: 32452996-32452996 on Build GRCh38
rs887811928 Chr.20: 32453078-32453078 on Build GRCh38
rs971112130 Chr.20: 32452850-32452850 on Build GRCh38
rs1737888 Chr.20: 32453229-32453229 on Build GRCh38
rs372537802 Chr.20: 32452853-32452853 on Build GRCh38
rs562071973 Chr.20: 32453095-32453095 on Build GRCh38
rs992249771 Chr.20: 32453109-32453109 on Build GRCh38
rs1040207452 Chr.20: 32453263-32453263 on Build GRCh38
rs780184820 Chr.20: 32453268-32453268 on Build GRCh38
rs952061569 Chr.20: 32453064-32453064 on Build GRCh38
rs766014578 Chr.20: 32453029-32453029 on Build GRCh38
rs901895399 Chr.20: 32453068-32453068 on Build GRCh38
rs755639817 Chr.20: 32452872-32452872 on Build GRCh38
rs771314160 Chr.20: 32452961-32452961 on Build GRCh38
rs144141778 Chr.20: 32452977-32452977 on Build GRCh38
rs771119815 Chr.20: 32452844-32452844 on Build GRCh38
rs1054403945 Chr.20: 32453030-32453030 on Build GRCh38
rs756343073 Chr.20: 32453253-32453253 on Build GRCh38
rs774473113 Chr.20: 32453013-32453013 on Build GRCh38
rs761664909 Chr.20: 32453017-32453017 on Build GRCh38

View Full Product Details