Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33085939-33086440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB4
Entrez Gene ID
149954
Gene Name
BPI fold containing family B member 4
Gene Aliases
C20orf186, LPLUNC4, RY2G5, dJ726C3.5
UniGene
Hs.38961
Gene Chromosome Location
Chr.20: 33081512-33111751 on Build GRCh38
Transcripts
NM_182519.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6849987 c.764G>A p.R255Q Chr.20: 33086119-33086119 on Build GRCh38
5565404 c.748C>T p.R250W Chr.20: 33086103-33086103 on Build GRCh38
3758518 c.685A>G p.I229V Chr.20: 33086040-33086040 on Build GRCh38
5614773 c.716T>C p.V239A Chr.20: 33086071-33086071 on Build GRCh38
4836872 c.767G>C p.C256S Chr.20: 33086122-33086122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760953021 Chr.20: 33086209-33086209 on Build GRCh38
rs148384178 Chr.20: 33086058-33086058 on Build GRCh38
rs550476702 Chr.20: 33086272-33086272 on Build GRCh38
rs781299785 Chr.20: 33086033-33086033 on Build GRCh38
rs745444866 Chr.20: 33086019-33086019 on Build GRCh38
rs950264078 Chr.20: 33086244-33086244 on Build GRCh38
rs529013098 Chr.20: 33086396-33086396 on Build GRCh38
rs147477530 Chr.20: 33086246-33086246 on Build GRCh38
rs535736291 Chr.20: 33086119-33086119 on Build GRCh38
rs376856849 Chr.20: 33086161-33086161 on Build GRCh38
rs752475726 Chr.20: 33086168-33086169 on Build GRCh38
rs557534278 Chr.20: 33086121-33086121 on Build GRCh38
rs188756520 Chr.20: 33086113-33086113 on Build GRCh38
rs901879515 Chr.20: 33086344-33086344 on Build GRCh38
rs780209201 Chr.20: 33086042-33086042 on Build GRCh38
rs140703275 Chr.20: 33085982-33085982 on Build GRCh38
rs373318056 Chr.20: 33086194-33086194 on Build GRCh38
rs774100558 Chr.20: 33086054-33086054 on Build GRCh38
rs41305787 Chr.20: 33086177-33086177 on Build GRCh38
rs541709460 Chr.20: 33086300-33086300 on Build GRCh38
rs777431886 Chr.20: 33086175-33086175 on Build GRCh38
rs906003483 Chr.20: 33086247-33086247 on Build GRCh38
rs772761772 Chr.20: 33086057-33086057 on Build GRCh38
rs998083169 Chr.20: 33086346-33086346 on Build GRCh38
rs745984159 Chr.20: 33086036-33086036 on Build GRCh38
rs761352164 Chr.20: 33085975-33085975 on Build GRCh38
rs758819681 Chr.20: 33086262-33086262 on Build GRCh38
rs1054506130 Chr.20: 33086261-33086261 on Build GRCh38
rs374496259 Chr.20: 33086043-33086043 on Build GRCh38
rs139770462 Chr.20: 33086163-33086163 on Build GRCh38
rs2070325 Chr.20: 33086040-33086040 on Build GRCh38
rs753959899 Chr.20: 33086212-33086212 on Build GRCh38
rs370634025 Chr.20: 33085979-33085979 on Build GRCh38
rs112859084 Chr.20: 33085963-33085963 on Build GRCh38
rs369833747 Chr.20: 33086129-33086129 on Build GRCh38
rs546003483 Chr.20: 33086389-33086389 on Build GRCh38
rs202190359 Chr.20: 33086103-33086103 on Build GRCh38
rs368074260 Chr.20: 33086088-33086088 on Build GRCh38
rs929885465 Chr.20: 33086390-33086390 on Build GRCh38
rs200540628 Chr.20: 33086089-33086089 on Build GRCh38
rs575310575 Chr.20: 33086379-33086379 on Build GRCh38
rs756173199 Chr.20: 33086139-33086139 on Build GRCh38
rs754015765 Chr.20: 33086141-33086141 on Build GRCh38
rs1010315975 Chr.20: 33086285-33086285 on Build GRCh38
rs368742859 Chr.20: 33086086-33086086 on Build GRCh38
rs576510614 Chr.20: 33086239-33086239 on Build GRCh38
rs939685154 Chr.20: 33085999-33085999 on Build GRCh38
rs191783938 Chr.20: 33086056-33086056 on Build GRCh38
rs753052746 Chr.20: 33085997-33085997 on Build GRCh38
rs373472303 Chr.20: 33086136-33086136 on Build GRCh38
rs983862154 Chr.20: 33086152-33086152 on Build GRCh38
rs760063308 Chr.20: 33085991-33085991 on Build GRCh38
rs150651189 Chr.20: 33086160-33086160 on Build GRCh38
rs964231196 Chr.20: 33086090-33086100 on Build GRCh38
rs542045948 Chr.20: 33086082-33086082 on Build GRCh38
rs758139078 Chr.20: 33086165-33086165 on Build GRCh38
rs267605884 Chr.20: 33086084-33086084 on Build GRCh38
rs113085566 Chr.20: 33086313-33086313 on Build GRCh38
rs772348892 Chr.20: 33086198-33086198 on Build GRCh38
rs377208700 Chr.20: 33086118-33086118 on Build GRCh38
rs985510501 Chr.20: 33086287-33086287 on Build GRCh38
rs143302622 Chr.20: 33086069-33086069 on Build GRCh38
rs761623500 Chr.20: 33085974-33085974 on Build GRCh38
rs762151160 Chr.20: 33086127-33086127 on Build GRCh38
rs771748280 Chr.20: 33086111-33086111 on Build GRCh38
rs143049942 Chr.20: 33086108-33086108 on Build GRCh38
rs775988803 Chr.20: 33086085-33086085 on Build GRCh38
rs574684272 Chr.20: 33086274-33086274 on Build GRCh38
rs764633887 Chr.20: 33086125-33086125 on Build GRCh38
rs770610795 Chr.20: 33086178-33086178 on Build GRCh38
rs138318222 Chr.20: 33086104-33086104 on Build GRCh38
rs372363944 Chr.20: 33086093-33086093 on Build GRCh38
rs1037245042 Chr.20: 33086034-33086034 on Build GRCh38
rs780656991 Chr.20: 33086179-33086179 on Build GRCh38
rs774766247 Chr.20: 33086126-33086126 on Build GRCh38
rs766760226 Chr.20: 33086210-33086210 on Build GRCh38
rs771095133 Chr.20: 33086375-33086375 on Build GRCh38
rs777912349 Chr.20: 33086002-33086002 on Build GRCh38
rs779119016 Chr.20: 33086235-33086235 on Build GRCh38
rs112296230 Chr.20: 33086092-33086092 on Build GRCh38
rs866143866 Chr.20: 33086070-33086070 on Build GRCh38
rs374179235 Chr.20: 33086095-33086095 on Build GRCh38
rs376957231 Chr.20: 33086101-33086101 on Build GRCh38
rs773446063 Chr.20: 33086201-33086201 on Build GRCh38
rs769147128 Chr.20: 33086186-33086186 on Build GRCh38
rs956991804 Chr.20: 33086204-33086204 on Build GRCh38
rs370847069 Chr.20: 33086018-33086018 on Build GRCh38
rs745440900 Chr.20: 33086184-33086184 on Build GRCh38
rs766600603 Chr.20: 33086140-33086140 on Build GRCh38
rs61739412 Chr.20: 33086087-33086087 on Build GRCh38
rs41311298 Chr.20: 33086255-33086255 on Build GRCh38
rs558154507 Chr.20: 33086026-33086026 on Build GRCh38
rs558268816 Chr.20: 33086234-33086234 on Build GRCh38
rs751800937 Chr.20: 33086025-33086025 on Build GRCh38
rs762180668 Chr.20: 33086196-33086196 on Build GRCh38
rs373989737 Chr.20: 33085992-33085992 on Build GRCh38

View Full Product Details