Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33288551-33288972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB1
Entrez Gene ID
92747
Gene Name
BPI fold containing family B member 1
Gene Aliases
C20orf114, LPLUNC1
UniGene
Hs.65551
Gene Chromosome Location
Chr.20: 33283135-33309878 on Build GRCh38
Transcripts
NM_033197.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1532986 c.187C>T p.R63W Chr.20: 33288812-33288812 on Build GRCh38
724031 c.238G>A p.V80I Chr.20: 33288863-33288863 on Build GRCh38
1025896 c.201C>T p.A67A Chr.20: 33288826-33288826 on Build GRCh38
4097656 c.222C>T p.G74G Chr.20: 33288847-33288847 on Build GRCh38
443659 c.129G>T p.E43D Chr.20: 33288754-33288754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771308005 Chr.20: 33288800-33288800 on Build GRCh38
rs561565814 Chr.20: 33288701-33288701 on Build GRCh38
rs779496486 Chr.20: 33288790-33288790 on Build GRCh38
rs773504875 Chr.20: 33288716-33288716 on Build GRCh38
rs746081066 Chr.20: 33288912-33288912 on Build GRCh38
rs750095021 Chr.20: 33288854-33288854 on Build GRCh38
rs963470910 Chr.20: 33288840-33288840 on Build GRCh38
rs780348900 Chr.20: 33288693-33288715 on Build GRCh38
rs745327935 Chr.20: 33288695-33288695 on Build GRCh38
rs981406132 Chr.20: 33288683-33288683 on Build GRCh38
rs746099218 Chr.20: 33288807-33288807 on Build GRCh38
rs770053081 Chr.20: 33288914-33288914 on Build GRCh38
rs767421343 Chr.20: 33288845-33288845 on Build GRCh38
rs1016419743 Chr.20: 33288837-33288837 on Build GRCh38
rs150010004 Chr.20: 33288796-33288796 on Build GRCh38
rs200424710 Chr.20: 33288927-33288927 on Build GRCh38
rs560066748 Chr.20: 33288674-33288674 on Build GRCh38
rs367680625 Chr.20: 33288895-33288895 on Build GRCh38
rs776871760 Chr.20: 33288736-33288736 on Build GRCh38
rs1007738173 Chr.20: 33288836-33288836 on Build GRCh38
rs370901362 Chr.20: 33288862-33288862 on Build GRCh38
rs1032946552 Chr.20: 33288595-33288595 on Build GRCh38
rs138442502 Chr.20: 33288935-33288935 on Build GRCh38
rs752616809 Chr.20: 33288762-33288762 on Build GRCh38
rs753327528 Chr.20: 33288866-33288866 on Build GRCh38
rs972079959 Chr.20: 33288635-33288635 on Build GRCh38
rs2295576 Chr.20: 33288679-33288679 on Build GRCh38
rs760927235 Chr.20: 33288724-33288724 on Build GRCh38
rs914324514 Chr.20: 33288751-33288751 on Build GRCh38
rs111428112 Chr.20: 33288942-33288942 on Build GRCh38
rs763895083 Chr.20: 33288820-33288820 on Build GRCh38
rs868840168 Chr.20: 33288920-33288920 on Build GRCh38
rs1024530387 Chr.20: 33288730-33288730 on Build GRCh38
rs763939076 Chr.20: 33288766-33288766 on Build GRCh38
rs761638589 Chr.20: 33288839-33288839 on Build GRCh38
rs762870231 Chr.20: 33288763-33288763 on Build GRCh38
rs1078761 Chr.20: 33288875-33288875 on Build GRCh38
rs367705404 Chr.20: 33288706-33288706 on Build GRCh38
rs537632311 Chr.20: 33288907-33288907 on Build GRCh38
rs755196186 Chr.20: 33288791-33288792 on Build GRCh38
rs139237364 Chr.20: 33288793-33288793 on Build GRCh38
rs771183559 Chr.20: 33288726-33288726 on Build GRCh38
rs145225110 Chr.20: 33288810-33288810 on Build GRCh38
rs779222797 Chr.20: 33288913-33288914 on Build GRCh38
rs1008703446 Chr.20: 33288634-33288634 on Build GRCh38
rs372358506 Chr.20: 33288689-33288689 on Build GRCh38
rs538717081 Chr.20: 33288941-33288941 on Build GRCh38
rs865867405 Chr.20: 33288913-33288913 on Build GRCh38
rs551400282 Chr.20: 33288827-33288827 on Build GRCh38
rs374640604 Chr.20: 33288863-33288863 on Build GRCh38
rs767225361 Chr.20: 33288770-33288770 on Build GRCh38
rs775809483 Chr.20: 33288926-33288926 on Build GRCh38
rs142208109 Chr.20: 33288868-33288868 on Build GRCh38
rs61737957 Chr.20: 33288741-33288741 on Build GRCh38
rs762929164 Chr.20: 33288813-33288813 on Build GRCh38
rs201956427 Chr.20: 33288795-33288795 on Build GRCh38
rs890388520 Chr.20: 33288617-33288617 on Build GRCh38
rs774085484 Chr.20: 33288929-33288929 on Build GRCh38
rs749386588 Chr.20: 33288765-33288767 on Build GRCh38
rs780814777 Chr.20: 33288693-33288693 on Build GRCh38
rs1038884389 Chr.20: 33288892-33288892 on Build GRCh38
rs776777296 Chr.20: 33288806-33288806 on Build GRCh38
rs761908126 Chr.20: 33288934-33288934 on Build GRCh38
rs150828161 Chr.20: 33288769-33288769 on Build GRCh38
rs928578755 Chr.20: 33288921-33288921 on Build GRCh38
rs919232924 Chr.20: 33288652-33288652 on Build GRCh38
rs201145500 Chr.20: 33288812-33288812 on Build GRCh38
rs1038894523 Chr.20: 33288663-33288663 on Build GRCh38
rs755617003 Chr.20: 33288789-33288789 on Build GRCh38
rs986700950 Chr.20: 33288576-33288576 on Build GRCh38
rs781606973 Chr.20: 33288902-33288902 on Build GRCh38
rs1016390391 Chr.20: 33288601-33288601 on Build GRCh38
rs754146128 Chr.20: 33288888-33288888 on Build GRCh38
rs900370317 Chr.20: 33288906-33288906 on Build GRCh38
rs759564093 Chr.20: 33288740-33288740 on Build GRCh38
rs750111009 Chr.20: 33288787-33288787 on Build GRCh38
rs963418924 Chr.20: 33288603-33288603 on Build GRCh38

View Full Product Details