Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33358761-33359263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP1
Entrez Gene ID
51654
Gene Name
CDK5 regulatory subunit associated protein 1
Gene Aliases
C20orf34, C42, CGI-05, HSPC167
UniGene
Hs.435952
Gene Chromosome Location
Chr.20: 33358839-33402768 on Build GRCh38
Transcripts
NM_001278167.1 NM_001278168.1 NM_001278169.1 NM_016082.4 NM_016408.3 XM_011528856.2 XM_011528857.2 XM_011528858.1 XM_011528859.2 XM_011528860.1 XM_017027876.1 XM_017027877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
478027 c.1689C>T p.T563T Chr.20: 33359118-33359118 on Build GRCh38
3363193 c.1686C>A p.I562I Chr.20: 33359121-33359121 on Build GRCh38
6467625 c.1727G>A p.C576Y Chr.20: 33359080-33359080 on Build GRCh38
1307284 c.1751C>G p.S584C Chr.20: 33359056-33359056 on Build GRCh38
6407965 c.1697G>T p.S566I Chr.20: 33359110-33359110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947313444 Chr.20: 33359114-33359114 on Build GRCh38
rs760707893 Chr.20: 33359087-33359087 on Build GRCh38
rs1043029939 Chr.20: 33358991-33358991 on Build GRCh38
rs770611478 Chr.20: 33359092-33359092 on Build GRCh38
rs764914439 Chr.20: 33359101-33359101 on Build GRCh38
rs903818749 Chr.20: 33359025-33359025 on Build GRCh38
rs113213388 Chr.20: 33358931-33358931 on Build GRCh38
rs1026999293 Chr.20: 33358914-33358914 on Build GRCh38
rs757985419 Chr.20: 33358995-33358995 on Build GRCh38
rs908694890 Chr.20: 33358961-33358961 on Build GRCh38
rs904518853 Chr.20: 33359234-33359234 on Build GRCh38
rs1055389557 Chr.20: 33358803-33358803 on Build GRCh38
rs563748318 Chr.20: 33358886-33358886 on Build GRCh38
rs1057475393 Chr.20: 33359211-33359211 on Build GRCh38
rs149419964 Chr.20: 33358869-33358869 on Build GRCh38
rs563284476 Chr.20: 33359193-33359193 on Build GRCh38
rs771366431 Chr.20: 33359157-33359157 on Build GRCh38
rs756427091 Chr.20: 33359021-33359021 on Build GRCh38
rs146671297 Chr.20: 33359046-33359046 on Build GRCh38
rs781741573 Chr.20: 33359006-33359006 on Build GRCh38
rs374958722 Chr.20: 33359086-33359086 on Build GRCh38
rs996818483 Chr.20: 33358891-33358891 on Build GRCh38
rs41287064 Chr.20: 33358927-33358927 on Build GRCh38
rs573775844 Chr.20: 33359039-33359039 on Build GRCh38
rs35714788 Chr.20: 33359005-33359005 on Build GRCh38
rs748172275 Chr.20: 33359061-33359061 on Build GRCh38
rs986553220 Chr.20: 33358830-33358830 on Build GRCh38
rs912284676 Chr.20: 33358832-33358832 on Build GRCh38
rs763723121 Chr.20: 33359119-33359119 on Build GRCh38
rs551721086 Chr.20: 33358885-33358885 on Build GRCh38
rs755352725 Chr.20: 33359165-33359165 on Build GRCh38
rs942358694 Chr.20: 33358851-33358851 on Build GRCh38
rs193301001 Chr.20: 33359196-33359196 on Build GRCh38
rs184316315 Chr.20: 33359205-33359205 on Build GRCh38
rs914246737 Chr.20: 33359105-33359105 on Build GRCh38
rs138912660 Chr.20: 33358824-33358824 on Build GRCh38
rs937047090 Chr.20: 33358789-33358789 on Build GRCh38
rs773019714 Chr.20: 33359077-33359077 on Build GRCh38
rs140207780 Chr.20: 33359095-33359095 on Build GRCh38
rs748436253 Chr.20: 33359172-33359172 on Build GRCh38
rs769660912 Chr.20: 33359141-33359143 on Build GRCh38
rs779114750 Chr.20: 33359052-33359052 on Build GRCh38
rs758442937 Chr.20: 33359175-33359175 on Build GRCh38
rs931552759 Chr.20: 33358881-33358881 on Build GRCh38
rs750992758 Chr.20: 33359014-33359014 on Build GRCh38
rs117988303 Chr.20: 33358825-33358825 on Build GRCh38
rs542745437 Chr.20: 33359044-33359044 on Build GRCh38
rs1008440083 Chr.20: 33359194-33359194 on Build GRCh38
rs754138775 Chr.20: 33359163-33359163 on Build GRCh38
rs756690799 Chr.20: 33359147-33359147 on Build GRCh38
rs562215755 Chr.20: 33358815-33358815 on Build GRCh38
rs908763823 Chr.20: 33359201-33359201 on Build GRCh38
rs979708280 Chr.20: 33358787-33358787 on Build GRCh38
rs777540865 Chr.20: 33359064-33359064 on Build GRCh38
rs1026966677 Chr.20: 33359223-33359223 on Build GRCh38
rs752411026 Chr.20: 33359104-33359104 on Build GRCh38
rs115726281 Chr.20: 33359140-33359140 on Build GRCh38
rs948803390 Chr.20: 33359229-33359229 on Build GRCh38
rs17124642 Chr.20: 33358855-33358855 on Build GRCh38
rs1020099524 Chr.20: 33358865-33358865 on Build GRCh38
rs74646485 Chr.20: 33359230-33359230 on Build GRCh38
rs916030498 Chr.20: 33359220-33359220 on Build GRCh38
rs1057234974 Chr.20: 33358904-33358904 on Build GRCh38
rs762537973 Chr.20: 33359111-33359111 on Build GRCh38
rs759433834 Chr.20: 33359100-33359100 on Build GRCh38
rs1047703042 Chr.20: 33358883-33358883 on Build GRCh38
rs754917411 Chr.20: 33358858-33358858 on Build GRCh38
rs541763921 Chr.20: 33359130-33359130 on Build GRCh38
rs116917900 Chr.20: 33358814-33358814 on Build GRCh38
rs776837347 Chr.20: 33359183-33359183 on Build GRCh38
rs372962722 Chr.20: 33359168-33359168 on Build GRCh38

View Full Product Details