Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33662414-33662957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf144
Entrez Gene ID
128864
Gene Name
chromosome 20 open reading frame 144
Gene Aliases
dJ63M2.6
UniGene
Hs.324104
Gene Chromosome Location
Chr.20: 33662286-33663915 on Build GRCh38
Transcripts
NM_080825.3 NM_031231.3 NM_031232.3 XM_005260510.1 XM_011528992.2 XM_017028015.1

Gene Symbol
NECAB3
Entrez Gene ID
63941
Gene Name
N-terminal EF-hand calcium binding protein 3
Gene Aliases
APBA2BP, EFCBP3, NIP1, STIP3, SYTIP2, XB51, dJ63M2.4, dJ63M2.5
UniGene
Hs.516986
Gene Chromosome Location
Chr.20: 33657087-33674458 on Build GRCh38
Transcripts
NM_080825.3 NM_031231.3 NM_031232.3 XM_005260510.1 XM_011528992.2 XM_017028015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7341164 c.112C>G p.R38G Chr.20: 33662457-33662457 on Build GRCh38
5391862 c.87G>A p.W29* Chr.20: 33662432-33662432 on Build GRCh38
5415029 c.72G>A p.M24I Chr.20: 33662417-33662417 on Build GRCh38
6034567 c.112C>T p.R38W Chr.20: 33662457-33662457 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs532237280 Chr.20: 33662716-33662716 on Build GRCh38
rs747175789 Chr.20: 33662501-33662501 on Build GRCh38
rs891493215 Chr.20: 33662608-33662608 on Build GRCh38
rs574890015 Chr.20: 33662480-33662480 on Build GRCh38
rs1026059396 Chr.20: 33662757-33662757 on Build GRCh38
rs1010365434 Chr.20: 33662609-33662609 on Build GRCh38
rs918617506 Chr.20: 33662511-33662512 on Build GRCh38
rs979895180 Chr.20: 33662807-33662807 on Build GRCh38
rs1042194951 Chr.20: 33662849-33662849 on Build GRCh38
rs776573787 Chr.20: 33662844-33662844 on Build GRCh38
rs549973372 Chr.20: 33662579-33662579 on Build GRCh38
rs564819129 Chr.20: 33662615-33662615 on Build GRCh38
rs912848804 Chr.20: 33662903-33662904 on Build GRCh38
rs139136851 Chr.20: 33662795-33662795 on Build GRCh38
rs537064884 Chr.20: 33662718-33662718 on Build GRCh38
rs1053845470 Chr.20: 33662558-33662558 on Build GRCh38
rs111367221 Chr.20: 33662916-33662916 on Build GRCh38
rs529396770 Chr.20: 33662632-33662632 on Build GRCh38
rs1016649099 Chr.20: 33662686-33662686 on Build GRCh38
rs372626931 Chr.20: 33662485-33662485 on Build GRCh38
rs35422291 Chr.20: 33662658-33662659 on Build GRCh38
rs746812996 Chr.20: 33662814-33662814 on Build GRCh38
rs761579956 Chr.20: 33662437-33662437 on Build GRCh38
rs778131605 Chr.20: 33662497-33662497 on Build GRCh38
rs909320290 Chr.20: 33662452-33662452 on Build GRCh38
rs955531744 Chr.20: 33662457-33662457 on Build GRCh38
rs756165170 Chr.20: 33662513-33662513 on Build GRCh38
rs935915240 Chr.20: 33662860-33662860 on Build GRCh38
rs755424666 Chr.20: 33662462-33662462 on Build GRCh38
rs986165431 Chr.20: 33662678-33662678 on Build GRCh38
rs371434634 Chr.20: 33662875-33662875 on Build GRCh38
rs749472805 Chr.20: 33662585-33662585 on Build GRCh38
rs951421990 Chr.20: 33662771-33662771 on Build GRCh38
rs995811254 Chr.20: 33662740-33662740 on Build GRCh38
rs115573317 Chr.20: 33662559-33662559 on Build GRCh38
rs73259865 Chr.20: 33662883-33662883 on Build GRCh38
rs949591607 Chr.20: 33662752-33662752 on Build GRCh38
rs766909314 Chr.20: 33662438-33662438 on Build GRCh38
rs1028269707 Chr.20: 33662657-33662657 on Build GRCh38
rs889721765 Chr.20: 33662922-33662922 on Build GRCh38
rs1028548454 Chr.20: 33662901-33662901 on Build GRCh38
rs901043996 Chr.20: 33662649-33662649 on Build GRCh38
rs1053470815 Chr.20: 33662805-33662805 on Build GRCh38
rs942084610 Chr.20: 33662467-33662467 on Build GRCh38
rs755162719 Chr.20: 33662650-33662650 on Build GRCh38
rs770777155 Chr.20: 33662830-33662830 on Build GRCh38
rs927229033 Chr.20: 33662591-33662591 on Build GRCh38
rs954078803 Chr.20: 33662677-33662677 on Build GRCh38
rs971177670 Chr.20: 33662799-33662801 on Build GRCh38
rs937721996 Chr.20: 33662533-33662533 on Build GRCh38
rs905437456 Chr.20: 33662520-33662520 on Build GRCh38
rs900910017 Chr.20: 33662880-33662880 on Build GRCh38
rs919341706 Chr.20: 33662722-33662722 on Build GRCh38
rs1044270193 Chr.20: 33662586-33662586 on Build GRCh38
rs149331538 Chr.20: 33662505-33662505 on Build GRCh38
rs765671829 Chr.20: 33662468-33662468 on Build GRCh38
rs935964467 Chr.20: 33662603-33662603 on Build GRCh38
rs368297450 Chr.20: 33662479-33662479 on Build GRCh38
rs982232677 Chr.20: 33662765-33662765 on Build GRCh38
rs750019606 Chr.20: 33662458-33662458 on Build GRCh38
rs1020802266 Chr.20: 33662628-33662628 on Build GRCh38
rs926919618 Chr.20: 33662789-33662789 on Build GRCh38
rs376150007 Chr.20: 33662800-33662800 on Build GRCh38
rs945748677 Chr.20: 33662930-33662930 on Build GRCh38
rs769385455 Chr.20: 33662909-33662909 on Build GRCh38
rs1123311 Chr.20: 33662566-33662566 on Build GRCh38
rs1050866923 Chr.20: 33662643-33662643 on Build GRCh38

View Full Product Details