Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33663439-33663940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf144
Entrez Gene ID
128864
Gene Name
chromosome 20 open reading frame 144
Gene Aliases
dJ63M2.6
UniGene
Hs.324104
Gene Chromosome Location
Chr.20: 33662286-33663915 on Build GRCh38
Transcripts
NM_080825.3 NM_031231.3 NM_031232.3 XM_005260510.1 XM_011528992.2 XM_017028015.1

Gene Symbol
NECAB3
Entrez Gene ID
63941
Gene Name
N-terminal EF-hand calcium binding protein 3
Gene Aliases
APBA2BP, EFCBP3, NIP1, STIP3, SYTIP2, XB51, dJ63M2.4, dJ63M2.5
UniGene
Hs.516986
Gene Chromosome Location
Chr.20: 33657087-33674458 on Build GRCh38
Transcripts
NM_080825.3 NM_031231.3 NM_031232.3 XM_005260510.1 XM_011528992.2 XM_017028015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
724005 c.239T>G p.L80W Chr.20: 33663644-33663644 on Build GRCh38
7358365 c.296T>G p.L99R Chr.20: 33663701-33663701 on Build GRCh38
6588341 c.268G>C p.E90Q Chr.20: 33663673-33663673 on Build GRCh38
6564646 c.346C>G p.L116V Chr.20: 33663751-33663751 on Build GRCh38
7207872 c.319C>T p.P107S Chr.20: 33663724-33663724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755984673 Chr.20: 33663690-33663691 on Build GRCh38
rs1004271119 Chr.20: 33663893-33663894 on Build GRCh38
rs143664136 Chr.20: 33663702-33663716 on Build GRCh38
rs771766237 Chr.20: 33663566-33663566 on Build GRCh38
rs541781071 Chr.20: 33663817-33663817 on Build GRCh38
rs752655284 Chr.20: 33663698-33663698 on Build GRCh38
rs568762996 Chr.20: 33663522-33663522 on Build GRCh38
rs762277407 Chr.20: 33663665-33663665 on Build GRCh38
rs1022441012 Chr.20: 33663536-33663536 on Build GRCh38
rs539258366 Chr.20: 33663583-33663583 on Build GRCh38
rs776158482 Chr.20: 33663585-33663585 on Build GRCh38
rs557286657 Chr.20: 33663586-33663586 on Build GRCh38
rs994016919 Chr.20: 33663472-33663472 on Build GRCh38
rs774903996 Chr.20: 33663716-33663716 on Build GRCh38
rs772515500 Chr.20: 33663720-33663720 on Build GRCh38
rs755118770 Chr.20: 33663626-33663626 on Build GRCh38
rs758953045 Chr.20: 33663529-33663529 on Build GRCh38
rs755043465 Chr.20: 33663683-33663683 on Build GRCh38
rs763960798 Chr.20: 33663759-33663759 on Build GRCh38
rs746633278 Chr.20: 33663641-33663641 on Build GRCh38
rs975899211 Chr.20: 33663613-33663630 on Build GRCh38
rs946697016 Chr.20: 33663825-33663825 on Build GRCh38
rs7260921 Chr.20: 33663590-33663590 on Build GRCh38
rs761370319 Chr.20: 33663513-33663513 on Build GRCh38
rs1042447442 Chr.20: 33663813-33663813 on Build GRCh38
rs557872161 Chr.20: 33663755-33663755 on Build GRCh38
rs907114759 Chr.20: 33663681-33663681 on Build GRCh38
rs780334186 Chr.20: 33663554-33663554 on Build GRCh38
rs770725346 Chr.20: 33663707-33663707 on Build GRCh38
rs999688083 Chr.20: 33663713-33663713 on Build GRCh38
rs775530210 Chr.20: 33663493-33663493 on Build GRCh38
rs747915946 Chr.20: 33663565-33663565 on Build GRCh38
rs777476928 Chr.20: 33663640-33663640 on Build GRCh38
rs540858485 Chr.20: 33663777-33663777 on Build GRCh38
rs938258147 Chr.20: 33663768-33663768 on Build GRCh38
rs745544415 Chr.20: 33663712-33663712 on Build GRCh38
rs775297656 Chr.20: 33663622-33663622 on Build GRCh38
rs758340134 Chr.20: 33663758-33663758 on Build GRCh38
rs941096356 Chr.20: 33663650-33663650 on Build GRCh38
rs776103523 Chr.20: 33663654-33663654 on Build GRCh38
rs770635433 Chr.20: 33663652-33663652 on Build GRCh38
rs774418052 Chr.20: 33663659-33663660 on Build GRCh38
rs758981545 Chr.20: 33663657-33663657 on Build GRCh38
rs553421335 Chr.20: 33663912-33663912 on Build GRCh38
rs759722540 Chr.20: 33663741-33663741 on Build GRCh38
rs529965841 Chr.20: 33663782-33663782 on Build GRCh38
rs981221433 Chr.20: 33663769-33663769 on Build GRCh38
rs890531753 Chr.20: 33663822-33663822 on Build GRCh38
rs908677074 Chr.20: 33663655-33663655 on Build GRCh38
rs895231590 Chr.20: 33663702-33663702 on Build GRCh38
rs1002171229 Chr.20: 33663460-33663460 on Build GRCh38
rs766653945 Chr.20: 33663677-33663677 on Build GRCh38
rs754032272 Chr.20: 33663615-33663615 on Build GRCh38
rs772876354 Chr.20: 33663573-33663573 on Build GRCh38
rs760338918 Chr.20: 33663575-33663575 on Build GRCh38
rs764332862 Chr.20: 33663530-33663530 on Build GRCh38
rs1047730305 Chr.20: 33663473-33663473 on Build GRCh38
rs929898819 Chr.20: 33663673-33663673 on Build GRCh38
rs766879492 Chr.20: 33663726-33663726 on Build GRCh38
rs762812321 Chr.20: 33663587-33663591 on Build GRCh38
rs928558901 Chr.20: 33663788-33663788 on Build GRCh38
rs760969269 Chr.20: 33663676-33663676 on Build GRCh38
rs1055543381 Chr.20: 33663800-33663800 on Build GRCh38
rs571018419 Chr.20: 33663510-33663510 on Build GRCh38
rs774005860 Chr.20: 33663561-33663561 on Build GRCh38
rs774872819 Chr.20: 33663661-33663661 on Build GRCh38
rs761908441 Chr.20: 33663688-33663689 on Build GRCh38
rs750331410 Chr.20: 33663557-33663557 on Build GRCh38
rs756217324 Chr.20: 33663555-33663555 on Build GRCh38
rs767896556 Chr.20: 33663666-33663666 on Build GRCh38
rs1043868980 Chr.20: 33663839-33663839 on Build GRCh38
rs778877884 Chr.20: 33663686-33663686 on Build GRCh38
rs749263681 Chr.20: 33663495-33663495 on Build GRCh38
rs906505629 Chr.20: 33663486-33663486 on Build GRCh38
rs997208461 Chr.20: 33663730-33663730 on Build GRCh38
rs918334022 Chr.20: 33663761-33663761 on Build GRCh38
rs748062711 Chr.20: 33663633-33663633 on Build GRCh38
rs769325464 Chr.20: 33663714-33663714 on Build GRCh38
rs530434940 Chr.20: 33663879-33663879 on Build GRCh38
rs762199165 Chr.20: 33663595-33663595 on Build GRCh38
rs535273746 Chr.20: 33663520-33663520 on Build GRCh38
rs199763056 Chr.20: 33663757-33663757 on Build GRCh38
rs770000414 Chr.20: 33663488-33663488 on Build GRCh38
rs553699048 Chr.20: 33663521-33663521 on Build GRCh38
rs768623097 Chr.20: 33663648-33663649 on Build GRCh38
rs1019561582 Chr.20: 33663567-33663567 on Build GRCh38
rs908312010 Chr.20: 33663752-33663752 on Build GRCh38
rs1007110745 Chr.20: 33663902-33663902 on Build GRCh38
rs902588830 Chr.20: 33663847-33663847 on Build GRCh38
rs938287722 Chr.20: 33663689-33663691 on Build GRCh38
rs746862374 Chr.20: 33663703-33663703 on Build GRCh38
rs758306790 Chr.20: 33663699-33663699 on Build GRCh38
rs570613367 Chr.20: 33663514-33663514 on Build GRCh38
rs748688980 Chr.20: 33663718-33663718 on Build GRCh38
rs760107824 Chr.20: 33663518-33663518 on Build GRCh38
rs559209641 Chr.20: 33663781-33663781 on Build GRCh38
rs777539750 Chr.20: 33663701-33663701 on Build GRCh38
rs757577041 Chr.20: 33663533-33663533 on Build GRCh38
rs1008078035 Chr.20: 33663708-33663708 on Build GRCh38
rs895107611 Chr.20: 33663823-33663823 on Build GRCh38
rs945388532 Chr.20: 33663805-33663805 on Build GRCh38
rs750655093 Chr.20: 33663609-33663609 on Build GRCh38
rs1017910420 Chr.20: 33663743-33663743 on Build GRCh38
rs746108951 Chr.20: 33663550-33663550 on Build GRCh38
rs937242793 Chr.20: 33663789-33663789 on Build GRCh38
rs763444244 Chr.20: 33663588-33663588 on Build GRCh38
rs781410258 Chr.20: 33663543-33663543 on Build GRCh38
rs776940228 Chr.20: 33663736-33663736 on Build GRCh38
rs899703295 Chr.20: 33663834-33663834 on Build GRCh38
rs1015234902 Chr.20: 33663899-33663899 on Build GRCh38
rs918386274 Chr.20: 33663667-33663667 on Build GRCh38
rs767796205 Chr.20: 33663606-33663606 on Build GRCh38
rs955633227 Chr.20: 33663732-33663732 on Build GRCh38
rs780785358 Chr.20: 33663710-33663710 on Build GRCh38
rs552305229 Chr.20: 33663504-33663504 on Build GRCh38
rs756999476 Chr.20: 33663801-33663801 on Build GRCh38
rs768782763 Chr.20: 33663559-33663559 on Build GRCh38
rs756215436 Chr.20: 33663552-33663552 on Build GRCh38
rs188273385 Chr.20: 33663827-33663827 on Build GRCh38
rs781014497 Chr.20: 33663861-33663861 on Build GRCh38
rs776946706 Chr.20: 33663582-33663611 on Build GRCh38
rs545844490 Chr.20: 33663634-33663634 on Build GRCh38
rs754114109 Chr.20: 33663678-33663678 on Build GRCh38
rs778841612 Chr.20: 33663630-33663630 on Build GRCh38
rs773872225 Chr.20: 33663722-33663722 on Build GRCh38
rs534852781 Chr.20: 33663883-33663883 on Build GRCh38
rs1054452095 Chr.20: 33663791-33663791 on Build GRCh38
rs750889799 Chr.20: 33663672-33663672 on Build GRCh38
rs1020107727 Chr.20: 33663494-33663494 on Build GRCh38
rs893833767 Chr.20: 33663695-33663695 on Build GRCh38
rs915206130 Chr.20: 33663802-33663802 on Build GRCh38
rs368305380 Chr.20: 33663623-33663623 on Build GRCh38
rs939371624 Chr.20: 33663688-33663688 on Build GRCh38
rs756448795 Chr.20: 33663610-33663610 on Build GRCh38
rs751530722 Chr.20: 33663767-33663767 on Build GRCh38
rs765328646 Chr.20: 33663744-33663744 on Build GRCh38
rs982224651 Chr.20: 33663764-33663764 on Build GRCh38
rs769219793 Chr.20: 33663658-33663658 on Build GRCh38
rs1040898934 Chr.20: 33663829-33663829 on Build GRCh38
rs745903315 Chr.20: 33663481-33663481 on Build GRCh38
rs753894540 Chr.20: 33663723-33663723 on Build GRCh38

View Full Product Details