Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33850868-33851371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP4B
Entrez Gene ID
128866
Gene Name
charged multivesicular body protein 4B
Gene Aliases
C20orf178, CHMP4A, CTPP3, CTRCT31, SNF7, SNF7-2, Shax1, VPS32B, Vps32-2, dJ553F4.4
UniGene
Hs.472471
Gene Chromosome Location
Chr.20: 33811304-33854367 on Build GRCh38
Transcripts
NM_176812.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672186 c.426A>G p.A142A Chr.20: 33851009-33851009 on Build GRCh38
274539 c.450G>A p.S150S Chr.20: 33851033-33851033 on Build GRCh38
723989 c.466G>C p.G156R Chr.20: 33851049-33851049 on Build GRCh38
723990 c.370G>C p.D124H Chr.20: 33850953-33850953 on Build GRCh38
5007383 c.481G>A p.E161K Chr.20: 33851064-33851064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878919488 Chr.20: 33851094-33851094 on Build GRCh38
rs201956567 Chr.20: 33851073-33851073 on Build GRCh38
rs377633958 Chr.20: 33851043-33851043 on Build GRCh38
rs745354594 Chr.20: 33850907-33850907 on Build GRCh38
rs776519557 Chr.20: 33851089-33851089 on Build GRCh38
rs747159062 Chr.20: 33851077-33851077 on Build GRCh38
rs1024243530 Chr.20: 33851141-33851141 on Build GRCh38
rs751110777 Chr.20: 33851039-33851039 on Build GRCh38
rs542431187 Chr.20: 33851231-33851231 on Build GRCh38
rs765293644 Chr.20: 33850925-33850926 on Build GRCh38
rs776803113 Chr.20: 33850958-33850958 on Build GRCh38
rs368007253 Chr.20: 33850919-33850919 on Build GRCh38
rs755457066 Chr.20: 33851060-33851060 on Build GRCh38
rs745999954 Chr.20: 33851101-33851101 on Build GRCh38
rs763982258 Chr.20: 33851037-33851037 on Build GRCh38
rs1026146855 Chr.20: 33851081-33851081 on Build GRCh38
rs766901041 Chr.20: 33851044-33851044 on Build GRCh38
rs979769840 Chr.20: 33851160-33851160 on Build GRCh38
rs771223331 Chr.20: 33851079-33851079 on Build GRCh38
rs912957999 Chr.20: 33851260-33851260 on Build GRCh38
rs534411001 Chr.20: 33851170-33851170 on Build GRCh38
rs865783046 Chr.20: 33851337-33851337 on Build GRCh38
rs775409766 Chr.20: 33851022-33851022 on Build GRCh38
rs118203966 Chr.20: 33851064-33851064 on Build GRCh38
rs779221149 Chr.20: 33850924-33850924 on Build GRCh38
rs886298704 Chr.20: 33851093-33851093 on Build GRCh38
rs972735356 Chr.20: 33851080-33851080 on Build GRCh38
rs778222288 Chr.20: 33851076-33851076 on Build GRCh38
rs989938581 Chr.20: 33851255-33851255 on Build GRCh38
rs373891576 Chr.20: 33850934-33850934 on Build GRCh38
rs759096292 Chr.20: 33851117-33851117 on Build GRCh38
rs765312860 Chr.20: 33851012-33851012 on Build GRCh38
rs971281815 Chr.20: 33851149-33851149 on Build GRCh38
rs748507989 Chr.20: 33851066-33851066 on Build GRCh38
rs548441020 Chr.20: 33850908-33850908 on Build GRCh38
rs780770988 Chr.20: 33850918-33850918 on Build GRCh38
rs917223756 Chr.20: 33851119-33851119 on Build GRCh38
rs118203965 Chr.20: 33850969-33850969 on Build GRCh38
rs563155854 Chr.20: 33850986-33850986 on Build GRCh38
rs769847123 Chr.20: 33851104-33851104 on Build GRCh38
rs776549014 Chr.20: 33851082-33851082 on Build GRCh38
rs148104800 Chr.20: 33851293-33851293 on Build GRCh38
rs770867018 Chr.20: 33850943-33850943 on Build GRCh38
rs773491442 Chr.20: 33850931-33850931 on Build GRCh38
rs946231232 Chr.20: 33851277-33851277 on Build GRCh38
rs942721583 Chr.20: 33851004-33851004 on Build GRCh38
rs535801835 Chr.20: 33851323-33851323 on Build GRCh38
rs570167861 Chr.20: 33851161-33851161 on Build GRCh38
rs376307592 Chr.20: 33850930-33850930 on Build GRCh38
rs146262818 Chr.20: 33851033-33851033 on Build GRCh38
rs769372339 Chr.20: 33851284-33851284 on Build GRCh38
rs772270578 Chr.20: 33850927-33850927 on Build GRCh38
rs1004706515 Chr.20: 33851105-33851105 on Build GRCh38
rs551868035 Chr.20: 33851063-33851063 on Build GRCh38
rs200647887 Chr.20: 33850906-33850906 on Build GRCh38
rs375126082 Chr.20: 33850926-33850926 on Build GRCh38
rs775174195 Chr.20: 33851144-33851144 on Build GRCh38
rs892051007 Chr.20: 33851314-33851314 on Build GRCh38
rs140995439 Chr.20: 33851344-33851344 on Build GRCh38
rs775497905 Chr.20: 33851116-33851116 on Build GRCh38
rs749993051 Chr.20: 33851051-33851051 on Build GRCh38
rs371861453 Chr.20: 33850939-33850939 on Build GRCh38

View Full Product Details