Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33853768-33854308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP4B
Entrez Gene ID
128866
Gene Name
charged multivesicular body protein 4B
Gene Aliases
C20orf178, CHMP4A, CTPP3, CTRCT31, SNF7, SNF7-2, Shax1, VPS32B, Vps32-2, dJ553F4.4
UniGene
Hs.472471
Gene Chromosome Location
Chr.20: 33811304-33854367 on Build GRCh38
Transcripts
NM_176812.4

SNP Detail

SNP ID Chromosome Location
rs780973758 Chr.20: 33853917-33853917 on Build GRCh38
rs894841725 Chr.20: 33854126-33854126 on Build GRCh38
rs558747282 Chr.20: 33853794-33853794 on Build GRCh38
rs955222375 Chr.20: 33854230-33854230 on Build GRCh38
rs896976805 Chr.20: 33854144-33854144 on Build GRCh38
rs1021881362 Chr.20: 33854153-33854153 on Build GRCh38
rs990622518 Chr.20: 33853878-33853878 on Build GRCh38
rs1038180352 Chr.20: 33854128-33854128 on Build GRCh38
rs943377593 Chr.20: 33853893-33853893 on Build GRCh38
rs190986165 Chr.20: 33853865-33853865 on Build GRCh38
rs574387269 Chr.20: 33854263-33854263 on Build GRCh38
rs887820812 Chr.20: 33854037-33854037 on Build GRCh38
rs977787599 Chr.20: 33854204-33854204 on Build GRCh38
rs140148563 Chr.20: 33853888-33853888 on Build GRCh38
rs961260182 Chr.20: 33853835-33853835 on Build GRCh38
rs187600351 Chr.20: 33853796-33853796 on Build GRCh38
rs933919934 Chr.20: 33853879-33853879 on Build GRCh38
rs1033925746 Chr.20: 33854106-33854106 on Build GRCh38
rs972404204 Chr.20: 33853851-33853851 on Build GRCh38
rs765492016 Chr.20: 33853856-33853856 on Build GRCh38
rs182495227 Chr.20: 33854157-33854157 on Build GRCh38
rs906680974 Chr.20: 33854023-33854023 on Build GRCh38
rs1005403246 Chr.20: 33854088-33854088 on Build GRCh38
rs968175161 Chr.20: 33854277-33854277 on Build GRCh38
rs1010528803 Chr.20: 33854149-33854149 on Build GRCh38
rs143881004 Chr.20: 33854130-33854130 on Build GRCh38
rs540944582 Chr.20: 33853951-33853951 on Build GRCh38
rs916306135 Chr.20: 33853887-33853887 on Build GRCh38
rs931968695 Chr.20: 33854018-33854018 on Build GRCh38
rs758583001 Chr.20: 33853916-33853916 on Build GRCh38
rs977902164 Chr.20: 33853859-33853859 on Build GRCh38
rs1034930904 Chr.20: 33854173-33854173 on Build GRCh38
rs1021995351 Chr.20: 33853800-33853800 on Build GRCh38
rs960436611 Chr.20: 33854180-33854180 on Build GRCh38
rs935616459 Chr.20: 33853948-33853948 on Build GRCh38
rs1004228577 Chr.20: 33854099-33854099 on Build GRCh38
rs572521901 Chr.20: 33853924-33853924 on Build GRCh38
rs754983268 Chr.20: 33853852-33853852 on Build GRCh38
rs11542525 Chr.20: 33854165-33854165 on Build GRCh38
rs543322188 Chr.20: 33854047-33854047 on Build GRCh38
rs371058569 Chr.20: 33853974-33853974 on Build GRCh38
rs979139695 Chr.20: 33854281-33854281 on Build GRCh38
rs545110394 Chr.20: 33853883-33853883 on Build GRCh38
rs1012289696 Chr.20: 33854222-33854222 on Build GRCh38
rs779848302 Chr.20: 33854220-33854220 on Build GRCh38
rs924210428 Chr.20: 33853931-33853931 on Build GRCh38

View Full Product Details