Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34851139-34851642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT7
Entrez Gene ID
2686
Gene Name
gamma-glutamyltransferase 7
Gene Aliases
D20S101, GGT4, GGTL3, GGTL5, dJ18C9.2
UniGene
Hs.433738
Gene Chromosome Location
Chr.20: 34844720-34872858 on Build GRCh38
Transcripts
NM_178026.2 XM_011528782.2 XM_011528783.2 XM_017027809.1 XM_017027810.1 XM_017027811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7241105 c.1675C>T p.L559F Chr.20: 34851281-34851281 on Build GRCh38
4291457 c.1677C>T p.L559L Chr.20: 34851279-34851279 on Build GRCh38
5057250 c.1650C>T p.P550P Chr.20: 34851306-34851306 on Build GRCh38
1411332 c.1652C>T p.A551V Chr.20: 34851304-34851304 on Build GRCh38
3972523 c.1701G>A p.A567A Chr.20: 34851255-34851255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368581486 Chr.20: 34851219-34851219 on Build GRCh38
rs868640488 Chr.20: 34851342-34851342 on Build GRCh38
rs960908355 Chr.20: 34851497-34851497 on Build GRCh38
rs371319730 Chr.20: 34851271-34851271 on Build GRCh38
rs768285988 Chr.20: 34851276-34851276 on Build GRCh38
rs776008502 Chr.20: 34851419-34851419 on Build GRCh38
rs1035268863 Chr.20: 34851493-34851493 on Build GRCh38
rs1017122351 Chr.20: 34851455-34851455 on Build GRCh38
rs979571447 Chr.20: 34851569-34851569 on Build GRCh38
rs973042171 Chr.20: 34851475-34851475 on Build GRCh38
rs540658672 Chr.20: 34851466-34851466 on Build GRCh38
rs375222269 Chr.20: 34851196-34851196 on Build GRCh38
rs760190557 Chr.20: 34851406-34851406 on Build GRCh38
rs761161712 Chr.20: 34851278-34851278 on Build GRCh38
rs772751695 Chr.20: 34851399-34851399 on Build GRCh38
rs780921464 Chr.20: 34851255-34851255 on Build GRCh38
rs781210882 Chr.20: 34851316-34851316 on Build GRCh38
rs961830119 Chr.20: 34851467-34851467 on Build GRCh38
rs745841534 Chr.20: 34851344-34851344 on Build GRCh38
rs1048391869 Chr.20: 34851427-34851427 on Build GRCh38
rs563504697 Chr.20: 34851608-34851608 on Build GRCh38
rs758665316 Chr.20: 34851243-34851243 on Build GRCh38
rs761408666 Chr.20: 34851394-34851394 on Build GRCh38
rs36084138 Chr.20: 34851282-34851283 on Build GRCh38
rs773835971 Chr.20: 34851392-34851392 on Build GRCh38
rs1022217612 Chr.20: 34851546-34851546 on Build GRCh38
rs1055216256 Chr.20: 34851495-34851495 on Build GRCh38
rs757240415 Chr.20: 34851310-34851310 on Build GRCh38
rs762471563 Chr.20: 34851198-34851198 on Build GRCh38
rs199778791 Chr.20: 34851220-34851220 on Build GRCh38
rs775662218 Chr.20: 34851260-34851260 on Build GRCh38
rs371027305 Chr.20: 34851256-34851256 on Build GRCh38
rs377674792 Chr.20: 34851225-34851225 on Build GRCh38
rs749463286 Chr.20: 34851399-34851400 on Build GRCh38
rs6060083 Chr.20: 34851599-34851599 on Build GRCh38
rs560041425 Chr.20: 34851529-34851529 on Build GRCh38
rs183946130 Chr.20: 34851448-34851448 on Build GRCh38
rs576222899 Chr.20: 34851306-34851306 on Build GRCh38
rs756028742 Chr.20: 34851566-34851566 on Build GRCh38
rs543513705 Chr.20: 34851376-34851376 on Build GRCh38
rs760102754 Chr.20: 34851294-34851294 on Build GRCh38
rs753022197 Chr.20: 34851242-34851242 on Build GRCh38
rs753110256 Chr.20: 34851303-34851303 on Build GRCh38
rs530895296 Chr.20: 34851197-34851197 on Build GRCh38
rs762633607 Chr.20: 34851267-34851267 on Build GRCh38
rs768304327 Chr.20: 34851390-34851390 on Build GRCh38
rs765829550 Chr.20: 34851411-34851411 on Build GRCh38
rs769687416 Chr.20: 34851366-34851366 on Build GRCh38
rs765739734 Chr.20: 34851300-34851300 on Build GRCh38
rs200653688 Chr.20: 34851305-34851305 on Build GRCh38
rs542051381 Chr.20: 34851595-34851595 on Build GRCh38
rs565836736 Chr.20: 34851279-34851279 on Build GRCh38
rs999463547 Chr.20: 34851570-34851570 on Build GRCh38
rs776545181 Chr.20: 34851191-34851191 on Build GRCh38
rs371365087 Chr.20: 34851420-34851420 on Build GRCh38
rs34687214 Chr.20: 34851253-34851253 on Build GRCh38
rs114477173 Chr.20: 34851164-34851164 on Build GRCh38
rs755259069 Chr.20: 34851234-34851234 on Build GRCh38
rs374372409 Chr.20: 34851265-34851265 on Build GRCh38
rs763641807 Chr.20: 34851200-34851200 on Build GRCh38
rs1026397975 Chr.20: 34851175-34851175 on Build GRCh38
rs773928220 Chr.20: 34851277-34851277 on Build GRCh38
rs567777161 Chr.20: 34851567-34851567 on Build GRCh38
rs955237323 Chr.20: 34851607-34851607 on Build GRCh38
rs1040310136 Chr.20: 34851592-34851592 on Build GRCh38
rs750179813 Chr.20: 34851499-34851499 on Build GRCh38
rs201808310 Chr.20: 34851304-34851304 on Build GRCh38
rs966639966 Chr.20: 34851548-34851548 on Build GRCh38
rs779304038 Chr.20: 34851239-34851239 on Build GRCh38
rs200359546 Chr.20: 34851207-34851207 on Build GRCh38
rs779889802 Chr.20: 34851371-34851371 on Build GRCh38
rs200306952 Chr.20: 34851398-34851398 on Build GRCh38
rs574770592 Chr.20: 34851547-34851547 on Build GRCh38
rs770766821 Chr.20: 34851184-34851184 on Build GRCh38
rs759191258 Chr.20: 34851192-34851192 on Build GRCh38
rs778069415 Chr.20: 34851249-34851249 on Build GRCh38
rs754254364 Chr.20: 34851287-34851287 on Build GRCh38
rs538566828 Chr.20: 34851254-34851254 on Build GRCh38
rs1029887785 Chr.20: 34851598-34851598 on Build GRCh38

View Full Product Details