Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34859884-34860374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT7
Entrez Gene ID
2686
Gene Name
gamma-glutamyltransferase 7
Gene Aliases
D20S101, GGT4, GGTL3, GGTL5, dJ18C9.2
UniGene
Hs.433738
Gene Chromosome Location
Chr.20: 34844720-34872858 on Build GRCh38
Transcripts
NM_178026.2 XM_011528782.2 XM_011528783.2 XM_017027809.1 XM_017027810.1 XM_017027811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5343580 c.727C>T p.H243Y Chr.20: 34860270-34860270 on Build GRCh38
3693448 c.737A>G p.Y246C Chr.20: 34860260-34860260 on Build GRCh38
4820302 c.701G>A p.G234E Chr.20: 34860296-34860296 on Build GRCh38
5391886 c.755C>T p.S252F Chr.20: 34860031-34860031 on Build GRCh38
5857727 c.784delG p.A262fs*7 Chr.20: 34860002-34860002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047086826 Chr.20: 34860100-34860100 on Build GRCh38
rs764688866 Chr.20: 34860298-34860298 on Build GRCh38
rs756418112 Chr.20: 34860333-34860333 on Build GRCh38
rs774875253 Chr.20: 34860222-34860223 on Build GRCh38
rs781600276 Chr.20: 34860215-34860215 on Build GRCh38
rs896085411 Chr.20: 34860106-34860106 on Build GRCh38
rs373147980 Chr.20: 34860039-34860039 on Build GRCh38
rs372362464 Chr.20: 34860260-34860260 on Build GRCh38
rs780252630 Chr.20: 34860030-34860030 on Build GRCh38
rs770050502 Chr.20: 34860024-34860024 on Build GRCh38
rs887197990 Chr.20: 34860105-34860105 on Build GRCh38
rs140622797 Chr.20: 34860262-34860262 on Build GRCh38
rs1005105513 Chr.20: 34860010-34860010 on Build GRCh38
rs181027490 Chr.20: 34860329-34860329 on Build GRCh38
rs765851466 Chr.20: 34859936-34859936 on Build GRCh38
rs369642023 Chr.20: 34859976-34859976 on Build GRCh38
rs573361809 Chr.20: 34860182-34860182 on Build GRCh38
rs1051947500 Chr.20: 34860007-34860007 on Build GRCh38
rs780340344 Chr.20: 34860220-34860220 on Build GRCh38
rs761474589 Chr.20: 34860047-34860047 on Build GRCh38
rs867920822 Chr.20: 34859975-34859975 on Build GRCh38
rs563588128 Chr.20: 34860354-34860354 on Build GRCh38
rs753329053 Chr.20: 34860094-34860094 on Build GRCh38
rs899099536 Chr.20: 34860088-34860088 on Build GRCh38
rs757517380 Chr.20: 34860214-34860214 on Build GRCh38
rs768702871 Chr.20: 34860045-34860045 on Build GRCh38
rs757509705 Chr.20: 34860003-34860003 on Build GRCh38
rs765937777 Chr.20: 34860092-34860092 on Build GRCh38
rs749322972 Chr.20: 34860222-34860222 on Build GRCh38
rs1034616607 Chr.20: 34860099-34860099 on Build GRCh38
rs369384619 Chr.20: 34860219-34860219 on Build GRCh38
rs866049288 Chr.20: 34859996-34859996 on Build GRCh38
rs769280181 Chr.20: 34860056-34860099 on Build GRCh38
rs781234854 Chr.20: 34860005-34860005 on Build GRCh38
rs8183445 Chr.20: 34860238-34860238 on Build GRCh38
rs745986271 Chr.20: 34860018-34860018 on Build GRCh38
rs6060095 Chr.20: 34860286-34860286 on Build GRCh38
rs896653663 Chr.20: 34860036-34860036 on Build GRCh38
rs774771343 Chr.20: 34860310-34860310 on Build GRCh38
rs753993169 Chr.20: 34860340-34860340 on Build GRCh38
rs968513634 Chr.20: 34860171-34860171 on Build GRCh38
rs979542803 Chr.20: 34860178-34860178 on Build GRCh38
rs919789984 Chr.20: 34860301-34860301 on Build GRCh38
rs768745642 Chr.20: 34860224-34860224 on Build GRCh38
rs750652638 Chr.20: 34860216-34860216 on Build GRCh38
rs375312421 Chr.20: 34859952-34859952 on Build GRCh38
rs764521693 Chr.20: 34860208-34860208 on Build GRCh38
rs778665554 Chr.20: 34860229-34860229 on Build GRCh38
rs1002395332 Chr.20: 34860192-34860194 on Build GRCh38
rs6142253 Chr.20: 34860338-34860338 on Build GRCh38
rs751810098 Chr.20: 34859994-34859994 on Build GRCh38
rs759112188 Chr.20: 34860044-34860045 on Build GRCh38
rs750788716 Chr.20: 34860332-34860332 on Build GRCh38
rs762354131 Chr.20: 34860325-34860325 on Build GRCh38
rs575789722 Chr.20: 34860103-34860103 on Build GRCh38
rs956737987 Chr.20: 34860198-34860198 on Build GRCh38
rs766643384 Chr.20: 34860334-34860334 on Build GRCh38
rs558000732 Chr.20: 34860175-34860175 on Build GRCh38
rs1023818222 Chr.20: 34860132-34860132 on Build GRCh38
rs771777366 Chr.20: 34860051-34860051 on Build GRCh38
rs776119649 Chr.20: 34860293-34860293 on Build GRCh38
rs1004508825 Chr.20: 34860079-34860079 on Build GRCh38
rs758899786 Chr.20: 34859957-34859957 on Build GRCh38
rs759010368 Chr.20: 34860297-34860297 on Build GRCh38
rs746719350 Chr.20: 34860271-34860271 on Build GRCh38
rs774209627 Chr.20: 34860046-34860046 on Build GRCh38
rs376736314 Chr.20: 34860200-34860200 on Build GRCh38
rs770508867 Chr.20: 34860283-34860283 on Build GRCh38
rs748094426 Chr.20: 34860252-34860252 on Build GRCh38
rs879023714 Chr.20: 34860314-34860314 on Build GRCh38
rs763460437 Chr.20: 34860207-34860207 on Build GRCh38
rs760341678 Chr.20: 34860062-34860062 on Build GRCh38
rs201952715 Chr.20: 34860345-34860345 on Build GRCh38
rs115343976 Chr.20: 34860213-34860213 on Build GRCh38
rs2295094 Chr.20: 34860112-34860112 on Build GRCh38
rs779017563 Chr.20: 34860351-34860351 on Build GRCh38
rs772787544 Chr.20: 34860052-34860052 on Build GRCh38

View Full Product Details