Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35142249-35142759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDEM2
Entrez Gene ID
55741
Gene Name
ER degradation enhancing alpha-mannosidase like protein 2
Gene Aliases
C20orf31, C20orf49, bA4204.1
UniGene
Hs.720177
Gene Chromosome Location
Chr.20: 35115357-35147358 on Build GRCh38
Transcripts
NM_001145025.1 NM_018217.2 NR_026728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4606636 c.191T>C p.V64A Chr.20: 35142423-35142423 on Build GRCh38
4186569 c.350A>T p.E117V Chr.20: 35142387-35142387 on Build GRCh38
4186570 c.227A>T p.E76V Chr.20: 35142387-35142387 on Build GRCh38
4186571 c.349G>T p.E117* Chr.20: 35142388-35142388 on Build GRCh38
4186572 c.226G>T p.E76* Chr.20: 35142388-35142388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771952256 Chr.20: 35142371-35142371 on Build GRCh38
rs6120849 Chr.20: 35142584-35142584 on Build GRCh38
rs750805682 Chr.20: 35142341-35142341 on Build GRCh38
rs773185360 Chr.20: 35142434-35142435 on Build GRCh38
rs371938796 Chr.20: 35142521-35142521 on Build GRCh38
rs373749622 Chr.20: 35142358-35142358 on Build GRCh38
rs764977629 Chr.20: 35142406-35142406 on Build GRCh38
rs532778574 Chr.20: 35142391-35142391 on Build GRCh38
rs761308105 Chr.20: 35142517-35142517 on Build GRCh38
rs184063388 Chr.20: 35142494-35142494 on Build GRCh38
rs780214317 Chr.20: 35142349-35142349 on Build GRCh38
rs755229780 Chr.20: 35142443-35142443 on Build GRCh38
rs781124804 Chr.20: 35142704-35142704 on Build GRCh38
rs763795566 Chr.20: 35142549-35142549 on Build GRCh38
rs776512001 Chr.20: 35142392-35142392 on Build GRCh38
rs748268998 Chr.20: 35142447-35142447 on Build GRCh38
rs145409721 Chr.20: 35142646-35142646 on Build GRCh38
rs775184458 Chr.20: 35142411-35142411 on Build GRCh38
rs758534825 Chr.20: 35142450-35142450 on Build GRCh38
rs141998071 Chr.20: 35142385-35142385 on Build GRCh38
rs1025348773 Chr.20: 35142701-35142701 on Build GRCh38
rs980703414 Chr.20: 35142510-35142510 on Build GRCh38
rs189031450 Chr.20: 35142497-35142497 on Build GRCh38
rs759321315 Chr.20: 35142404-35142404 on Build GRCh38
rs538872790 Chr.20: 35142653-35142653 on Build GRCh38
rs540044847 Chr.20: 35142356-35142356 on Build GRCh38
rs144209865 Chr.20: 35142424-35142424 on Build GRCh38
rs748195066 Chr.20: 35142364-35142364 on Build GRCh38
rs754249409 Chr.20: 35142526-35142526 on Build GRCh38
rs776635644 Chr.20: 35142493-35142493 on Build GRCh38
rs946047765 Chr.20: 35142614-35142614 on Build GRCh38
rs1019039440 Chr.20: 35142499-35142499 on Build GRCh38
rs760773345 Chr.20: 35142487-35142487 on Build GRCh38
rs892524081 Chr.20: 35142377-35142377 on Build GRCh38
rs773717513 Chr.20: 35142511-35142511 on Build GRCh38
rs140217956 Chr.20: 35142446-35142446 on Build GRCh38
rs1025092041 Chr.20: 35142721-35142721 on Build GRCh38
rs147639867 Chr.20: 35142381-35142381 on Build GRCh38
rs142283407 Chr.20: 35142732-35142732 on Build GRCh38
rs995596868 Chr.20: 35142694-35142694 on Build GRCh38
rs375577548 Chr.20: 35142360-35142360 on Build GRCh38
rs763671919 Chr.20: 35142500-35142500 on Build GRCh38
rs962119178 Chr.20: 35142275-35142275 on Build GRCh38
rs997048963 Chr.20: 35142481-35142481 on Build GRCh38
rs369596170 Chr.20: 35142335-35142335 on Build GRCh38
rs149129007 Chr.20: 35142705-35142705 on Build GRCh38
rs1056887525 Chr.20: 35142372-35142372 on Build GRCh38
rs971892866 Chr.20: 35142279-35142279 on Build GRCh38
rs3746427 Chr.20: 35142661-35142661 on Build GRCh38
rs1018676402 Chr.20: 35142271-35142271 on Build GRCh38
rs978051693 Chr.20: 35142436-35142436 on Build GRCh38
rs917861015 Chr.20: 35142505-35142505 on Build GRCh38
rs891853190 Chr.20: 35142604-35142605 on Build GRCh38
rs770765592 Chr.20: 35142388-35142388 on Build GRCh38
rs777646292 Chr.20: 35142452-35142452 on Build GRCh38
rs891347972 Chr.20: 35142370-35142370 on Build GRCh38
rs369753267 Chr.20: 35142421-35142421 on Build GRCh38
rs571429549 Chr.20: 35142650-35142650 on Build GRCh38
rs936515623 Chr.20: 35142551-35142551 on Build GRCh38
rs756043577 Chr.20: 35142713-35142713 on Build GRCh38
rs370930437 Chr.20: 35142496-35142496 on Build GRCh38
rs891841745 Chr.20: 35142303-35142303 on Build GRCh38
rs763466253 Chr.20: 35142414-35142414 on Build GRCh38
rs554212872 Chr.20: 35142731-35142731 on Build GRCh38
rs766916281 Chr.20: 35142495-35142495 on Build GRCh38
rs565408534 Chr.20: 35142293-35142293 on Build GRCh38
rs151130172 Chr.20: 35142425-35142425 on Build GRCh38
rs947938828 Chr.20: 35142509-35142509 on Build GRCh38
rs570069875 Chr.20: 35142575-35142575 on Build GRCh38
rs150074155 Chr.20: 35142527-35142527 on Build GRCh38
rs920436570 Chr.20: 35142333-35142333 on Build GRCh38
rs1013220194 Chr.20: 35142708-35142708 on Build GRCh38
rs553674967 Chr.20: 35142656-35142656 on Build GRCh38
rs761344824 Chr.20: 35142298-35142298 on Build GRCh38
rs756526754 Chr.20: 35142344-35142344 on Build GRCh38
rs762505697 Chr.20: 35142413-35142413 on Build GRCh38
rs376269911 Chr.20: 35142468-35142468 on Build GRCh38
rs556734937 Chr.20: 35142375-35142375 on Build GRCh38
rs375028628 Chr.20: 35142464-35142464 on Build GRCh38
rs1051063579 Chr.20: 35142634-35142634 on Build GRCh38
rs375436239 Chr.20: 35142337-35142337 on Build GRCh38
rs756517367 Chr.20: 35142423-35142423 on Build GRCh38

View Full Product Details