Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35437599-35438103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF5
Entrez Gene ID
8200
Gene Name
growth differentiation factor 5
Gene Aliases
BDA1C, BMP-14, BMP14, CDMP1, LAP-4, LAP4, OS5, SYM1B, SYNS2
UniGene
Hs.1573
Gene Chromosome Location
Chr.20: 35433347-35454746 on Build GRCh38
Transcripts
NM_000557.4 NM_001319138.1 XM_011529075.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1615509 c.192C>A p.S64R Chr.20: 35437737-35437737 on Build GRCh38
1615508 c.297C>A p.P99P Chr.20: 35437632-35437632 on Build GRCh38
6670218 c.250G>T p.G84* Chr.20: 35437679-35437679 on Build GRCh38
723912 c.322C>A p.P108T Chr.20: 35437607-35437607 on Build GRCh38
5741847 c.308G>A p.G103D Chr.20: 35437621-35437621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142244310 Chr.20: 35437777-35437777 on Build GRCh38
rs752622490 Chr.20: 35437772-35437773 on Build GRCh38
rs886042462 Chr.20: 35437771-35437771 on Build GRCh38
rs754670993 Chr.20: 35437732-35437732 on Build GRCh38
rs763641226 Chr.20: 35437927-35437927 on Build GRCh38
rs373973964 Chr.20: 35437904-35437904 on Build GRCh38
rs748012215 Chr.20: 35437674-35437674 on Build GRCh38
rs754422467 Chr.20: 35437956-35437956 on Build GRCh38
rs150002208 Chr.20: 35437627-35437627 on Build GRCh38
rs781718444 Chr.20: 35437680-35437680 on Build GRCh38
rs960967052 Chr.20: 35438025-35438025 on Build GRCh38
rs371564366 Chr.20: 35437765-35437765 on Build GRCh38
rs111902988 Chr.20: 35437773-35437773 on Build GRCh38
rs767477220 Chr.20: 35437632-35437632 on Build GRCh38
rs776550586 Chr.20: 35437894-35437894 on Build GRCh38
rs537885107 Chr.20: 35438000-35438000 on Build GRCh38
rs754587274 Chr.20: 35437655-35437655 on Build GRCh38
rs916896047 Chr.20: 35437715-35437715 on Build GRCh38
rs768978933 Chr.20: 35437703-35437703 on Build GRCh38
rs763690438 Chr.20: 35437792-35437792 on Build GRCh38
rs184304612 Chr.20: 35437665-35437665 on Build GRCh38
rs749564274 Chr.20: 35437694-35437694 on Build GRCh38
rs757930848 Chr.20: 35437745-35437745 on Build GRCh38
rs752406359 Chr.20: 35437778-35437778 on Build GRCh38
rs545542357 Chr.20: 35438075-35438075 on Build GRCh38
rs755912564 Chr.20: 35437634-35437634 on Build GRCh38
rs776465124 Chr.20: 35437776-35437776 on Build GRCh38
rs375024998 Chr.20: 35437958-35437958 on Build GRCh38
rs374059504 Chr.20: 35437724-35437724 on Build GRCh38
rs867399514 Chr.20: 35437830-35437830 on Build GRCh38
rs774637826 Chr.20: 35437770-35437770 on Build GRCh38
rs868380625 Chr.20: 35437727-35437727 on Build GRCh38
rs756745099 Chr.20: 35437805-35437805 on Build GRCh38
rs977851749 Chr.20: 35437717-35437717 on Build GRCh38
rs200445104 Chr.20: 35437760-35437760 on Build GRCh38
rs144074930 Chr.20: 35437628-35437628 on Build GRCh38
rs764289151 Chr.20: 35437626-35437626 on Build GRCh38
rs889039427 Chr.20: 35437826-35437826 on Build GRCh38
rs745708689 Chr.20: 35437960-35437960 on Build GRCh38
rs751093117 Chr.20: 35437795-35437795 on Build GRCh38
rs542574339 Chr.20: 35437723-35437723 on Build GRCh38
rs754882664 Chr.20: 35437724-35437726 on Build GRCh38
rs920670845 Chr.20: 35437623-35437623 on Build GRCh38
rs376503178 Chr.20: 35437864-35437864 on Build GRCh38
rs767079839 Chr.20: 35437947-35437947 on Build GRCh38
rs565928127 Chr.20: 35437942-35437942 on Build GRCh38
rs767789406 Chr.20: 35437718-35437720 on Build GRCh38
rs369738392 Chr.20: 35437689-35437689 on Build GRCh38
rs140058823 Chr.20: 35437653-35437653 on Build GRCh38
rs147847468 Chr.20: 35437900-35437900 on Build GRCh38
rs777876942 Chr.20: 35437867-35437867 on Build GRCh38
rs755054277 Chr.20: 35437766-35437766 on Build GRCh38
rs746980493 Chr.20: 35437872-35437872 on Build GRCh38
rs966374627 Chr.20: 35438006-35438006 on Build GRCh38
rs779771024 Chr.20: 35437649-35437649 on Build GRCh38
rs778834209 Chr.20: 35437771-35437772 on Build GRCh38
rs914146150 Chr.20: 35438057-35438057 on Build GRCh38
rs746294424 Chr.20: 35437686-35437686 on Build GRCh38
rs369817738 Chr.20: 35437865-35437865 on Build GRCh38
rs753597147 Chr.20: 35437730-35437730 on Build GRCh38
rs945714416 Chr.20: 35437725-35437725 on Build GRCh38
rs755386910 Chr.20: 35437832-35437832 on Build GRCh38
rs777088932 Chr.20: 35437749-35437749 on Build GRCh38
rs746399130 Chr.20: 35437755-35437755 on Build GRCh38
rs747669190 Chr.20: 35437666-35437666 on Build GRCh38
rs144924248 Chr.20: 35438062-35438062 on Build GRCh38
rs774353141 Chr.20: 35437708-35437708 on Build GRCh38
rs779102835 Chr.20: 35437848-35437848 on Build GRCh38
rs992435308 Chr.20: 35438037-35438037 on Build GRCh38
rs748265022 Chr.20: 35437771-35437771 on Build GRCh38
rs747076228 Chr.20: 35437874-35437874 on Build GRCh38
rs762705770 Chr.20: 35437915-35437915 on Build GRCh38
rs143384 Chr.20: 35437976-35437976 on Build GRCh38
rs549803995 Chr.20: 35437845-35437845 on Build GRCh38
rs926866819 Chr.20: 35437637-35437637 on Build GRCh38
rs532328832 Chr.20: 35437901-35437901 on Build GRCh38
rs767434438 Chr.20: 35437719-35437719 on Build GRCh38
rs770917734 Chr.20: 35437880-35437880 on Build GRCh38
rs199666386 Chr.20: 35437761-35437761 on Build GRCh38
rs1048841167 Chr.20: 35437859-35437859 on Build GRCh38
rs567301733 Chr.20: 35437988-35437988 on Build GRCh38
rs763209567 Chr.20: 35437622-35437622 on Build GRCh38
rs926752271 Chr.20: 35438007-35438007 on Build GRCh38
rs752211507 Chr.20: 35437744-35437744 on Build GRCh38
rs749747287 Chr.20: 35437829-35437829 on Build GRCh38
rs748435907 Chr.20: 35437850-35437850 on Build GRCh38
rs753691079 Chr.20: 35437723-35437724 on Build GRCh38
rs773128415 Chr.20: 35437772-35437772 on Build GRCh38
rs73094730 Chr.20: 35438063-35438063 on Build GRCh38
rs148658511 Chr.20: 35437688-35437688 on Build GRCh38
rs780702378 Chr.20: 35437812-35437812 on Build GRCh38
rs371869707 Chr.20: 35437728-35437728 on Build GRCh38
rs757938728 Chr.20: 35437782-35437782 on Build GRCh38
rs149907722 Chr.20: 35437967-35437967 on Build GRCh38
rs761962752 Chr.20: 35437631-35437632 on Build GRCh38
rs895019955 Chr.20: 35437975-35437975 on Build GRCh38
rs547280273 Chr.20: 35437941-35437941 on Build GRCh38
rs764925735 Chr.20: 35437743-35437743 on Build GRCh38

View Full Product Details