Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35466880-35467374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP250
Entrez Gene ID
11190
Gene Name
centrosomal protein 250
Gene Aliases
C-NAP1, CEP2, CNAP1
UniGene
Hs.443976
Gene Chromosome Location
Chr.20: 35455138-35517531 on Build GRCh38
Transcripts
NM_001318219.1 NM_007186.5 XM_005260262.4 XM_005260263.4 XM_005260264.4 XM_006723690.3 XM_006723691.1 XM_006723692.3 XM_006723693.3 XM_006723694.3 XM_011528517.2 XM_011528518.2 XM_011528519.2 XM_017027617.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363194 c.566G>A p.R189H Chr.20: 35467039-35467039 on Build GRCh38
6627012 c.614T>C p.L205P Chr.20: 35467318-35467318 on Build GRCh38
7176188 c.510G>A p.L170L Chr.20: 35466983-35466983 on Build GRCh38
268875 c.549G>A p.E183E Chr.20: 35467022-35467022 on Build GRCh38
3545738 c.664C>A p.R222S Chr.20: 35467368-35467368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007340158 Chr.20: 35466974-35466974 on Build GRCh38
rs987862631 Chr.20: 35467218-35467218 on Build GRCh38
rs912896813 Chr.20: 35467202-35467202 on Build GRCh38
rs755314316 Chr.20: 35467036-35467036 on Build GRCh38
rs747074479 Chr.20: 35467058-35467058 on Build GRCh38
rs2296402 Chr.20: 35466904-35466904 on Build GRCh38
rs969078666 Chr.20: 35467162-35467162 on Build GRCh38
rs770748663 Chr.20: 35466995-35466995 on Build GRCh38
rs780053752 Chr.20: 35467282-35467282 on Build GRCh38
rs746986257 Chr.20: 35466986-35466986 on Build GRCh38
rs935563204 Chr.20: 35467148-35467148 on Build GRCh38
rs770269509 Chr.20: 35467353-35467353 on Build GRCh38
rs887995783 Chr.20: 35467175-35467175 on Build GRCh38
rs377351050 Chr.20: 35467211-35467211 on Build GRCh38
rs567008977 Chr.20: 35467171-35467171 on Build GRCh38
rs761414182 Chr.20: 35467091-35467091 on Build GRCh38
rs758674972 Chr.20: 35467110-35467110 on Build GRCh38
rs144992146 Chr.20: 35466929-35466929 on Build GRCh38
rs946230087 Chr.20: 35467238-35467238 on Build GRCh38
rs778723730 Chr.20: 35467312-35467312 on Build GRCh38
rs900369302 Chr.20: 35467243-35467243 on Build GRCh38
rs749092245 Chr.20: 35467296-35467296 on Build GRCh38
rs775018359 Chr.20: 35467017-35467017 on Build GRCh38
rs772751738 Chr.20: 35467332-35467332 on Build GRCh38
rs1051811400 Chr.20: 35466931-35466931 on Build GRCh38
rs35257858 Chr.20: 35467289-35467290 on Build GRCh38
rs745823057 Chr.20: 35467271-35467271 on Build GRCh38
rs774073842 Chr.20: 35467088-35467088 on Build GRCh38
rs150110885 Chr.20: 35466999-35466999 on Build GRCh38
rs757295577 Chr.20: 35467261-35467261 on Build GRCh38
rs780994840 Chr.20: 35467062-35467062 on Build GRCh38
rs551595644 Chr.20: 35467161-35467161 on Build GRCh38
rs200083040 Chr.20: 35466965-35466965 on Build GRCh38
rs763738378 Chr.20: 35467021-35467021 on Build GRCh38
rs748315594 Chr.20: 35466939-35466939 on Build GRCh38
rs201927235 Chr.20: 35467000-35467000 on Build GRCh38
rs112836240 Chr.20: 35466909-35466909 on Build GRCh38
rs764294331 Chr.20: 35467121-35467121 on Build GRCh38
rs777876722 Chr.20: 35467043-35467043 on Build GRCh38
rs942971751 Chr.20: 35467274-35467274 on Build GRCh38
rs932285833 Chr.20: 35467166-35467166 on Build GRCh38
rs924958178 Chr.20: 35467176-35467176 on Build GRCh38
rs772166343 Chr.20: 35466948-35466948 on Build GRCh38
rs761323783 Chr.20: 35467023-35467023 on Build GRCh38
rs6060429 Chr.20: 35466914-35466914 on Build GRCh38
rs776545208 Chr.20: 35466996-35466996 on Build GRCh38
rs751711345 Chr.20: 35467253-35467253 on Build GRCh38
rs533431347 Chr.20: 35467087-35467087 on Build GRCh38
rs1023843258 Chr.20: 35467097-35467097 on Build GRCh38
rs765556337 Chr.20: 35467100-35467100 on Build GRCh38
rs376695125 Chr.20: 35467026-35467026 on Build GRCh38
rs982407136 Chr.20: 35467128-35467128 on Build GRCh38
rs765592288 Chr.20: 35467038-35467038 on Build GRCh38
rs779010891 Chr.20: 35466938-35466938 on Build GRCh38
rs781284061 Chr.20: 35467270-35467270 on Build GRCh38
rs371345185 Chr.20: 35467338-35467338 on Build GRCh38
rs748997441 Chr.20: 35467084-35467084 on Build GRCh38
rs183055406 Chr.20: 35467229-35467229 on Build GRCh38
rs747791151 Chr.20: 35467316-35467316 on Build GRCh38
rs769497701 Chr.20: 35467070-35467070 on Build GRCh38
rs768501038 Chr.20: 35467311-35467311 on Build GRCh38
rs760053107 Chr.20: 35467095-35467095 on Build GRCh38
rs766996907 Chr.20: 35467093-35467093 on Build GRCh38
rs753034196 Chr.20: 35467105-35467105 on Build GRCh38
rs757282804 Chr.20: 35467060-35467060 on Build GRCh38
rs928219584 Chr.20: 35467141-35467141 on Build GRCh38
rs969946779 Chr.20: 35467126-35467126 on Build GRCh38
rs745759891 Chr.20: 35467069-35467069 on Build GRCh38
rs777029853 Chr.20: 35467094-35467094 on Build GRCh38
rs973480161 Chr.20: 35467051-35467051 on Build GRCh38
rs760005480 Chr.20: 35467337-35467337 on Build GRCh38
rs138289150 Chr.20: 35467039-35467039 on Build GRCh38
rs766793865 Chr.20: 35467024-35467024 on Build GRCh38
rs771517134 Chr.20: 35467331-35467331 on Build GRCh38
rs988886286 Chr.20: 35467159-35467159 on Build GRCh38
rs763398872 Chr.20: 35467351-35467361 on Build GRCh38
rs756056295 Chr.20: 35467276-35467276 on Build GRCh38
rs758583260 Chr.20: 35467041-35467041 on Build GRCh38

View Full Product Details