Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35522590-35523092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf173
Entrez Gene ID
140873
Gene Name
chromosome 20 open reading frame 173
Gene Aliases
dJ477O4.4
UniGene
Hs.158680
Gene Chromosome Location
Chr.20: 35520741-35529652 on Build GRCh38
Transcripts
NR_026933.2

SNP Detail

SNP ID Chromosome Location
rs939620468 Chr.20: 35522888-35522888 on Build GRCh38
rs147755834 Chr.20: 35523050-35523050 on Build GRCh38
rs373185209 Chr.20: 35522794-35522794 on Build GRCh38
rs1006855386 Chr.20: 35522795-35522795 on Build GRCh38
rs902593484 Chr.20: 35522723-35522724 on Build GRCh38
rs936589644 Chr.20: 35522782-35522782 on Build GRCh38
rs768200272 Chr.20: 35522854-35522854 on Build GRCh38
rs949238799 Chr.20: 35522666-35522666 on Build GRCh38
rs1025798458 Chr.20: 35522899-35522899 on Build GRCh38
rs529455065 Chr.20: 35522720-35522720 on Build GRCh38
rs537586149 Chr.20: 35522967-35522967 on Build GRCh38
rs192623926 Chr.20: 35522971-35522971 on Build GRCh38
rs952977096 Chr.20: 35522944-35522944 on Build GRCh38
rs999401374 Chr.20: 35522733-35522733 on Build GRCh38
rs1053826450 Chr.20: 35522787-35522787 on Build GRCh38
rs539661224 Chr.20: 35522774-35522774 on Build GRCh38
rs188610760 Chr.20: 35522852-35522852 on Build GRCh38
rs565804639 Chr.20: 35523000-35523000 on Build GRCh38
rs8118442 Chr.20: 35522646-35522646 on Build GRCh38
rs569255465 Chr.20: 35522747-35522747 on Build GRCh38
rs6060447 Chr.20: 35522976-35522976 on Build GRCh38
rs951652428 Chr.20: 35522805-35522805 on Build GRCh38
rs112216746 Chr.20: 35522965-35522965 on Build GRCh38
rs1028952328 Chr.20: 35523068-35523068 on Build GRCh38
rs966006024 Chr.20: 35522786-35522786 on Build GRCh38
rs916584516 Chr.20: 35522901-35522901 on Build GRCh38
rs185344392 Chr.20: 35522641-35522641 on Build GRCh38
rs552020457 Chr.20: 35522790-35522790 on Build GRCh38
rs972947384 Chr.20: 35522882-35522884 on Build GRCh38
rs141700633 Chr.20: 35522855-35522855 on Build GRCh38
rs1044110089 Chr.20: 35522951-35522951 on Build GRCh38
rs567052904 Chr.20: 35522833-35522833 on Build GRCh38
rs1022340795 Chr.20: 35522723-35522723 on Build GRCh38
rs895125000 Chr.20: 35522693-35522693 on Build GRCh38
rs767775641 Chr.20: 35523036-35523036 on Build GRCh38
rs999993753 Chr.20: 35523001-35523001 on Build GRCh38
rs573382269 Chr.20: 35522856-35522856 on Build GRCh38
rs996097106 Chr.20: 35523056-35523056 on Build GRCh38
rs551172619 Chr.20: 35522742-35522742 on Build GRCh38
rs1025632007 Chr.20: 35523007-35523007 on Build GRCh38
rs928099696 Chr.20: 35522882-35522882 on Build GRCh38
rs764255714 Chr.20: 35523031-35523031 on Build GRCh38
rs6058256 Chr.20: 35522673-35522673 on Build GRCh38
rs923884033 Chr.20: 35522758-35522758 on Build GRCh38
rs1037597782 Chr.20: 35522634-35522634 on Build GRCh38
rs369254526 Chr.20: 35522770-35522770 on Build GRCh38
rs994364575 Chr.20: 35522894-35522894 on Build GRCh38
rs1040219421 Chr.20: 35523023-35523023 on Build GRCh38
rs751695523 Chr.20: 35523037-35523037 on Build GRCh38

View Full Product Details