Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36173574-36174029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L1
Entrez Gene ID
2036
Gene Name
erythrocyte membrane protein band 4.1 like 1
Gene Aliases
4.1N, MRD11
UniGene
Hs.437422
Gene Chromosome Location
Chr.20: 36064891-36232799 on Build GRCh38
Transcripts
NM_001258329.1 NM_001258330.1 NM_001258331.1 NM_012156.2 NM_177996.2 XM_011528667.1 XM_011528671.1 XM_011528673.1 XM_011528677.1 XM_011528681.1 XM_011528682.1 XM_011528683.1 XM_011528685.1 XM_017027711.1 XM_017027712.1 XM_017027713.1 XM_017027714.1 XM_017027715.1 XM_017027716.1 XM_017027717.1 XM_017027718.1 XM_017027719.1 XM_017027720.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5814035 c.2T>A p.M1K Chr.20: 36173779-36173779 on Build GRCh38
5365963 c.76G>A p.A26T Chr.20: 36173853-36173853 on Build GRCh38
81138 c.111C>T p.H37H Chr.20: 36173888-36173888 on Build GRCh38
4504612 c.66C>T p.P22P Chr.20: 36173843-36173843 on Build GRCh38
4921872 c.114C>A p.G38G Chr.20: 36173891-36173891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775455374 Chr.20: 36173829-36173829 on Build GRCh38
rs371069159 Chr.20: 36173739-36173739 on Build GRCh38
rs181702972 Chr.20: 36173628-36173628 on Build GRCh38
rs957513104 Chr.20: 36173991-36173991 on Build GRCh38
rs200618454 Chr.20: 36173747-36173747 on Build GRCh38
rs908201468 Chr.20: 36174006-36174006 on Build GRCh38
rs1034681701 Chr.20: 36173657-36173657 on Build GRCh38
rs556499983 Chr.20: 36173652-36173652 on Build GRCh38
rs955260419 Chr.20: 36173981-36173981 on Build GRCh38
rs750211774 Chr.20: 36173897-36173897 on Build GRCh38
rs111619149 Chr.20: 36173825-36173825 on Build GRCh38
rs375684940 Chr.20: 36173980-36173980 on Build GRCh38
rs762083362 Chr.20: 36173740-36173740 on Build GRCh38
rs977923404 Chr.20: 36173717-36173717 on Build GRCh38
rs372761608 Chr.20: 36173855-36173855 on Build GRCh38
rs956080291 Chr.20: 36173621-36173621 on Build GRCh38
rs562835358 Chr.20: 36173941-36173941 on Build GRCh38
rs538096119 Chr.20: 36173645-36173645 on Build GRCh38
rs772016286 Chr.20: 36173817-36173817 on Build GRCh38
rs751452904 Chr.20: 36173997-36173997 on Build GRCh38
rs763709856 Chr.20: 36173879-36173879 on Build GRCh38
rs377478345 Chr.20: 36173777-36173777 on Build GRCh38
rs773788038 Chr.20: 36173843-36173843 on Build GRCh38
rs898660383 Chr.20: 36173758-36173758 on Build GRCh38
rs752178389 Chr.20: 36173865-36173865 on Build GRCh38
rs201658763 Chr.20: 36173812-36173812 on Build GRCh38
rs766975752 Chr.20: 36173860-36173860 on Build GRCh38
rs756943586 Chr.20: 36173808-36173808 on Build GRCh38
rs968326133 Chr.20: 36173662-36173662 on Build GRCh38
rs138256745 Chr.20: 36173921-36173921 on Build GRCh38
rs752229233 Chr.20: 36173785-36173785 on Build GRCh38
rs774939320 Chr.20: 36173955-36173955 on Build GRCh38
rs753490096 Chr.20: 36173883-36173883 on Build GRCh38
rs371405277 Chr.20: 36173938-36173938 on Build GRCh38
rs768666836 Chr.20: 36173926-36173926 on Build GRCh38
rs755743855 Chr.20: 36173798-36173798 on Build GRCh38
rs374844407 Chr.20: 36173964-36173964 on Build GRCh38
rs773359834 Chr.20: 36173940-36173940 on Build GRCh38
rs766997205 Chr.20: 36173961-36173961 on Build GRCh38
rs538440238 Chr.20: 36173686-36173686 on Build GRCh38
rs573229607 Chr.20: 36173704-36173704 on Build GRCh38
rs777563514 Chr.20: 36173799-36173799 on Build GRCh38
rs571614792 Chr.20: 36173677-36173677 on Build GRCh38
rs779965572 Chr.20: 36173909-36173909 on Build GRCh38
rs948934312 Chr.20: 36173765-36173765 on Build GRCh38
rs760195022 Chr.20: 36173866-36173866 on Build GRCh38
rs770031656 Chr.20: 36173939-36173939 on Build GRCh38
rs763401527 Chr.20: 36173708-36173708 on Build GRCh38
rs753358733 Chr.20: 36173974-36173974 on Build GRCh38
rs964830786 Chr.20: 36173927-36173927 on Build GRCh38
rs140731421 Chr.20: 36173888-36173888 on Build GRCh38
rs986408682 Chr.20: 36173688-36173688 on Build GRCh38
rs748952207 Chr.20: 36173801-36173801 on Build GRCh38
rs369402278 Chr.20: 36173846-36173846 on Build GRCh38
rs1014208690 Chr.20: 36173681-36173681 on Build GRCh38
rs553961201 Chr.20: 36173687-36173687 on Build GRCh38
rs367558996 Chr.20: 36173965-36173965 on Build GRCh38
rs111421912 Chr.20: 36173856-36173856 on Build GRCh38
rs145893462 Chr.20: 36173889-36173889 on Build GRCh38
rs768793627 Chr.20: 36173834-36173834 on Build GRCh38
rs562902192 Chr.20: 36173759-36173759 on Build GRCh38
rs761280005 Chr.20: 36173976-36173976 on Build GRCh38
rs778792843 Chr.20: 36173809-36173809 on Build GRCh38
rs758189925 Chr.20: 36173906-36173906 on Build GRCh38
rs370818122 Chr.20: 36173977-36173977 on Build GRCh38
rs758972199 Chr.20: 36173749-36173749 on Build GRCh38
rs975349406 Chr.20: 36173685-36173685 on Build GRCh38
rs573702846 Chr.20: 36173931-36173931 on Build GRCh38
rs894380826 Chr.20: 36173904-36173904 on Build GRCh38
rs1031679243 Chr.20: 36173609-36173609 on Build GRCh38
rs747082511 Chr.20: 36173833-36173833 on Build GRCh38
rs765641945 Chr.20: 36173741-36173741 on Build GRCh38
rs994738486 Chr.20: 36173655-36173656 on Build GRCh38

View Full Product Details