Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36240057-36240564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAR2
Entrez Gene ID
25980
Gene Name
AAR2 splicing factor homolog
Gene Aliases
C20orf4, CGI-23
UniGene
Hs.11314
Gene Chromosome Location
Chr.20: 36236136-36256941 on Build GRCh38
Transcripts
NM_001271874.1 NM_015511.4 XM_006723770.3 XM_011528762.2 XM_011528763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6857416 c.499A>G p.M167V Chr.20: 36240367-36240367 on Build GRCh38
6897576 c.220C>T p.R74C Chr.20: 36240088-36240088 on Build GRCh38
328766 c.685A>T p.S229C Chr.20: 36240553-36240553 on Build GRCh38
309691 c.301T>C p.S101P Chr.20: 36240169-36240169 on Build GRCh38
2759599 c.618C>T p.I206I Chr.20: 36240486-36240486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955002400 Chr.20: 36240534-36240534 on Build GRCh38
rs149423857 Chr.20: 36240474-36240474 on Build GRCh38
rs999598615 Chr.20: 36240224-36240224 on Build GRCh38
rs754532671 Chr.20: 36240176-36240176 on Build GRCh38
rs777278054 Chr.20: 36240173-36240173 on Build GRCh38
rs780673670 Chr.20: 36240154-36240154 on Build GRCh38
rs758879813 Chr.20: 36240151-36240151 on Build GRCh38
rs147662826 Chr.20: 36240093-36240093 on Build GRCh38
rs778338068 Chr.20: 36240380-36240380 on Build GRCh38
rs1032562842 Chr.20: 36240208-36240208 on Build GRCh38
rs201549531 Chr.20: 36240092-36240092 on Build GRCh38
rs779633484 Chr.20: 36240387-36240387 on Build GRCh38
rs781615324 Chr.20: 36240299-36240299 on Build GRCh38
rs749356105 Chr.20: 36240488-36240488 on Build GRCh38
rs750676328 Chr.20: 36240291-36240291 on Build GRCh38
rs372602653 Chr.20: 36240349-36240349 on Build GRCh38
rs779689445 Chr.20: 36240219-36240220 on Build GRCh38
rs716431 Chr.20: 36240537-36240537 on Build GRCh38
rs373216629 Chr.20: 36240119-36240119 on Build GRCh38
rs768037630 Chr.20: 36240352-36240352 on Build GRCh38
rs746020778 Chr.20: 36240513-36240513 on Build GRCh38
rs768628432 Chr.20: 36240326-36240326 on Build GRCh38
rs373127299 Chr.20: 36240302-36240302 on Build GRCh38
rs762334505 Chr.20: 36240532-36240532 on Build GRCh38
rs777915080 Chr.20: 36240487-36240487 on Build GRCh38
rs764187400 Chr.20: 36240132-36240132 on Build GRCh38
rs751659734 Chr.20: 36240188-36240188 on Build GRCh38
rs1038239797 Chr.20: 36240415-36240415 on Build GRCh38
rs772345367 Chr.20: 36240515-36240515 on Build GRCh38
rs770612106 Chr.20: 36240181-36240181 on Build GRCh38
rs138517690 Chr.20: 36240399-36240399 on Build GRCh38
rs747476412 Chr.20: 36240160-36240160 on Build GRCh38
rs780790498 Chr.20: 36240471-36240471 on Build GRCh38
rs748751730 Chr.20: 36240178-36240178 on Build GRCh38
rs761105630 Chr.20: 36240421-36240421 on Build GRCh38
rs759834621 Chr.20: 36240414-36240414 on Build GRCh38
rs1002180324 Chr.20: 36240113-36240113 on Build GRCh38
rs748920139 Chr.20: 36240095-36240095 on Build GRCh38
rs774065363 Chr.20: 36240114-36240114 on Build GRCh38
rs769382754 Chr.20: 36240089-36240089 on Build GRCh38
rs150337322 Chr.20: 36240285-36240285 on Build GRCh38
rs778587109 Chr.20: 36240183-36240183 on Build GRCh38
rs570444876 Chr.20: 36240301-36240301 on Build GRCh38
rs142460450 Chr.20: 36240118-36240118 on Build GRCh38
rs748581464 Chr.20: 36240292-36240305 on Build GRCh38
rs189919055 Chr.20: 36240234-36240234 on Build GRCh38
rs200342935 Chr.20: 36240508-36240508 on Build GRCh38
rs201038575 Chr.20: 36240507-36240507 on Build GRCh38
rs761088980 Chr.20: 36240523-36240523 on Build GRCh38
rs141691493 Chr.20: 36240420-36240420 on Build GRCh38
rs147834617 Chr.20: 36240211-36240211 on Build GRCh38
rs745632232 Chr.20: 36240184-36240184 on Build GRCh38
rs763296550 Chr.20: 36240246-36240246 on Build GRCh38
rs374608307 Chr.20: 36240422-36240422 on Build GRCh38
rs201609790 Chr.20: 36240346-36240346 on Build GRCh38
rs1046622376 Chr.20: 36240469-36240469 on Build GRCh38
rs749926117 Chr.20: 36240429-36240429 on Build GRCh38
rs896478501 Chr.20: 36240433-36240433 on Build GRCh38
rs76403264 Chr.20: 36240381-36240381 on Build GRCh38
rs768485629 Chr.20: 36240228-36240228 on Build GRCh38
rs774478093 Chr.20: 36240413-36240413 on Build GRCh38
rs775715913 Chr.20: 36240516-36240516 on Build GRCh38
rs751882156 Chr.20: 36240282-36240282 on Build GRCh38
rs772413491 Chr.20: 36240534-36240534 on Build GRCh38
rs781069168 Chr.20: 36240396-36240396 on Build GRCh38
rs34390439 Chr.20: 36240328-36240328 on Build GRCh38
rs201761855 Chr.20: 36240330-36240330 on Build GRCh38
rs375291820 Chr.20: 36240137-36240137 on Build GRCh38
rs755461449 Chr.20: 36240172-36240172 on Build GRCh38
rs377708694 Chr.20: 36240329-36240329 on Build GRCh38
rs368104861 Chr.20: 36240448-36240448 on Build GRCh38
rs201316006 Chr.20: 36240449-36240449 on Build GRCh38
rs775287748 Chr.20: 36240191-36240191 on Build GRCh38
rs970697472 Chr.20: 36240219-36240219 on Build GRCh38
rs537933325 Chr.20: 36240200-36240200 on Build GRCh38
rs138947526 Chr.20: 36240407-36240407 on Build GRCh38
rs548506013 Chr.20: 36240136-36240136 on Build GRCh38
rs1032942833 Chr.20: 36240490-36240490 on Build GRCh38
rs769126744 Chr.20: 36240386-36240386 on Build GRCh38
rs777166312 Chr.20: 36240531-36240531 on Build GRCh38
rs757956329 Chr.20: 36240430-36240430 on Build GRCh38
rs770503157 Chr.20: 36240102-36240102 on Build GRCh38
rs750800240 Chr.20: 36240143-36240143 on Build GRCh38
rs759876242 Chr.20: 36240351-36240351 on Build GRCh38
rs530309921 Chr.20: 36240080-36240080 on Build GRCh38
rs370233623 Chr.20: 36240416-36240416 on Build GRCh38
rs200929659 Chr.20: 36240406-36240406 on Build GRCh38
rs879221823 Chr.20: 36240510-36240510 on Build GRCh38
rs761860611 Chr.20: 36240236-36240236 on Build GRCh38
rs1021086767 Chr.20: 36240165-36240165 on Build GRCh38
rs747984876 Chr.20: 36240398-36240398 on Build GRCh38
rs771729009 Chr.20: 36240313-36240313 on Build GRCh38
rs746800707 Chr.20: 36240388-36240388 on Build GRCh38
rs144623771 Chr.20: 36240348-36240348 on Build GRCh38
rs369464017 Chr.20: 36240318-36240318 on Build GRCh38
rs745475861 Chr.20: 36240310-36240310 on Build GRCh38
rs766799042 Chr.20: 36240260-36240260 on Build GRCh38
rs753104874 Chr.20: 36240353-36240353 on Build GRCh38
rs757481964 Chr.20: 36240275-36240275 on Build GRCh38
rs760563020 Chr.20: 36240124-36240124 on Build GRCh38
rs748957834 Chr.20: 36240345-36240345 on Build GRCh38
rs146546733 Chr.20: 36240133-36240133 on Build GRCh38
rs747959052 Chr.20: 36240472-36240472 on Build GRCh38
rs756638120 Chr.20: 36240354-36240354 on Build GRCh38
rs199966492 Chr.20: 36240245-36240245 on Build GRCh38
rs374203470 Chr.20: 36240325-36240325 on Build GRCh38
rs6121183 Chr.20: 36240238-36240238 on Build GRCh38
rs764566378 Chr.20: 36240524-36240524 on Build GRCh38
rs1013661836 Chr.20: 36240161-36240161 on Build GRCh38
rs754678537 Chr.20: 36240456-36240456 on Build GRCh38
rs771094947 Chr.20: 36240496-36240496 on Build GRCh38
rs773445065 Chr.20: 36240334-36240334 on Build GRCh38
rs775448962 Chr.20: 36240121-36240121 on Build GRCh38

View Full Product Details