Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36892725-36893244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMHD1
Entrez Gene ID
25939
Gene Name
SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1
Gene Aliases
CHBL2, DCIP, HDDC1, MOP-5, SBBI88
UniGene
Hs.580681
Gene Chromosome Location
Chr.20: 36890882-36951843 on Build GRCh38
Transcripts
NM_015474.3 XM_005260384.3 XM_011528761.2 NM_001304783.1 NM_080628.2

Gene Symbol
TLDC2
Entrez Gene ID
140711
Gene Name
TBC/LysM-associated domain containing 2
Gene Aliases
C20orf118
UniGene
Hs.472630
Gene Chromosome Location
Chr.20: 36873071-36894235 on Build GRCh38
Transcripts
NM_015474.3 XM_005260384.3 XM_011528761.2 NM_001304783.1 NM_080628.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6093226 c.1761A>C p.I587I Chr.20: 36893052-36893052 on Build GRCh38
1411557 c.1787delA p.K596fs*>31 Chr.20: 36893026-36893026 on Build GRCh38
368312 c.1753G>A p.D585N Chr.20: 36893060-36893060 on Build GRCh38
5643776 c.1867G>A p.D623N Chr.20: 36892946-36892946 on Build GRCh38
4412992 c.1880G>A p.*627* Chr.20: 36892933-36892933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372533307 Chr.20: 36892965-36892965 on Build GRCh38
rs898149223 Chr.20: 36892752-36892752 on Build GRCh38
rs776940625 Chr.20: 36892821-36892821 on Build GRCh38
rs773683280 Chr.20: 36892961-36892961 on Build GRCh38
rs771446566 Chr.20: 36893091-36893091 on Build GRCh38
rs746419935 Chr.20: 36893100-36893100 on Build GRCh38
rs139005678 Chr.20: 36893222-36893222 on Build GRCh38
rs767155031 Chr.20: 36892937-36892937 on Build GRCh38
rs761596589 Chr.20: 36893015-36893015 on Build GRCh38
rs1031442313 Chr.20: 36893145-36893145 on Build GRCh38
rs368268178 Chr.20: 36892895-36892895 on Build GRCh38
rs188941712 Chr.20: 36892812-36892812 on Build GRCh38
rs763620244 Chr.20: 36892868-36892868 on Build GRCh38
rs200106463 Chr.20: 36893093-36893093 on Build GRCh38
rs45595032 Chr.20: 36892872-36892872 on Build GRCh38
rs755019176 Chr.20: 36893055-36893055 on Build GRCh38
rs62208115 Chr.20: 36892972-36892972 on Build GRCh38
rs190893470 Chr.20: 36893012-36893012 on Build GRCh38
rs750147783 Chr.20: 36893020-36893020 on Build GRCh38
rs747049626 Chr.20: 36892929-36892929 on Build GRCh38
rs768050438 Chr.20: 36893103-36893103 on Build GRCh38
rs770049426 Chr.20: 36892944-36892944 on Build GRCh38
rs777863189 Chr.20: 36893066-36893066 on Build GRCh38
rs750326939 Chr.20: 36892914-36892914 on Build GRCh38
rs770133980 Chr.20: 36892784-36892784 on Build GRCh38
rs150658755 Chr.20: 36893064-36893064 on Build GRCh38
rs35102927 Chr.20: 36893016-36893016 on Build GRCh38
rs1012548822 Chr.20: 36892811-36892811 on Build GRCh38
rs968013512 Chr.20: 36892898-36892898 on Build GRCh38
rs1041748449 Chr.20: 36893181-36893181 on Build GRCh38
rs927154496 Chr.20: 36893147-36893147 on Build GRCh38
rs756982727 Chr.20: 36892904-36892904 on Build GRCh38
rs768797500 Chr.20: 36892815-36892815 on Build GRCh38
rs749714514 Chr.20: 36893082-36893082 on Build GRCh38
rs1021530519 Chr.20: 36893212-36893212 on Build GRCh38
rs138927042 Chr.20: 36892861-36892861 on Build GRCh38
rs780697696 Chr.20: 36893088-36893088 on Build GRCh38
rs542086135 Chr.20: 36893148-36893148 on Build GRCh38
rs954400529 Chr.20: 36893029-36893029 on Build GRCh38
rs775082384 Chr.20: 36892878-36892878 on Build GRCh38
rs758217797 Chr.20: 36893023-36893023 on Build GRCh38
rs115645414 Chr.20: 36892887-36892887 on Build GRCh38
rs760217203 Chr.20: 36892982-36892982 on Build GRCh38
rs779967417 Chr.20: 36892919-36892919 on Build GRCh38
rs776120597 Chr.20: 36893003-36893003 on Build GRCh38
rs147220022 Chr.20: 36892837-36892837 on Build GRCh38
rs776267197 Chr.20: 36893115-36893115 on Build GRCh38
rs749631141 Chr.20: 36893089-36893089 on Build GRCh38
rs550047744 Chr.20: 36892943-36892943 on Build GRCh38
rs1051223670 Chr.20: 36893011-36893011 on Build GRCh38
rs1057261870 Chr.20: 36893161-36893161 on Build GRCh38
rs773828572 Chr.20: 36892873-36892873 on Build GRCh38
rs774995485 Chr.20: 36893076-36893077 on Build GRCh38
rs751545200 Chr.20: 36893051-36893051 on Build GRCh38
rs570535296 Chr.20: 36892828-36892828 on Build GRCh38
rs868061483 Chr.20: 36893032-36893032 on Build GRCh38
rs1003030445 Chr.20: 36893133-36893133 on Build GRCh38
rs1011430017 Chr.20: 36893166-36893166 on Build GRCh38
rs200398507 Chr.20: 36892930-36892930 on Build GRCh38
rs534458534 Chr.20: 36893158-36893158 on Build GRCh38
rs765124468 Chr.20: 36892910-36892910 on Build GRCh38
rs150713148 Chr.20: 36892981-36892981 on Build GRCh38
rs753611896 Chr.20: 36893061-36893061 on Build GRCh38
rs940227703 Chr.20: 36893057-36893057 on Build GRCh38
rs770241845 Chr.20: 36892841-36892841 on Build GRCh38
rs895922901 Chr.20: 36893163-36893163 on Build GRCh38
rs762361576 Chr.20: 36893082-36893083 on Build GRCh38
rs895851053 Chr.20: 36892850-36892850 on Build GRCh38
rs1044047789 Chr.20: 36893109-36893109 on Build GRCh38
rs569571932 Chr.20: 36892918-36892918 on Build GRCh38
rs766253283 Chr.20: 36893037-36893037 on Build GRCh38
rs368771802 Chr.20: 36893096-36893096 on Build GRCh38
rs905658920 Chr.20: 36892800-36892800 on Build GRCh38
rs202165710 Chr.20: 36893060-36893060 on Build GRCh38
rs139841806 Chr.20: 36893001-36893001 on Build GRCh38

View Full Product Details