Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37524295-37524791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLCAP
Entrez Gene ID
10904
Gene Name
bladder cancer associated protein
Gene Aliases
BC10
UniGene
Hs.472651
Gene Chromosome Location
Chr.20: 37517417-37527931 on Build GRCh38
Transcripts
NM_001167820.1 NM_001167821.1 NM_001167822.2 NM_001167823.1 NM_006698.3

SNP Detail

SNP ID Chromosome Location
rs1000243615 Chr.20: 37524736-37524736 on Build GRCh38
rs901416602 Chr.20: 37524383-37524383 on Build GRCh38
rs554334295 Chr.20: 37524402-37524402 on Build GRCh38
rs145718145 Chr.20: 37524637-37524637 on Build GRCh38
rs543033921 Chr.20: 37524436-37524436 on Build GRCh38
rs549584552 Chr.20: 37524713-37524713 on Build GRCh38
rs905736255 Chr.20: 37524554-37524554 on Build GRCh38
rs1017950657 Chr.20: 37524742-37524742 on Build GRCh38
rs895690887 Chr.20: 37524568-37524568 on Build GRCh38
rs968377737 Chr.20: 37524381-37524381 on Build GRCh38
rs1041023027 Chr.20: 37524679-37524679 on Build GRCh38
rs992475743 Chr.20: 37524473-37524473 on Build GRCh38
rs964640026 Chr.20: 37524754-37524754 on Build GRCh38
rs1007111918 Chr.20: 37524322-37524322 on Build GRCh38
rs1045091154 Chr.20: 37524538-37524538 on Build GRCh38
rs527928364 Chr.20: 37524700-37524700 on Build GRCh38
rs936767525 Chr.20: 37524582-37524582 on Build GRCh38
rs753560332 Chr.20: 37524393-37524393 on Build GRCh38
rs766189657 Chr.20: 37524356-37524356 on Build GRCh38
rs977665353 Chr.20: 37524487-37524487 on Build GRCh38
rs1001467113 Chr.20: 37524558-37524558 on Build GRCh38
rs968450397 Chr.20: 37524660-37524660 on Build GRCh38
rs139992835 Chr.20: 37524411-37524411 on Build GRCh38
rs1019903677 Chr.20: 37524361-37524361 on Build GRCh38
rs149844774 Chr.20: 37524486-37524486 on Build GRCh38
rs547424683 Chr.20: 37524517-37524517 on Build GRCh38
rs942729887 Chr.20: 37524677-37524677 on Build GRCh38
rs988553661 Chr.20: 37524357-37524357 on Build GRCh38
rs778898974 Chr.20: 37524516-37524516 on Build GRCh38
rs754919861 Chr.20: 37524457-37524457 on Build GRCh38
rs563371307 Chr.20: 37524455-37524455 on Build GRCh38
rs1053844827 Chr.20: 37524612-37524612 on Build GRCh38
rs1009921795 Chr.20: 37524609-37524609 on Build GRCh38
rs889758014 Chr.20: 37524321-37524321 on Build GRCh38
rs994396621 Chr.20: 37524730-37524730 on Build GRCh38
rs565879091 Chr.20: 37524697-37524697 on Build GRCh38
rs1019854743 Chr.20: 37524359-37524359 on Build GRCh38
rs1057359129 Chr.20: 37524565-37524565 on Build GRCh38
rs564503546 Chr.20: 37524480-37524480 on Build GRCh38
rs925768511 Chr.20: 37524530-37524530 on Build GRCh38
rs747126546 Chr.20: 37524684-37524684 on Build GRCh38
rs574412514 Chr.20: 37524384-37524384 on Build GRCh38
rs952577972 Chr.20: 37524416-37524416 on Build GRCh38
rs567989524 Chr.20: 37524744-37524744 on Build GRCh38

View Full Product Details