Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37765106-37765609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNBL1
Entrez Gene ID
56259
Gene Name
catenin beta like 1
Gene Aliases
C20orf33, NAP, P14L, PP8304, dJ633O20.1
UniGene
Hs.472667
Gene Chromosome Location
Chr.20: 37693955-37872129 on Build GRCh38
Transcripts
NM_001281495.1 NM_030877.4 XM_011528917.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4943744 c.658+8G>C p.? Chr.20: 37765298-37765298 on Build GRCh38
1632277 c.597G>C p.Q199H Chr.20: 37765229-37765229 on Build GRCh38
6004393 c.618G>A p.E206E Chr.20: 37765250-37765250 on Build GRCh38
6558732 c.642C>T p.G214G Chr.20: 37765274-37765274 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1016523708 Chr.20: 37765267-37765267 on Build GRCh38
rs994879814 Chr.20: 37765347-37765348 on Build GRCh38
rs977855173 Chr.20: 37765146-37765146 on Build GRCh38
rs766279280 Chr.20: 37765229-37765229 on Build GRCh38
rs1033476546 Chr.20: 37765435-37765435 on Build GRCh38
rs940401166 Chr.20: 37765297-37765297 on Build GRCh38
rs975794502 Chr.20: 37765462-37765462 on Build GRCh38
rs761436253 Chr.20: 37765151-37765151 on Build GRCh38
rs943628446 Chr.20: 37765150-37765150 on Build GRCh38
rs990302036 Chr.20: 37765396-37765396 on Build GRCh38
rs921856375 Chr.20: 37765480-37765480 on Build GRCh38
rs530483499 Chr.20: 37765284-37765284 on Build GRCh38
rs967473710 Chr.20: 37765417-37765417 on Build GRCh38
rs781389055 Chr.20: 37765298-37765298 on Build GRCh38
rs533179207 Chr.20: 37765133-37765133 on Build GRCh38
rs999214111 Chr.20: 37765371-37765371 on Build GRCh38
rs756139187 Chr.20: 37765347-37765347 on Build GRCh38
rs554336045 Chr.20: 37765393-37765393 on Build GRCh38
rs931819059 Chr.20: 37765242-37765242 on Build GRCh38
rs894707835 Chr.20: 37765374-37765374 on Build GRCh38
rs933329656 Chr.20: 37765493-37765493 on Build GRCh38
rs930344642 Chr.20: 37765528-37765528 on Build GRCh38
rs914730577 Chr.20: 37765420-37765420 on Build GRCh38
rs998940510 Chr.20: 37765424-37765424 on Build GRCh38
rs74975514 Chr.20: 37765308-37765308 on Build GRCh38
rs1047775950 Chr.20: 37765494-37765494 on Build GRCh38
rs6096107 Chr.20: 37765582-37765582 on Build GRCh38
rs886253006 Chr.20: 37765240-37765240 on Build GRCh38
rs767651139 Chr.20: 37765232-37765232 on Build GRCh38
rs750455079 Chr.20: 37765250-37765250 on Build GRCh38
rs7262900 Chr.20: 37765135-37765135 on Build GRCh38
rs762050034 Chr.20: 37765184-37765184 on Build GRCh38
rs1047092641 Chr.20: 37765551-37765551 on Build GRCh38
rs367782973 Chr.20: 37765558-37765558 on Build GRCh38
rs1037501263 Chr.20: 37765534-37765534 on Build GRCh38
rs538871489 Chr.20: 37765379-37765379 on Build GRCh38
rs898825678 Chr.20: 37765334-37765334 on Build GRCh38
rs867056026 Chr.20: 37765269-37765269 on Build GRCh38
rs754318729 Chr.20: 37765442-37765442 on Build GRCh38
rs546100560 Chr.20: 37765466-37765466 on Build GRCh38
rs536525318 Chr.20: 37765510-37765510 on Build GRCh38
rs779207103 Chr.20: 37765495-37765495 on Build GRCh38
rs755517443 Chr.20: 37765492-37765492 on Build GRCh38
rs758269905 Chr.20: 37765209-37765209 on Build GRCh38
rs765035294 Chr.20: 37765183-37765183 on Build GRCh38
rs190928177 Chr.20: 37765482-37765482 on Build GRCh38
rs942117162 Chr.20: 37765502-37765502 on Build GRCh38
rs895028985 Chr.20: 37765274-37765274 on Build GRCh38
rs150168338 Chr.20: 37765286-37765286 on Build GRCh38
rs1014442570 Chr.20: 37765565-37765565 on Build GRCh38
rs188004269 Chr.20: 37765188-37765188 on Build GRCh38
rs1055360158 Chr.20: 37765550-37765550 on Build GRCh38
rs548237340 Chr.20: 37765187-37765187 on Build GRCh38

View Full Product Details