Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38588053-38588556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIG
Entrez Gene ID
149685
Gene Name
adipogenin
Gene Aliases
SMAF1
UniGene
Hs.368028
Gene Chromosome Location
Chr.20: 38581195-38588463 on Build GRCh38
Transcripts
NM_001018082.2 NR_130907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4447857 c.470_471GG>AA p.G157E Chr.20: 38588346-38588347 on Build GRCh38
4445310 c.379C>T p.P127S Chr.20: 38588255-38588255 on Build GRCh38
4445308 c.270G>A p.W90* Chr.20: 38588146-38588146 on Build GRCh38
6988432 c.427C>T p.R143C Chr.20: 38588303-38588303 on Build GRCh38
4291897 c.361G>A p.V121M Chr.20: 38588237-38588237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954405361 Chr.20: 38588168-38588168 on Build GRCh38
rs766608262 Chr.20: 38588336-38588336 on Build GRCh38
rs538040922 Chr.20: 38588352-38588352 on Build GRCh38
rs940527702 Chr.20: 38588290-38588290 on Build GRCh38
rs534126664 Chr.20: 38588260-38588260 on Build GRCh38
rs78725800 Chr.20: 38588504-38588504 on Build GRCh38
rs566728510 Chr.20: 38588248-38588248 on Build GRCh38
rs35940408 Chr.20: 38588210-38588211 on Build GRCh38
rs964319882 Chr.20: 38588082-38588082 on Build GRCh38
rs553765821 Chr.20: 38588511-38588511 on Build GRCh38
rs145214707 Chr.20: 38588437-38588437 on Build GRCh38
rs775161565 Chr.20: 38588167-38588167 on Build GRCh38
rs751985067 Chr.20: 38588346-38588346 on Build GRCh38
rs540293153 Chr.20: 38588192-38588192 on Build GRCh38
rs773867033 Chr.20: 38588160-38588160 on Build GRCh38
rs865939728 Chr.20: 38588375-38588375 on Build GRCh38
rs1011066622 Chr.20: 38588443-38588443 on Build GRCh38
rs1045994319 Chr.20: 38588495-38588495 on Build GRCh38
rs773232152 Chr.20: 38588376-38588376 on Build GRCh38
rs753802347 Chr.20: 38588197-38588197 on Build GRCh38
rs977259293 Chr.20: 38588442-38588455 on Build GRCh38
rs567539405 Chr.20: 38588307-38588307 on Build GRCh38
rs755307735 Chr.20: 38588353-38588353 on Build GRCh38
rs1003580037 Chr.20: 38588380-38588380 on Build GRCh38
rs759146226 Chr.20: 38588161-38588161 on Build GRCh38
rs771689230 Chr.20: 38588163-38588163 on Build GRCh38
rs370029546 Chr.20: 38588397-38588397 on Build GRCh38
rs186341754 Chr.20: 38588118-38588118 on Build GRCh38
rs3171210 Chr.20: 38588455-38588455 on Build GRCh38
rs372230740 Chr.20: 38588461-38588461 on Build GRCh38
rs1022444229 Chr.20: 38588457-38588457 on Build GRCh38
rs748839046 Chr.20: 38588100-38588100 on Build GRCh38
rs115965434 Chr.20: 38588232-38588232 on Build GRCh38
rs145364287 Chr.20: 38588303-38588303 on Build GRCh38
rs1054871497 Chr.20: 38588533-38588533 on Build GRCh38
rs998837437 Chr.20: 38588527-38588527 on Build GRCh38
rs947949898 Chr.20: 38588372-38588372 on Build GRCh38
rs191196275 Chr.20: 38588180-38588180 on Build GRCh38
rs141690880 Chr.20: 38588157-38588158 on Build GRCh38
rs867215598 Chr.20: 38588121-38588121 on Build GRCh38
rs987194913 Chr.20: 38588516-38588516 on Build GRCh38
rs756608486 Chr.20: 38588456-38588456 on Build GRCh38
rs538707856 Chr.20: 38588378-38588378 on Build GRCh38
rs975181463 Chr.20: 38588231-38588231 on Build GRCh38
rs560022739 Chr.20: 38588212-38588212 on Build GRCh38
rs964225362 Chr.20: 38588475-38588475 on Build GRCh38
rs1054920211 Chr.20: 38588448-38588448 on Build GRCh38
rs903170357 Chr.20: 38588094-38588094 on Build GRCh38
rs200838830 Chr.20: 38588503-38588503 on Build GRCh38
rs867108685 Chr.20: 38588281-38588281 on Build GRCh38
rs1037524755 Chr.20: 38588314-38588314 on Build GRCh38
rs758558589 Chr.20: 38588310-38588310 on Build GRCh38
rs760574995 Chr.20: 38588445-38588445 on Build GRCh38
rs770770979 Chr.20: 38588218-38588220 on Build GRCh38

View Full Product Details