Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44247121-44247619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP1L1
Entrez Gene ID
78997
Gene Name
ganglioside induced differentiation associated protein 1 like 1
Gene Aliases
dJ881L22.1, dJ995J12.1.1
UniGene
Hs.517059
Gene Chromosome Location
Chr.20: 44247099-44280917 on Build GRCh38
Transcripts
NM_001256737.1 NM_001256738.1 NM_001256739.1 NM_001256740.1 NM_024034.4 NR_046353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6004405 c.55G>A p.A19T Chr.20: 44247389-44247389 on Build GRCh38
6004404 c.55G>A p.A19T Chr.20: 44247389-44247389 on Build GRCh38
5952827 c.117C>T p.P39P Chr.20: 44247451-44247451 on Build GRCh38
5952826 c.117C>T p.P39P Chr.20: 44247451-44247451 on Build GRCh38
3292672 c.56C>T p.A19V Chr.20: 44247390-44247390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748624632 Chr.20: 44247413-44247413 on Build GRCh38
rs757040831 Chr.20: 44247375-44247375 on Build GRCh38
rs755013644 Chr.20: 44247401-44247401 on Build GRCh38
rs747445473 Chr.20: 44247301-44247301 on Build GRCh38
rs914365119 Chr.20: 44247448-44247448 on Build GRCh38
rs990835159 Chr.20: 44247225-44247225 on Build GRCh38
rs892884979 Chr.20: 44247430-44247430 on Build GRCh38
rs764988544 Chr.20: 44247464-44247464 on Build GRCh38
rs1004904881 Chr.20: 44247599-44247599 on Build GRCh38
rs562714366 Chr.20: 44247294-44247294 on Build GRCh38
rs768083106 Chr.20: 44247450-44247450 on Build GRCh38
rs966964300 Chr.20: 44247147-44247147 on Build GRCh38
rs765043496 Chr.20: 44247376-44247376 on Build GRCh38
rs762748059 Chr.20: 44247471-44247471 on Build GRCh38
rs779718999 Chr.20: 44247559-44247559 on Build GRCh38
rs780091513 Chr.20: 44247395-44247395 on Build GRCh38
rs576303211 Chr.20: 44247511-44247511 on Build GRCh38
rs1041604420 Chr.20: 44247522-44247522 on Build GRCh38
rs1056492011 Chr.20: 44247255-44247255 on Build GRCh38
rs758662793 Chr.20: 44247388-44247388 on Build GRCh38
rs761981611 Chr.20: 44247314-44247314 on Build GRCh38
rs756751004 Chr.20: 44247525-44247525 on Build GRCh38
rs375779631 Chr.20: 44247440-44247440 on Build GRCh38
rs933805498 Chr.20: 44247363-44247363 on Build GRCh38
rs945825161 Chr.20: 44247201-44247201 on Build GRCh38
rs549942097 Chr.20: 44247260-44247260 on Build GRCh38
rs991237037 Chr.20: 44247566-44247566 on Build GRCh38
rs1012346397 Chr.20: 44247278-44247279 on Build GRCh38
rs895115145 Chr.20: 44247258-44247258 on Build GRCh38
rs887891433 Chr.20: 44247596-44247596 on Build GRCh38
rs370907086 Chr.20: 44247254-44247254 on Build GRCh38
rs758428645 Chr.20: 44247289-44247289 on Build GRCh38
rs760518324 Chr.20: 44247361-44247361 on Build GRCh38
rs776586206 Chr.20: 44247452-44247452 on Build GRCh38
rs775208864 Chr.20: 44247358-44247358 on Build GRCh38
rs781692296 Chr.20: 44247403-44247403 on Build GRCh38
rs771428365 Chr.20: 44247541-44247541 on Build GRCh38
rs567838751 Chr.20: 44247232-44247232 on Build GRCh38
rs1041573220 Chr.20: 44247226-44247227 on Build GRCh38
rs959395427 Chr.20: 44247210-44247210 on Build GRCh38
rs780159199 Chr.20: 44247296-44247296 on Build GRCh38
rs114191782 Chr.20: 44247536-44247536 on Build GRCh38
rs749607423 Chr.20: 44247538-44247538 on Build GRCh38
rs777015313 Chr.20: 44247309-44247309 on Build GRCh38
rs147883207 Chr.20: 44247423-44247423 on Build GRCh38
rs373063886 Chr.20: 44247367-44247367 on Build GRCh38
rs771910437 Chr.20: 44247437-44247437 on Build GRCh38
rs758243650 Chr.20: 44247161-44247161 on Build GRCh38
rs745509349 Chr.20: 44247436-44247436 on Build GRCh38
rs931767067 Chr.20: 44247143-44247143 on Build GRCh38
rs543683527 Chr.20: 44247587-44247587 on Build GRCh38
rs746124320 Chr.20: 44247287-44247287 on Build GRCh38
rs554464197 Chr.20: 44247457-44247457 on Build GRCh38
rs1037288472 Chr.20: 44247257-44247257 on Build GRCh38
rs1021087767 Chr.20: 44247143-44247143 on Build GRCh38
rs867982565 Chr.20: 44247407-44247407 on Build GRCh38
rs760165610 Chr.20: 44247449-44247449 on Build GRCh38
rs1049042812 Chr.20: 44247570-44247570 on Build GRCh38
rs1003190674 Chr.20: 44247247-44247247 on Build GRCh38
rs928188845 Chr.20: 44247185-44247185 on Build GRCh38
rs907155173 Chr.20: 44247244-44247244 on Build GRCh38
rs753725320 Chr.20: 44247368-44247368 on Build GRCh38
rs750288581 Chr.20: 44247469-44247469 on Build GRCh38
rs902712368 Chr.20: 44247528-44247528 on Build GRCh38
rs201862996 Chr.20: 44247348-44247348 on Build GRCh38
rs376618654 Chr.20: 44247378-44247378 on Build GRCh38
rs144892364 Chr.20: 44247334-44247334 on Build GRCh38
rs1012700079 Chr.20: 44247569-44247569 on Build GRCh38
rs373331422 Chr.20: 44247326-44247326 on Build GRCh38
rs755033248 Chr.20: 44247505-44247505 on Build GRCh38
rs952126379 Chr.20: 44247327-44247327 on Build GRCh38
rs370601048 Chr.20: 44247416-44247416 on Build GRCh38
rs928433854 Chr.20: 44247534-44247534 on Build GRCh38
rs982385220 Chr.20: 44247173-44247173 on Build GRCh38
rs747050378 Chr.20: 44247400-44247400 on Build GRCh38
rs769261755 Chr.20: 44247305-44247305 on Build GRCh38
rs781177575 Chr.20: 44247507-44247507 on Build GRCh38
rs777821001 Chr.20: 44247227-44247234 on Build GRCh38
rs199890970 Chr.20: 44247297-44247297 on Build GRCh38
rs766519426 Chr.20: 44247473-44247473 on Build GRCh38
rs1003705800 Chr.20: 44247529-44247529 on Build GRCh38
rs751757180 Chr.20: 44247491-44247491 on Build GRCh38
rs942407355 Chr.20: 44247408-44247408 on Build GRCh38
rs534476167 Chr.20: 44247424-44247424 on Build GRCh38
rs1011493058 Chr.20: 44247140-44247140 on Build GRCh38

View Full Product Details