Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44310873-44311365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FITM2
Entrez Gene ID
128486
Gene Name
fat storage inducing transmembrane protein 2
Gene Aliases
C20orf142, Fit2, dJ881L22.2
UniGene
Hs.632269
Gene Chromosome Location
Chr.20: 44306557-44311249 on Build GRCh38
Transcripts
NM_001080472.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5017628 c.122delT p.L41fs*43 Chr.20: 44311027-44311027 on Build GRCh38
4599221 c.116A>G p.K39R Chr.20: 44311033-44311033 on Build GRCh38
4509862 c.81C>T p.A27A Chr.20: 44311068-44311068 on Build GRCh38
5781390 c.96G>A p.M32I Chr.20: 44311053-44311053 on Build GRCh38
4581828 c.39_40insG p.T14fs*138 Chr.20: 44311109-44311110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185786331 Chr.20: 44311220-44311220 on Build GRCh38
rs897348484 Chr.20: 44311153-44311153 on Build GRCh38
rs985479020 Chr.20: 44310991-44310991 on Build GRCh38
rs536563717 Chr.20: 44311345-44311345 on Build GRCh38
rs767085102 Chr.20: 44311022-44311022 on Build GRCh38
rs375586401 Chr.20: 44310950-44310950 on Build GRCh38
rs1048772307 Chr.20: 44311115-44311115 on Build GRCh38
rs937233373 Chr.20: 44311106-44311106 on Build GRCh38
rs1001218170 Chr.20: 44310896-44310896 on Build GRCh38
rs919345947 Chr.20: 44311213-44311213 on Build GRCh38
rs758621850 Chr.20: 44311205-44311205 on Build GRCh38
rs1039265829 Chr.20: 44311257-44311257 on Build GRCh38
rs770142931 Chr.20: 44311111-44311111 on Build GRCh38
rs764188779 Chr.20: 44311009-44311009 on Build GRCh38
rs747058586 Chr.20: 44311095-44311095 on Build GRCh38
rs1031846241 Chr.20: 44311137-44311137 on Build GRCh38
rs150052004 Chr.20: 44311081-44311081 on Build GRCh38
rs547595301 Chr.20: 44311317-44311317 on Build GRCh38
rs571358142 Chr.20: 44310942-44310942 on Build GRCh38
rs199527707 Chr.20: 44311177-44311177 on Build GRCh38
rs771707542 Chr.20: 44311121-44311121 on Build GRCh38
rs374581060 Chr.20: 44311181-44311181 on Build GRCh38
rs764053395 Chr.20: 44311063-44311063 on Build GRCh38
rs762122051 Chr.20: 44310986-44310986 on Build GRCh38
rs763326107 Chr.20: 44311116-44311116 on Build GRCh38
rs957463537 Chr.20: 44311011-44311011 on Build GRCh38
rs142368699 Chr.20: 44311127-44311127 on Build GRCh38
rs777021731 Chr.20: 44310968-44310968 on Build GRCh38
rs770232661 Chr.20: 44311180-44311180 on Build GRCh38
rs1051283819 Chr.20: 44311339-44311339 on Build GRCh38
rs140366349 Chr.20: 44311150-44311150 on Build GRCh38
rs760305499 Chr.20: 44311193-44311193 on Build GRCh38
rs747063104 Chr.20: 44310945-44310945 on Build GRCh38
rs771241095 Chr.20: 44311185-44311185 on Build GRCh38
rs375636278 Chr.20: 44311168-44311168 on Build GRCh38
rs867953326 Chr.20: 44311074-44311074 on Build GRCh38
rs956072234 Chr.20: 44310894-44310894 on Build GRCh38
rs753762137 Chr.20: 44311068-44311068 on Build GRCh38
rs971849276 Chr.20: 44311255-44311255 on Build GRCh38
rs973392510 Chr.20: 44311208-44311209 on Build GRCh38
rs746081619 Chr.20: 44311088-44311088 on Build GRCh38
rs146812591 Chr.20: 44311129-44311129 on Build GRCh38
rs954336725 Chr.20: 44311179-44311179 on Build GRCh38
rs1051908329 Chr.20: 44311037-44311037 on Build GRCh38
rs757098052 Chr.20: 44311072-44311072 on Build GRCh38
rs369629822 Chr.20: 44310992-44310992 on Build GRCh38
rs372613519 Chr.20: 44311149-44311149 on Build GRCh38
rs554959292 Chr.20: 44311021-44311021 on Build GRCh38
rs540810697 Chr.20: 44311155-44311155 on Build GRCh38
rs775187515 Chr.20: 44311187-44311187 on Build GRCh38
rs929382065 Chr.20: 44311225-44311225 on Build GRCh38
rs933040394 Chr.20: 44311338-44311338 on Build GRCh38
rs753812368 Chr.20: 44311142-44311142 on Build GRCh38
rs6073401 Chr.20: 44311110-44311110 on Build GRCh38
rs763562381 Chr.20: 44311140-44311140 on Build GRCh38
rs891516151 Chr.20: 44311148-44311148 on Build GRCh38
rs148377517 Chr.20: 44311053-44311053 on Build GRCh38
rs925930292 Chr.20: 44311005-44311005 on Build GRCh38
rs769145443 Chr.20: 44310961-44310961 on Build GRCh38
rs531687026 Chr.20: 44311226-44311226 on Build GRCh38
rs551813456 Chr.20: 44310923-44310923 on Build GRCh38
rs780152118 Chr.20: 44311093-44311093 on Build GRCh38
rs200289087 Chr.20: 44310958-44310958 on Build GRCh38
rs112118897 Chr.20: 44311289-44311289 on Build GRCh38
rs777161804 Chr.20: 44311100-44311100 on Build GRCh38
rs549232349 Chr.20: 44311214-44311214 on Build GRCh38
rs998882269 Chr.20: 44310962-44310962 on Build GRCh38
rs751692747 Chr.20: 44311268-44311268 on Build GRCh38
rs1048722074 Chr.20: 44310921-44310921 on Build GRCh38
rs981317405 Chr.20: 44311264-44311264 on Build GRCh38
rs576070517 Chr.20: 44311099-44311099 on Build GRCh38
rs901595398 Chr.20: 44311327-44311327 on Build GRCh38
rs979996753 Chr.20: 44310934-44310934 on Build GRCh38
rs773501722 Chr.20: 44311112-44311112 on Build GRCh38
rs1014032573 Chr.20: 44311291-44311291 on Build GRCh38
rs35390759 Chr.20: 44311032-44311033 on Build GRCh38
rs890719881 Chr.20: 44311044-44311044 on Build GRCh38
rs1029905501 Chr.20: 44310965-44310965 on Build GRCh38
rs772264012 Chr.20: 44310931-44310931 on Build GRCh38
rs3746569 Chr.20: 44311245-44311245 on Build GRCh38
rs937303376 Chr.20: 44311271-44311271 on Build GRCh38
rs891438122 Chr.20: 44311282-44311282 on Build GRCh38
rs913109234 Chr.20: 44311031-44311031 on Build GRCh38
rs533782994 Chr.20: 44311309-44311309 on Build GRCh38
rs1052764320 Chr.20: 44311280-44311280 on Build GRCh38
rs140049914 Chr.20: 44311151-44311151 on Build GRCh38
rs912099074 Chr.20: 44311233-44311233 on Build GRCh38
rs988840253 Chr.20: 44311014-44311014 on Build GRCh38
rs556214658 Chr.20: 44311092-44311092 on Build GRCh38
rs117858854 Chr.20: 44311033-44311033 on Build GRCh38
rs371149406 Chr.20: 44311158-44311158 on Build GRCh38
rs1024656547 Chr.20: 44310900-44310900 on Build GRCh38
rs571707675 Chr.20: 44311259-44311259 on Build GRCh38
rs1004658892 Chr.20: 44311145-44311145 on Build GRCh38
rs930323232 Chr.20: 44310967-44310967 on Build GRCh38
rs368277008 Chr.20: 44311156-44311156 on Build GRCh38
rs567443045 Chr.20: 44311324-44311324 on Build GRCh38

View Full Product Details