Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45891322-45891819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSA
Entrez Gene ID
5476
Gene Name
cathepsin A
Gene Aliases
GLB2, GSL, NGBE, PPCA, PPGB
UniGene
Hs.609336
Gene Chromosome Location
Chr.20: 45890144-45898820 on Build GRCh38
Transcripts
NM_000308.3 NM_001127695.2 NM_001167594.2 NR_133656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6499051 c.116C>T p.S39F Chr.20: 45891630-45891630 on Build GRCh38
3713282 c.222C>A p.G74G Chr.20: 45891736-45891736 on Build GRCh38
6453591 c.122C>T p.A41V Chr.20: 45891636-45891636 on Build GRCh38
6539433 c.230C>G p.S77C Chr.20: 45891744-45891744 on Build GRCh38
5034732 c.54+3G>C p.? Chr.20: 45891382-45891382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575214387 Chr.20: 45891747-45891747 on Build GRCh38
rs777864836 Chr.20: 45891691-45891691 on Build GRCh38
rs758265394 Chr.20: 45891345-45891345 on Build GRCh38
rs757836846 Chr.20: 45891697-45891697 on Build GRCh38
rs1053560945 Chr.20: 45891551-45891551 on Build GRCh38
rs780220124 Chr.20: 45891470-45891470 on Build GRCh38
rs769772351 Chr.20: 45891599-45891610 on Build GRCh38
rs752201518 Chr.20: 45891618-45891622 on Build GRCh38
rs891163030 Chr.20: 45891736-45891736 on Build GRCh38
rs766204578 Chr.20: 45891677-45891677 on Build GRCh38
rs544157818 Chr.20: 45891623-45891625 on Build GRCh38
rs376350093 Chr.20: 45891653-45891653 on Build GRCh38
rs774681531 Chr.20: 45891609-45891610 on Build GRCh38
rs752927732 Chr.20: 45891680-45891680 on Build GRCh38
rs753979268 Chr.20: 45891786-45891786 on Build GRCh38
rs994209634 Chr.20: 45891668-45891668 on Build GRCh38
rs768166872 Chr.20: 45891428-45891428 on Build GRCh38
rs377390291 Chr.20: 45891372-45891372 on Build GRCh38
rs772808136 Chr.20: 45891670-45891670 on Build GRCh38
rs534135757 Chr.20: 45891528-45891528 on Build GRCh38
rs749819661 Chr.20: 45891621-45891621 on Build GRCh38
rs776723059 Chr.20: 45891613-45891613 on Build GRCh38
rs748154467 Chr.20: 45891367-45891367 on Build GRCh38
rs749726417 Chr.20: 45891386-45891386 on Build GRCh38
rs137854541 Chr.20: 45891714-45891714 on Build GRCh38
rs138615064 Chr.20: 45891728-45891728 on Build GRCh38
rs750393087 Chr.20: 45891531-45891531 on Build GRCh38
rs569267789 Chr.20: 45891411-45891411 on Build GRCh38
rs6104389 Chr.20: 45891436-45891436 on Build GRCh38
rs919085389 Chr.20: 45891467-45891467 on Build GRCh38
rs368879245 Chr.20: 45891682-45891682 on Build GRCh38
rs957153733 Chr.20: 45891569-45891569 on Build GRCh38
rs1018343995 Chr.20: 45891794-45891794 on Build GRCh38
rs770967656 Chr.20: 45891735-45891735 on Build GRCh38
rs752837853 Chr.20: 45891576-45891576 on Build GRCh38
rs188792776 Chr.20: 45891365-45891365 on Build GRCh38
rs762928002 Chr.20: 45891537-45891537 on Build GRCh38
rs781257090 Chr.20: 45891564-45891564 on Build GRCh38
rs746200419 Chr.20: 45891642-45891642 on Build GRCh38
rs755113833 Chr.20: 45891356-45891356 on Build GRCh38
rs775222023 Chr.20: 45891769-45891769 on Build GRCh38
rs757280969 Chr.20: 45891792-45891792 on Build GRCh38
rs769444725 Chr.20: 45891656-45891656 on Build GRCh38
rs761395072 Chr.20: 45891524-45891524 on Build GRCh38
rs566581282 Chr.20: 45891493-45891493 on Build GRCh38
rs540291608 Chr.20: 45891601-45891602 on Build GRCh38
rs770340993 Chr.20: 45891368-45891368 on Build GRCh38
rs910228649 Chr.20: 45891520-45891520 on Build GRCh38
rs576896599 Chr.20: 45891619-45891620 on Build GRCh38
rs753886584 Chr.20: 45891696-45891696 on Build GRCh38
rs538562022 Chr.20: 45891720-45891720 on Build GRCh38
rs1007018466 Chr.20: 45891561-45891561 on Build GRCh38
rs779279066 Chr.20: 45891700-45891700 on Build GRCh38
rs1006892424 Chr.20: 45891540-45891540 on Build GRCh38
rs150140984 Chr.20: 45891673-45891673 on Build GRCh38
rs1003805541 Chr.20: 45891628-45891628 on Build GRCh38
rs989584126 Chr.20: 45891737-45891737 on Build GRCh38
rs749496723 Chr.20: 45891601-45891601 on Build GRCh38
rs1055355836 Chr.20: 45891632-45891632 on Build GRCh38
rs759243010 Chr.20: 45891679-45891679 on Build GRCh38
rs530287837 Chr.20: 45891622-45891622 on Build GRCh38
rs772896149 Chr.20: 45891725-45891725 on Build GRCh38
rs578201110 Chr.20: 45891511-45891511 on Build GRCh38
rs372800409 Chr.20: 45891363-45891363 on Build GRCh38
rs746332491 Chr.20: 45891599-45891607 on Build GRCh38
rs542208835 Chr.20: 45891793-45891793 on Build GRCh38
rs532494728 Chr.20: 45891347-45891347 on Build GRCh38
rs777770494 Chr.20: 45891591-45891591 on Build GRCh38
rs779648193 Chr.20: 45891346-45891346 on Build GRCh38
rs564815597 Chr.20: 45891619-45891619 on Build GRCh38
rs979557944 Chr.20: 45891567-45891567 on Build GRCh38
rs760929674 Chr.20: 45891770-45891770 on Build GRCh38
rs766226288 Chr.20: 45891539-45891539 on Build GRCh38
rs758543261 Chr.20: 45891440-45891440 on Build GRCh38
rs776143304 Chr.20: 45891647-45891647 on Build GRCh38
rs373507043 Chr.20: 45891544-45891544 on Build GRCh38
rs201331162 Chr.20: 45891399-45891399 on Build GRCh38
rs776257120 Chr.20: 45891429-45891429 on Build GRCh38
rs28934603 Chr.20: 45891761-45891761 on Build GRCh38
rs879061920 Chr.20: 45891589-45891589 on Build GRCh38
rs987312318 Chr.20: 45891501-45891501 on Build GRCh38
rs764282392 Chr.20: 45891777-45891777 on Build GRCh38
rs78991921 Chr.20: 45891708-45891708 on Build GRCh38
rs994196870 Chr.20: 45891469-45891469 on Build GRCh38
rs370694703 Chr.20: 45891400-45891400 on Build GRCh38
rs751427255 Chr.20: 45891678-45891678 on Build GRCh38
rs756186589 Chr.20: 45891586-45891586 on Build GRCh38
rs769046478 Chr.20: 45891608-45891609 on Build GRCh38
rs893572063 Chr.20: 45891555-45891555 on Build GRCh38
rs933281472 Chr.20: 45891494-45891494 on Build GRCh38
rs759450806 Chr.20: 45891615-45891622 on Build GRCh38
rs751962489 Chr.20: 45891599-45891604 on Build GRCh38
rs868058474 Chr.20: 45891379-45891379 on Build GRCh38
rs72555383 Chr.20: 45891599-45891601 on Build GRCh38
rs886039898 Chr.20: 45891619-45891622 on Build GRCh38
rs535700546 Chr.20: 45891483-45891483 on Build GRCh38
rs968222440 Chr.20: 45891565-45891565 on Build GRCh38
rs770831518 Chr.20: 45891612-45891612 on Build GRCh38
rs562182818 Chr.20: 45891621-45891622 on Build GRCh38
rs769538991 Chr.20: 45891722-45891722 on Build GRCh38
rs779378519 Chr.20: 45891634-45891634 on Build GRCh38
rs910439232 Chr.20: 45891705-45891705 on Build GRCh38
rs797045505 Chr.20: 45891599-45891624 on Build GRCh38
rs774156870 Chr.20: 45891744-45891744 on Build GRCh38
rs759456788 Chr.20: 45891757-45891757 on Build GRCh38
rs764683890 Chr.20: 45891616-45891616 on Build GRCh38
rs746311995 Chr.20: 45891702-45891702 on Build GRCh38
rs181943893 Chr.20: 45891622-45891622 on Build GRCh38
rs760609872 Chr.20: 45891611-45891611 on Build GRCh38
rs2090364 Chr.20: 45891495-45891495 on Build GRCh38
rs750928198 Chr.20: 45891628-45891628 on Build GRCh38
rs397784956 Chr.20: 45891622-45891624 on Build GRCh38
rs142125327 Chr.20: 45891612-45891612 on Build GRCh38
rs771295717 Chr.20: 45891392-45891392 on Build GRCh38
rs961795659 Chr.20: 45891659-45891659 on Build GRCh38
rs751278595 Chr.20: 45891543-45891543 on Build GRCh38
rs201198067 Chr.20: 45891759-45891759 on Build GRCh38
rs781707830 Chr.20: 45891598-45891599 on Build GRCh38
rs780510269 Chr.20: 45891711-45891711 on Build GRCh38

View Full Product Details