Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46122522-46123023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD40
Entrez Gene ID
958
Gene Name
CD40 molecule
Gene Aliases
Bp50, CDW40, TNFRSF5, p50
UniGene
Hs.472860
Gene Chromosome Location
Chr.20: 46118242-46129745 on Build GRCh38
Transcripts
NM_001250.5 NM_001302753.1 NM_001322421.1 NM_001322422.1 NM_152854.3 NR_126502.1 NR_136327.1 XM_005260619.3 XM_011529109.2 XM_017028135.1 XM_017028136.1 XM_017028137.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3972574 c.294A>G p.E98E Chr.20: 46122647-46122647 on Build GRCh38
127578 c.363G>T p.L121L Chr.20: 46122716-46122716 on Build GRCh38
1235823 c.364C>A p.H122N Chr.20: 46122717-46122717 on Build GRCh38
3963619 c.312C>T p.T104T Chr.20: 46122665-46122665 on Build GRCh38
4430203 c.279G>C p.Q93H Chr.20: 46122632-46122632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74174998 Chr.20: 46122973-46122973 on Build GRCh38
rs371997367 Chr.20: 46122754-46122754 on Build GRCh38
rs774170070 Chr.20: 46122795-46122795 on Build GRCh38
rs768090085 Chr.20: 46122728-46122728 on Build GRCh38
rs144542285 Chr.20: 46122621-46122621 on Build GRCh38
rs969322911 Chr.20: 46122860-46122860 on Build GRCh38
rs267605960 Chr.20: 46122689-46122689 on Build GRCh38
rs771189629 Chr.20: 46122683-46122683 on Build GRCh38
rs933531196 Chr.20: 46122595-46122595 on Build GRCh38
rs555476733 Chr.20: 46122707-46122707 on Build GRCh38
rs771888321 Chr.20: 46122585-46122585 on Build GRCh38
rs933351403 Chr.20: 46122991-46122991 on Build GRCh38
rs1024910207 Chr.20: 46122827-46122827 on Build GRCh38
rs761229326 Chr.20: 46122622-46122622 on Build GRCh38
rs878952715 Chr.20: 46122995-46122995 on Build GRCh38
rs763263083 Chr.20: 46122734-46122734 on Build GRCh38
rs766205073 Chr.20: 46122718-46122718 on Build GRCh38
rs11699100 Chr.20: 46122776-46122776 on Build GRCh38
rs764882779 Chr.20: 46122624-46122624 on Build GRCh38
rs963784749 Chr.20: 46122937-46122937 on Build GRCh38
rs751182588 Chr.20: 46122597-46122601 on Build GRCh38
rs775974865 Chr.20: 46122825-46122825 on Build GRCh38
rs1048633563 Chr.20: 46122880-46122880 on Build GRCh38
rs768637864 Chr.20: 46122612-46122612 on Build GRCh38
rs150312341 Chr.20: 46122731-46122731 on Build GRCh38
rs769831127 Chr.20: 46122788-46122788 on Build GRCh38
rs571974174 Chr.20: 46122597-46122597 on Build GRCh38
rs762684377 Chr.20: 46122711-46122711 on Build GRCh38
rs376448796 Chr.20: 46122587-46122587 on Build GRCh38
rs754006524 Chr.20: 46122735-46122735 on Build GRCh38
rs778017892 Chr.20: 46122680-46122680 on Build GRCh38
rs756171667 Chr.20: 46122678-46122678 on Build GRCh38
rs61760052 Chr.20: 46122753-46122753 on Build GRCh38
rs748815394 Chr.20: 46122793-46122793 on Build GRCh38
rs11569321 Chr.20: 46122724-46122724 on Build GRCh38
rs1015743694 Chr.20: 46122925-46122925 on Build GRCh38
rs763334863 Chr.20: 46122641-46122641 on Build GRCh38
rs537489349 Chr.20: 46122697-46122697 on Build GRCh38
rs752031207 Chr.20: 46122644-46122644 on Build GRCh38
rs752764084 Chr.20: 46122675-46122675 on Build GRCh38
rs972180690 Chr.20: 46122960-46122960 on Build GRCh38
rs767743485 Chr.20: 46122807-46122807 on Build GRCh38
rs772829518 Chr.20: 46122699-46122699 on Build GRCh38
rs888734492 Chr.20: 46122908-46122908 on Build GRCh38
rs755540822 Chr.20: 46122646-46122646 on Build GRCh38
rs377499066 Chr.20: 46122799-46122799 on Build GRCh38
rs886056717 Chr.20: 46122692-46122692 on Build GRCh38
rs749195546 Chr.20: 46122794-46122794 on Build GRCh38
rs761071478 Chr.20: 46122849-46122849 on Build GRCh38
rs147816161 Chr.20: 46122698-46122698 on Build GRCh38
rs143155409 Chr.20: 46122806-46122806 on Build GRCh38
rs757465117 Chr.20: 46122737-46122737 on Build GRCh38
rs564200263 Chr.20: 46122682-46122682 on Build GRCh38
rs762898461 Chr.20: 46122575-46122575 on Build GRCh38
rs748001541 Chr.20: 46122782-46122782 on Build GRCh38
rs539204755 Chr.20: 46122963-46122963 on Build GRCh38
rs143913014 Chr.20: 46122879-46122879 on Build GRCh38
rs779810076 Chr.20: 46122688-46122688 on Build GRCh38
rs777873139 Chr.20: 46122579-46122579 on Build GRCh38
rs369901991 Chr.20: 46122602-46122602 on Build GRCh38
rs199588140 Chr.20: 46122578-46122578 on Build GRCh38
rs781140849 Chr.20: 46122779-46122779 on Build GRCh38
rs772597083 Chr.20: 46122791-46122791 on Build GRCh38
rs569911350 Chr.20: 46122556-46122556 on Build GRCh38
rs578123474 Chr.20: 46122911-46122911 on Build GRCh38
rs751910371 Chr.20: 46122721-46122721 on Build GRCh38
rs764100837 Chr.20: 46122591-46122594 on Build GRCh38
rs753176464 Chr.20: 46122730-46122730 on Build GRCh38
rs781766854 Chr.20: 46122665-46122665 on Build GRCh38
rs750063104 Chr.20: 46122625-46122625 on Build GRCh38
rs1801293 Chr.20: 46122743-46122743 on Build GRCh38

View Full Product Details