Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46128198-46128711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD40
Entrez Gene ID
958
Gene Name
CD40 molecule
Gene Aliases
Bp50, CDW40, TNFRSF5, p50
UniGene
Hs.472860
Gene Chromosome Location
Chr.20: 46118242-46129745 on Build GRCh38
Transcripts
NM_001250.5 NM_001302753.1 NM_001322421.1 NM_001322422.1 NM_152854.3 NR_126502.1 NR_136327.1 XM_005260619.3 XM_017028135.1 XM_017028136.1 XM_017028137.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4594022 c.653T>G p.V218G Chr.20: 46128336-46128336 on Build GRCh38
4822883 c.645C>T p.I215I Chr.20: 46128223-46128223 on Build GRCh38
1661056 c.648A>G p.K216K Chr.20: 46128331-46128331 on Build GRCh38
377918 c.647A>G p.K216R Chr.20: 46128330-46128330 on Build GRCh38
1326700 c.652G>T p.V218L Chr.20: 46128335-46128335 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765889602 Chr.20: 46128249-46128249 on Build GRCh38
rs149938657 Chr.20: 46128483-46128483 on Build GRCh38
rs73310754 Chr.20: 46128379-46128379 on Build GRCh38
rs746007204 Chr.20: 46128531-46128531 on Build GRCh38
rs544008602 Chr.20: 46128301-46128301 on Build GRCh38
rs368055606 Chr.20: 46128389-46128389 on Build GRCh38
rs952713943 Chr.20: 46128668-46128668 on Build GRCh38
rs75892394 Chr.20: 46128519-46128519 on Build GRCh38
rs775731299 Chr.20: 46128611-46128611 on Build GRCh38
rs78782129 Chr.20: 46128329-46128329 on Build GRCh38
rs765214644 Chr.20: 46128238-46128238 on Build GRCh38
rs374618623 Chr.20: 46128271-46128271 on Build GRCh38
rs759022099 Chr.20: 46128522-46128522 on Build GRCh38
rs371799172 Chr.20: 46128342-46128342 on Build GRCh38
rs150736777 Chr.20: 46128416-46128416 on Build GRCh38
rs948282712 Chr.20: 46128609-46128609 on Build GRCh38
rs772360039 Chr.20: 46128302-46128302 on Build GRCh38
rs749590513 Chr.20: 46128304-46128305 on Build GRCh38
rs552022022 Chr.20: 46128455-46128455 on Build GRCh38
rs752316532 Chr.20: 46128367-46128367 on Build GRCh38
rs567876375 Chr.20: 46128508-46128508 on Build GRCh38
rs1029317951 Chr.20: 46128453-46128453 on Build GRCh38
rs565954224 Chr.20: 46128573-46128573 on Build GRCh38
rs750401984 Chr.20: 46128247-46128247 on Build GRCh38
rs749684570 Chr.20: 46128380-46128380 on Build GRCh38
rs775748987 Chr.20: 46128425-46128425 on Build GRCh38
rs554002816 Chr.20: 46128535-46128535 on Build GRCh38
rs760318564 Chr.20: 46128502-46128502 on Build GRCh38
rs745937396 Chr.20: 46128291-46128291 on Build GRCh38
rs896899558 Chr.20: 46128681-46128681 on Build GRCh38
rs200195817 Chr.20: 46128610-46128610 on Build GRCh38
rs781292706 Chr.20: 46128603-46128603 on Build GRCh38
rs3765456 Chr.20: 46128252-46128252 on Build GRCh38
rs762624705 Chr.20: 46128480-46128481 on Build GRCh38
rs368614125 Chr.20: 46128377-46128377 on Build GRCh38
rs750924973 Chr.20: 46128520-46128520 on Build GRCh38
rs756464016 Chr.20: 46128302-46128303 on Build GRCh38
rs549558844 Chr.20: 46128500-46128500 on Build GRCh38
rs750023774 Chr.20: 46128239-46128240 on Build GRCh38
rs923140172 Chr.20: 46128400-46128400 on Build GRCh38
rs766292495 Chr.20: 46128336-46128336 on Build GRCh38
rs780267119 Chr.20: 46128572-46128572 on Build GRCh38
rs942183229 Chr.20: 46128661-46128661 on Build GRCh38
rs768818751 Chr.20: 46128438-46128438 on Build GRCh38
rs774187457 Chr.20: 46128473-46128473 on Build GRCh38
rs11569337 Chr.20: 46128251-46128251 on Build GRCh38
rs11569341 Chr.20: 46128488-46128488 on Build GRCh38
rs3765457 Chr.20: 46128574-46128574 on Build GRCh38
rs759459933 Chr.20: 46128662-46128662 on Build GRCh38
rs754507112 Chr.20: 46128343-46128343 on Build GRCh38
rs4813000 Chr.20: 46128269-46128269 on Build GRCh38
rs758308480 Chr.20: 46128248-46128248 on Build GRCh38
rs905363044 Chr.20: 46128590-46128590 on Build GRCh38
rs749773841 Chr.20: 46128305-46128305 on Build GRCh38
rs761656711 Chr.20: 46128305-46128305 on Build GRCh38
rs11569342 Chr.20: 46128650-46128650 on Build GRCh38
rs780836857 Chr.20: 46128263-46128263 on Build GRCh38
rs772084859 Chr.20: 46128305-46128306 on Build GRCh38
rs778043845 Chr.20: 46128270-46128270 on Build GRCh38
rs771402513 Chr.20: 46128282-46128282 on Build GRCh38
rs779297695 Chr.20: 46128411-46128411 on Build GRCh38
rs1052684216 Chr.20: 46128513-46128513 on Build GRCh38
rs780414883 Chr.20: 46128303-46128304 on Build GRCh38
rs867188253 Chr.20: 46128469-46128469 on Build GRCh38
rs767118191 Chr.20: 46128355-46128355 on Build GRCh38
rs11569340 Chr.20: 46128475-46128475 on Build GRCh38
rs75736856 Chr.20: 46128328-46128328 on Build GRCh38
rs747210864 Chr.20: 46128427-46128427 on Build GRCh38
rs868675588 Chr.20: 46128596-46128596 on Build GRCh38
rs765092482 Chr.20: 46128327-46128327 on Build GRCh38
rs770601285 Chr.20: 46128633-46128633 on Build GRCh38
rs775404861 Chr.20: 46128401-46128401 on Build GRCh38
rs761711766 Chr.20: 46128232-46128232 on Build GRCh38
rs11569338 Chr.20: 46128264-46128264 on Build GRCh38
rs779391263 Chr.20: 46128467-46128467 on Build GRCh38
rs527246432 Chr.20: 46128417-46128417 on Build GRCh38
rs371353981 Chr.20: 46128346-46128346 on Build GRCh38
rs760912929 Chr.20: 46128310-46128311 on Build GRCh38
rs757582967 Chr.20: 46128510-46128510 on Build GRCh38
rs774486667 Chr.20: 46128287-46128287 on Build GRCh38
rs1005683552 Chr.20: 46128305-46128316 on Build GRCh38
rs11569339 Chr.20: 46128303-46128303 on Build GRCh38
rs368619894 Chr.20: 46128338-46128338 on Build GRCh38
rs769499805 Chr.20: 46128327-46128328 on Build GRCh38

View Full Product Details