Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46128894-46129389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD40
Entrez Gene ID
958
Gene Name
CD40 molecule
Gene Aliases
Bp50, CDW40, TNFRSF5, p50
UniGene
Hs.472860
Gene Chromosome Location
Chr.20: 46118242-46129745 on Build GRCh38
Transcripts
NM_001250.5 NM_001302753.1 NM_001322421.1 NM_001322422.1 NM_152854.3 NR_126502.1 NR_136327.1 XM_005260619.3 XM_017028135.1 XM_017028136.1 XM_017028137.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3294375 c.809G>A p.R270H Chr.20: 46129015-46129015 on Build GRCh38
5468989 c.754C>G p.Q252E Chr.20: 46128960-46128960 on Build GRCh38
6623771 c.690G>A p.K230K Chr.20: 46128896-46128896 on Build GRCh38
5762738 c.799A>G p.K267E Chr.20: 46129005-46129005 on Build GRCh38
3389794 c.717C>T p.P239P Chr.20: 46128923-46128923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776099682 Chr.20: 46128989-46128989 on Build GRCh38
rs753974727 Chr.20: 46129025-46129025 on Build GRCh38
rs913525522 Chr.20: 46129044-46129044 on Build GRCh38
rs369546762 Chr.20: 46129237-46129237 on Build GRCh38
rs1052340852 Chr.20: 46129321-46129321 on Build GRCh38
rs746182207 Chr.20: 46129159-46129159 on Build GRCh38
rs755836869 Chr.20: 46129102-46129104 on Build GRCh38
rs778653844 Chr.20: 46129140-46129140 on Build GRCh38
rs957745543 Chr.20: 46129004-46129004 on Build GRCh38
rs926902514 Chr.20: 46129156-46129156 on Build GRCh38
rs139300926 Chr.20: 46129015-46129015 on Build GRCh38
rs772009573 Chr.20: 46129346-46129346 on Build GRCh38
rs777419143 Chr.20: 46129120-46129120 on Build GRCh38
rs150890139 Chr.20: 46128986-46128986 on Build GRCh38
rs780950226 Chr.20: 46129105-46129105 on Build GRCh38
rs776624029 Chr.20: 46129171-46129171 on Build GRCh38
rs199980487 Chr.20: 46128960-46128960 on Build GRCh38
rs147672904 Chr.20: 46128926-46128926 on Build GRCh38
rs376780996 Chr.20: 46128956-46128956 on Build GRCh38
rs1016241905 Chr.20: 46129363-46129363 on Build GRCh38
rs748815275 Chr.20: 46129137-46129137 on Build GRCh38
rs759961210 Chr.20: 46128985-46128985 on Build GRCh38
rs751626954 Chr.20: 46129056-46129056 on Build GRCh38
rs777475990 Chr.20: 46128977-46128977 on Build GRCh38
rs940067560 Chr.20: 46129192-46129192 on Build GRCh38
rs779807906 Chr.20: 46128936-46128936 on Build GRCh38
rs754026379 Chr.20: 46128929-46128929 on Build GRCh38
rs963361078 Chr.20: 46129364-46129364 on Build GRCh38
rs762015042 Chr.20: 46129060-46129060 on Build GRCh38
rs761892578 Chr.20: 46129173-46129173 on Build GRCh38
rs748935911 Chr.20: 46128980-46128980 on Build GRCh38
rs936546325 Chr.20: 46129302-46129302 on Build GRCh38
rs757516011 Chr.20: 46128934-46128934 on Build GRCh38
rs886056718 Chr.20: 46129125-46129125 on Build GRCh38
rs774515822 Chr.20: 46128984-46128984 on Build GRCh38
rs570025940 Chr.20: 46129304-46129304 on Build GRCh38
rs539494893 Chr.20: 46129227-46129227 on Build GRCh38
rs537564344 Chr.20: 46129318-46129318 on Build GRCh38
rs987910986 Chr.20: 46129009-46129009 on Build GRCh38
rs985984295 Chr.20: 46128945-46128945 on Build GRCh38
rs765500316 Chr.20: 46129068-46129068 on Build GRCh38
rs770653224 Chr.20: 46129138-46129138 on Build GRCh38
rs565941222 Chr.20: 46129218-46129218 on Build GRCh38
rs6131019 Chr.20: 46129130-46129130 on Build GRCh38
rs11569344 Chr.20: 46129356-46129356 on Build GRCh38
rs982770421 Chr.20: 46129154-46129154 on Build GRCh38
rs750063120 Chr.20: 46129007-46129008 on Build GRCh38
rs555863322 Chr.20: 46129342-46129342 on Build GRCh38
rs769214684 Chr.20: 46129169-46129169 on Build GRCh38
rs754823732 Chr.20: 46129090-46129090 on Build GRCh38
rs998235214 Chr.20: 46129256-46129256 on Build GRCh38
rs551789939 Chr.20: 46129269-46129269 on Build GRCh38
rs767094199 Chr.20: 46128968-46128969 on Build GRCh38
rs747561680 Chr.20: 46128961-46128961 on Build GRCh38
rs752657781 Chr.20: 46129112-46129112 on Build GRCh38
rs529049263 Chr.20: 46129069-46129069 on Build GRCh38
rs988084488 Chr.20: 46129198-46129198 on Build GRCh38
rs761207472 Chr.20: 46129010-46129010 on Build GRCh38
rs756178814 Chr.20: 46129119-46129119 on Build GRCh38
rs11569343 Chr.20: 46129175-46129175 on Build GRCh38
rs1037125026 Chr.20: 46129216-46129216 on Build GRCh38
rs889065677 Chr.20: 46129350-46129350 on Build GRCh38
rs903736094 Chr.20: 46128940-46128940 on Build GRCh38
rs942453778 Chr.20: 46129238-46129238 on Build GRCh38
rs781200622 Chr.20: 46128957-46128957 on Build GRCh38
rs886056719 Chr.20: 46129256-46129256 on Build GRCh38
rs775762947 Chr.20: 46129167-46129167 on Build GRCh38
rs886056720 Chr.20: 46129350-46129350 on Build GRCh38
rs772447675 Chr.20: 46129160-46129160 on Build GRCh38
rs41282790 Chr.20: 46129351-46129351 on Build GRCh38
rs768078284 Chr.20: 46128916-46128916 on Build GRCh38
rs751437274 Chr.20: 46128949-46128949 on Build GRCh38
rs369693842 Chr.20: 46128983-46128983 on Build GRCh38
rs769399729 Chr.20: 46128973-46128973 on Build GRCh38
rs371282411 Chr.20: 46129306-46129306 on Build GRCh38
rs144466131 Chr.20: 46128923-46128923 on Build GRCh38
rs761154625 Chr.20: 46129168-46129168 on Build GRCh38
rs764257725 Chr.20: 46128927-46128927 on Build GRCh38

View Full Product Details