Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50191822-50192330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPB
Entrez Gene ID
1051
Gene Name
CCAAT/enhancer binding protein beta
Gene Aliases
C/EBP-beta, IL6DBP, NF-IL6, TCF5
UniGene
Hs.517106
Gene Chromosome Location
Chr.20: 50190583-50192690 on Build GRCh38
Transcripts
NM_001285878.1 NM_001285879.1 NM_005194.3 NR_125739.1

Gene Symbol
CEBPB-AS1
Entrez Gene ID
101927559
Gene Name
CEBPB antisense RNA 1
Gene Aliases
-
UniGene
Hs.442486
Gene Chromosome Location
Chr.20: 50184603-50192069 on Build GRCh38
Transcripts
NM_001285878.1 NM_001285879.1 NM_005194.3 NR_125739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2762518 c.909C>G p.V303V Chr.20: 50191942-50191942 on Build GRCh38
1412416 c.879G>T p.K293N Chr.20: 50191912-50191912 on Build GRCh38
724219 c.906G>A p.K302K Chr.20: 50191939-50191939 on Build GRCh38
6999511 c.925G>T p.E309* Chr.20: 50191958-50191958 on Build GRCh38
1307515 c.927G>A p.E309E Chr.20: 50191960-50191960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767096943 Chr.20: 50192062-50192062 on Build GRCh38
rs775696986 Chr.20: 50192103-50192103 on Build GRCh38
rs775399191 Chr.20: 50192014-50192014 on Build GRCh38
rs759627700 Chr.20: 50191885-50191885 on Build GRCh38
rs200435791 Chr.20: 50191957-50191957 on Build GRCh38
rs919642184 Chr.20: 50192219-50192219 on Build GRCh38
rs774845694 Chr.20: 50191999-50191999 on Build GRCh38
rs991197826 Chr.20: 50192158-50192158 on Build GRCh38
rs749640653 Chr.20: 50191988-50191988 on Build GRCh38
rs898163623 Chr.20: 50192008-50192008 on Build GRCh38
rs780522860 Chr.20: 50191948-50191948 on Build GRCh38
rs891398872 Chr.20: 50192114-50192114 on Build GRCh38
rs1017293001 Chr.20: 50192293-50192293 on Build GRCh38
rs754052559 Chr.20: 50192047-50192047 on Build GRCh38
rs915880813 Chr.20: 50192117-50192117 on Build GRCh38
rs200365245 Chr.20: 50191900-50191900 on Build GRCh38
rs868421494 Chr.20: 50191916-50191916 on Build GRCh38
rs1008414622 Chr.20: 50192119-50192119 on Build GRCh38
rs758466280 Chr.20: 50191918-50191918 on Build GRCh38
rs777416244 Chr.20: 50192077-50192077 on Build GRCh38
rs779200975 Chr.20: 50192098-50192098 on Build GRCh38
rs772450886 Chr.20: 50192102-50192102 on Build GRCh38
rs759997646 Chr.20: 50192005-50192005 on Build GRCh38
rs751561149 Chr.20: 50192054-50192054 on Build GRCh38
rs1049883526 Chr.20: 50192256-50192259 on Build GRCh38
rs757651622 Chr.20: 50192092-50192092 on Build GRCh38
rs771228331 Chr.20: 50191991-50191991 on Build GRCh38
rs746535168 Chr.20: 50192160-50192160 on Build GRCh38
rs1007707663 Chr.20: 50192255-50192255 on Build GRCh38
rs756312550 Chr.20: 50192097-50192097 on Build GRCh38
rs939796566 Chr.20: 50192209-50192209 on Build GRCh38
rs1008467942 Chr.20: 50192282-50192282 on Build GRCh38
rs751473169 Chr.20: 50191942-50191942 on Build GRCh38
rs929437390 Chr.20: 50192191-50192191 on Build GRCh38
rs932407115 Chr.20: 50192227-50192228 on Build GRCh38
rs926101428 Chr.20: 50191981-50191981 on Build GRCh38
rs867148328 Chr.20: 50192015-50192015 on Build GRCh38
rs1021117795 Chr.20: 50192122-50192127 on Build GRCh38
rs966015956 Chr.20: 50191882-50191882 on Build GRCh38
rs747071489 Chr.20: 50192120-50192120 on Build GRCh38
rs747887242 Chr.20: 50191954-50191954 on Build GRCh38
rs574993056 Chr.20: 50191906-50191906 on Build GRCh38
rs774473666 Chr.20: 50191845-50191845 on Build GRCh38
rs560057016 Chr.20: 50192212-50192212 on Build GRCh38
rs750238105 Chr.20: 50191912-50191912 on Build GRCh38
rs890793877 Chr.20: 50192274-50192274 on Build GRCh38
rs772241254 Chr.20: 50192012-50192012 on Build GRCh38
rs200273644 Chr.20: 50192209-50192209 on Build GRCh38
rs529249404 Chr.20: 50192143-50192143 on Build GRCh38
rs561942729 Chr.20: 50192088-50192088 on Build GRCh38
rs201972813 Chr.20: 50191924-50191924 on Build GRCh38
rs745903016 Chr.20: 50192076-50192077 on Build GRCh38
rs761024259 Chr.20: 50192028-50192028 on Build GRCh38
rs778180638 Chr.20: 50191985-50191985 on Build GRCh38
rs966842459 Chr.20: 50192133-50192133 on Build GRCh38
rs766233869 Chr.20: 50192051-50192051 on Build GRCh38
rs767430489 Chr.20: 50191891-50191891 on Build GRCh38
rs746426283 Chr.20: 50192099-50192099 on Build GRCh38
rs45491391 Chr.20: 50192210-50192210 on Build GRCh38
rs898006913 Chr.20: 50192151-50192151 on Build GRCh38
rs762842653 Chr.20: 50192048-50192048 on Build GRCh38
rs754545518 Chr.20: 50191945-50191945 on Build GRCh38
rs754851484 Chr.20: 50192056-50192056 on Build GRCh38
rs887567829 Chr.20: 50192251-50192251 on Build GRCh38
rs1050023817 Chr.20: 50192081-50192082 on Build GRCh38
rs971345491 Chr.20: 50192188-50192188 on Build GRCh38
rs949986667 Chr.20: 50191847-50191847 on Build GRCh38
rs368807189 Chr.20: 50192038-50192038 on Build GRCh38
rs755856576 Chr.20: 50192065-50192065 on Build GRCh38
rs917908550 Chr.20: 50192162-50192162 on Build GRCh38
rs45554839 Chr.20: 50192211-50192211 on Build GRCh38
rs777282272 Chr.20: 50192239-50192239 on Build GRCh38
rs1032800115 Chr.20: 50192154-50192154 on Build GRCh38

View Full Product Details