Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50589594-50590094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
UniGene
Hs.372578
Gene Chromosome Location
Chr.20: 50586108-50692004 on Build GRCh38
Transcripts
NM_001290268.1 NM_080829.3 NR_110890.1 XM_005260294.3 XM_006723713.3 XM_011528578.2 XM_011528579.2 XM_011528580.2 XM_011528581.2 XM_011528585.2 XM_017027681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644592 c.2604C>G p.D868E Chr.20: 50589731-50589731 on Build GRCh38
4434175 c.2637C>T p.L879L Chr.20: 50589698-50589698 on Build GRCh38
1615721 c.2566G>C p.A856P Chr.20: 50589769-50589769 on Build GRCh38
1615720 c.2586C>T p.N862N Chr.20: 50589749-50589749 on Build GRCh38
3740175 c.2585A>T p.N862I Chr.20: 50589750-50589750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755293539 Chr.20: 50589751-50589751 on Build GRCh38
rs769568139 Chr.20: 50589817-50589817 on Build GRCh38
rs976310366 Chr.20: 50589948-50589948 on Build GRCh38
rs940968412 Chr.20: 50589702-50589702 on Build GRCh38
rs748256451 Chr.20: 50589681-50589681 on Build GRCh38
rs1055033130 Chr.20: 50589827-50589827 on Build GRCh38
rs201535929 Chr.20: 50589730-50589730 on Build GRCh38
rs372668224 Chr.20: 50589781-50589781 on Build GRCh38
rs902920019 Chr.20: 50590044-50590044 on Build GRCh38
rs531976013 Chr.20: 50589944-50589944 on Build GRCh38
rs527900156 Chr.20: 50590060-50590060 on Build GRCh38
rs749539240 Chr.20: 50589771-50589771 on Build GRCh38
rs776745638 Chr.20: 50589796-50589796 on Build GRCh38
rs750048177 Chr.20: 50589644-50589644 on Build GRCh38
rs747434914 Chr.20: 50589682-50589682 on Build GRCh38
rs763204438 Chr.20: 50589701-50589701 on Build GRCh38
rs547366377 Chr.20: 50590059-50590059 on Build GRCh38
rs879861079 Chr.20: 50589652-50589652 on Build GRCh38
rs967328274 Chr.20: 50589631-50589631 on Build GRCh38
rs773355508 Chr.20: 50589690-50589690 on Build GRCh38
rs1013527986 Chr.20: 50589851-50589851 on Build GRCh38
rs1015239630 Chr.20: 50589875-50589875 on Build GRCh38
rs375508730 Chr.20: 50589749-50589749 on Build GRCh38
rs183977103 Chr.20: 50589713-50589713 on Build GRCh38
rs945436901 Chr.20: 50590021-50590021 on Build GRCh38
rs369796961 Chr.20: 50589755-50589755 on Build GRCh38
rs141783722 Chr.20: 50589901-50589901 on Build GRCh38
rs761800960 Chr.20: 50589809-50589809 on Build GRCh38
rs930872988 Chr.20: 50589993-50589993 on Build GRCh38
rs879377013 Chr.20: 50589653-50589653 on Build GRCh38
rs116847542 Chr.20: 50589731-50589731 on Build GRCh38
rs748566083 Chr.20: 50589806-50589806 on Build GRCh38
rs543815868 Chr.20: 50589879-50589879 on Build GRCh38
rs758725688 Chr.20: 50589647-50589647 on Build GRCh38
rs985800860 Chr.20: 50590008-50590009 on Build GRCh38
rs774655122 Chr.20: 50589787-50589787 on Build GRCh38
rs560723233 Chr.20: 50590040-50590040 on Build GRCh38
rs542834572 Chr.20: 50589979-50589979 on Build GRCh38
rs866506047 Chr.20: 50589753-50589753 on Build GRCh38
rs1035205951 Chr.20: 50589923-50589923 on Build GRCh38
rs576383011 Chr.20: 50589829-50589829 on Build GRCh38
rs558119832 Chr.20: 50589769-50589769 on Build GRCh38
rs774505345 Chr.20: 50589864-50589864 on Build GRCh38
rs188302508 Chr.20: 50590028-50590028 on Build GRCh38
rs540553339 Chr.20: 50589748-50589748 on Build GRCh38
rs879452076 Chr.20: 50589935-50589935 on Build GRCh38
rs945417676 Chr.20: 50589800-50589800 on Build GRCh38
rs982125088 Chr.20: 50589922-50589922 on Build GRCh38
rs769161590 Chr.20: 50589674-50589674 on Build GRCh38
rs745880935 Chr.20: 50589689-50589690 on Build GRCh38
rs903682506 Chr.20: 50589811-50589811 on Build GRCh38
rs867186894 Chr.20: 50589890-50589890 on Build GRCh38
rs1010903147 Chr.20: 50589891-50589891 on Build GRCh38
rs749956543 Chr.20: 50589821-50589821 on Build GRCh38
rs761255631 Chr.20: 50589733-50589733 on Build GRCh38
rs549164783 Chr.20: 50590049-50590049 on Build GRCh38
rs760566652 Chr.20: 50589788-50589788 on Build GRCh38
rs373403544 Chr.20: 50589654-50589654 on Build GRCh38
rs949250883 Chr.20: 50590064-50590064 on Build GRCh38
rs564927382 Chr.20: 50589642-50589642 on Build GRCh38
rs769549112 Chr.20: 50589734-50589736 on Build GRCh38
rs372296012 Chr.20: 50589675-50589675 on Build GRCh38
rs780839566 Chr.20: 50589637-50589640 on Build GRCh38
rs769233161 Chr.20: 50589720-50589720 on Build GRCh38
rs920841086 Chr.20: 50589710-50589710 on Build GRCh38
rs771829904 Chr.20: 50589703-50589703 on Build GRCh38
rs369475644 Chr.20: 50589828-50589828 on Build GRCh38
rs552125977 Chr.20: 50590032-50590032 on Build GRCh38
rs368271108 Chr.20: 50589667-50589667 on Build GRCh38
rs764786515 Chr.20: 50589734-50589734 on Build GRCh38
rs74561863 Chr.20: 50589945-50589945 on Build GRCh38
rs374324537 Chr.20: 50589760-50589760 on Build GRCh38
rs750106125 Chr.20: 50589739-50589739 on Build GRCh38
rs986338581 Chr.20: 50589939-50589939 on Build GRCh38
rs762696548 Chr.20: 50589905-50589905 on Build GRCh38
rs913865444 Chr.20: 50589973-50589973 on Build GRCh38
rs374774516 Chr.20: 50589791-50589791 on Build GRCh38
rs769813417 Chr.20: 50589768-50589768 on Build GRCh38
rs775413820 Chr.20: 50589706-50589706 on Build GRCh38

View Full Product Details