Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50601837-50602359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
UniGene
Hs.372578
Gene Chromosome Location
Chr.20: 50586108-50692004 on Build GRCh38
Transcripts
NM_001290268.1 NM_080829.3 NR_110890.1 XM_005260294.3 XM_006723713.3 XM_011528578.2 XM_011528579.2 XM_011528580.2 XM_011528581.2 XM_011528584.2 XM_011528585.2 XM_011528586.2 XM_017027681.1 XM_017027682.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6426142 c.1619A>G p.Q540R Chr.20: 50602100-50602100 on Build GRCh38
6412990 c.1502C>T p.A501V Chr.20: 50602217-50602217 on Build GRCh38
4099331 c.1444G>T p.G482C Chr.20: 50602275-50602275 on Build GRCh38
4099330 c.1461C>T p.S487S Chr.20: 50602258-50602258 on Build GRCh38
5974093 c.1427G>A p.S476N Chr.20: 50602292-50602292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73272554 Chr.20: 50601863-50601863 on Build GRCh38
rs900713457 Chr.20: 50601966-50601966 on Build GRCh38
rs752844500 Chr.20: 50602197-50602197 on Build GRCh38
rs577488890 Chr.20: 50602086-50602086 on Build GRCh38
rs761448115 Chr.20: 50602085-50602085 on Build GRCh38
rs975349726 Chr.20: 50602045-50602045 on Build GRCh38
rs758270641 Chr.20: 50602211-50602211 on Build GRCh38
rs367985662 Chr.20: 50602179-50602179 on Build GRCh38
rs764576044 Chr.20: 50602115-50602115 on Build GRCh38
rs774013116 Chr.20: 50602300-50602300 on Build GRCh38
rs776106672 Chr.20: 50602246-50602246 on Build GRCh38
rs754217023 Chr.20: 50602119-50602119 on Build GRCh38
rs769469827 Chr.20: 50602173-50602173 on Build GRCh38
rs1011101381 Chr.20: 50602080-50602080 on Build GRCh38
rs756420215 Chr.20: 50602134-50602134 on Build GRCh38
rs568143686 Chr.20: 50601907-50601907 on Build GRCh38
rs772155963 Chr.20: 50602063-50602063 on Build GRCh38
rs988512381 Chr.20: 50601989-50601989 on Build GRCh38
rs748305817 Chr.20: 50602055-50602055 on Build GRCh38
rs147584980 Chr.20: 50602247-50602247 on Build GRCh38
rs775362353 Chr.20: 50602293-50602295 on Build GRCh38
rs78064464 Chr.20: 50602333-50602333 on Build GRCh38
rs779521133 Chr.20: 50602257-50602257 on Build GRCh38
rs763265423 Chr.20: 50602093-50602093 on Build GRCh38
rs999877478 Chr.20: 50602310-50602310 on Build GRCh38
rs267605991 Chr.20: 50602175-50602175 on Build GRCh38
rs561715087 Chr.20: 50602328-50602328 on Build GRCh38
rs949423848 Chr.20: 50601915-50601915 on Build GRCh38
rs962615097 Chr.20: 50602042-50602042 on Build GRCh38
rs761901198 Chr.20: 50602334-50602334 on Build GRCh38
rs549214415 Chr.20: 50602092-50602092 on Build GRCh38
rs1003462118 Chr.20: 50602113-50602113 on Build GRCh38
rs751019874 Chr.20: 50601867-50601868 on Build GRCh38
rs1048165868 Chr.20: 50601943-50601943 on Build GRCh38
rs755147742 Chr.20: 50602116-50602116 on Build GRCh38
rs1023575140 Chr.20: 50601944-50601944 on Build GRCh38
rs747697920 Chr.20: 50602225-50602225 on Build GRCh38
rs775249600 Chr.20: 50602065-50602065 on Build GRCh38
rs373678317 Chr.20: 50602050-50602050 on Build GRCh38
rs761252784 Chr.20: 50602164-50602164 on Build GRCh38
rs548799196 Chr.20: 50601950-50601950 on Build GRCh38
rs772538647 Chr.20: 50602305-50602305 on Build GRCh38
rs890609070 Chr.20: 50602143-50602143 on Build GRCh38
rs149659268 Chr.20: 50602273-50602273 on Build GRCh38
rs754772770 Chr.20: 50602261-50602261 on Build GRCh38
rs760026090 Chr.20: 50602044-50602049 on Build GRCh38
rs894041455 Chr.20: 50601906-50601906 on Build GRCh38
rs535587278 Chr.20: 50601945-50601945 on Build GRCh38
rs756219778 Chr.20: 50602198-50602198 on Build GRCh38
rs759898087 Chr.20: 50602237-50602237 on Build GRCh38
rs745685220 Chr.20: 50602139-50602139 on Build GRCh38
rs770515279 Chr.20: 50602299-50602299 on Build GRCh38
rs765419339 Chr.20: 50602337-50602337 on Build GRCh38
rs774295149 Chr.20: 50602059-50602059 on Build GRCh38
rs752760847 Chr.20: 50602125-50602125 on Build GRCh38
rs770917317 Chr.20: 50602058-50602058 on Build GRCh38
rs181586857 Chr.20: 50601904-50601904 on Build GRCh38
rs766951532 Chr.20: 50602107-50602107 on Build GRCh38
rs141361182 Chr.20: 50602180-50602180 on Build GRCh38
rs775759564 Chr.20: 50601960-50601960 on Build GRCh38
rs746907602 Chr.20: 50602153-50602153 on Build GRCh38
rs781526412 Chr.20: 50602054-50602054 on Build GRCh38
rs754087238 Chr.20: 50602210-50602210 on Build GRCh38
rs376794842 Chr.20: 50602183-50602183 on Build GRCh38
rs150809495 Chr.20: 50602213-50602213 on Build GRCh38
rs369361044 Chr.20: 50602274-50602274 on Build GRCh38
rs760271998 Chr.20: 50602068-50602068 on Build GRCh38
rs780379658 Chr.20: 50602049-50602049 on Build GRCh38
rs566907555 Chr.20: 50601981-50601981 on Build GRCh38
rs768013136 Chr.20: 50602189-50602189 on Build GRCh38
rs894001197 Chr.20: 50602075-50602075 on Build GRCh38
rs6096021 Chr.20: 50602234-50602234 on Build GRCh38
rs776399232 Chr.20: 50602079-50602079 on Build GRCh38
rs186308971 Chr.20: 50602212-50602212 on Build GRCh38
rs375187412 Chr.20: 50602251-50602251 on Build GRCh38
rs887137638 Chr.20: 50602292-50602292 on Build GRCh38
rs1035492403 Chr.20: 50601968-50601968 on Build GRCh38
rs777272607 Chr.20: 50602286-50602286 on Build GRCh38
rs529861332 Chr.20: 50602194-50602194 on Build GRCh38
rs767938043 Chr.20: 50602243-50602243 on Build GRCh38
rs909749052 Chr.20: 50602022-50602022 on Build GRCh38
rs951501620 Chr.20: 50601976-50601976 on Build GRCh38
rs754179429 Chr.20: 50602252-50602252 on Build GRCh38
rs780084072 Chr.20: 50602152-50602152 on Build GRCh38
rs748939000 Chr.20: 50602233-50602233 on Build GRCh38
rs558951276 Chr.20: 50602053-50602053 on Build GRCh38
rs140418486 Chr.20: 50602267-50602267 on Build GRCh38
rs1000078272 Chr.20: 50601870-50601870 on Build GRCh38
rs543434720 Chr.20: 50602316-50602316 on Build GRCh38
rs917188923 Chr.20: 50602059-50602059 on Build GRCh38
rs765370813 Chr.20: 50602290-50602291 on Build GRCh38
rs768025692 Chr.20: 50602071-50602071 on Build GRCh38
rs368287723 Chr.20: 50602258-50602258 on Build GRCh38
rs545997871 Chr.20: 50602228-50602228 on Build GRCh38
rs927137340 Chr.20: 50601886-50601886 on Build GRCh38
rs139663528 Chr.20: 50602232-50602232 on Build GRCh38
rs778177803 Chr.20: 50602207-50602207 on Build GRCh38
rs937147480 Chr.20: 50601901-50601901 on Build GRCh38
rs771729586 Chr.20: 50602142-50602142 on Build GRCh38
rs148677587 Chr.20: 50602162-50602162 on Build GRCh38
rs748777109 Chr.20: 50602287-50602287 on Build GRCh38
rs761258423 Chr.20: 50602319-50602319 on Build GRCh38
rs776193130 Chr.20: 50602314-50602314 on Build GRCh38
rs762026467 Chr.20: 50602156-50602156 on Build GRCh38
rs189577349 Chr.20: 50601898-50601898 on Build GRCh38
rs572110299 Chr.20: 50602227-50602227 on Build GRCh38
rs144244193 Chr.20: 50602154-50602154 on Build GRCh38
rs201413126 Chr.20: 50602138-50602138 on Build GRCh38
rs746255705 Chr.20: 50602301-50602301 on Build GRCh38
rs370389293 Chr.20: 50602188-50602188 on Build GRCh38

View Full Product Details