Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56392283-56392786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKA
Entrez Gene ID
6790
Gene Name
aurora kinase A
Gene Aliases
AIK, ARK1, AURA, BTAK, PPP1R47, STK15, STK6, STK7
UniGene
Hs.250822
Gene Chromosome Location
Chr.20: 56369389-56392337 on Build GRCh38
Transcripts
NM_001323303.1 NM_001323304.1 NM_001323305.1 NM_003600.3 NM_198433.2 NM_198434.2 NM_198435.2 NM_198436.2 NM_198437.2 XM_017028034.1 NM_001033521.1 NM_001033522.1 NM_001324.2 XM_011528600.1

Gene Symbol
CSTF1
Entrez Gene ID
1477
Gene Name
cleavage stimulation factor subunit 1
Gene Aliases
CstF-50, CstFp50
UniGene
Hs.172865
Gene Chromosome Location
Chr.20: 56392371-56404526 on Build GRCh38
Transcripts
NM_001323303.1 NM_001323304.1 NM_001323305.1 NM_003600.3 NM_198433.2 NM_198434.2 NM_198435.2 NM_198436.2 NM_198437.2 XM_017028034.1 NM_001033521.1 NM_001033522.1 NM_001324.2 XM_011528600.1

SNP Detail

SNP ID Chromosome Location
rs553728383 Chr.20: 56392386-56392386 on Build GRCh38
rs953384710 Chr.20: 56392316-56392316 on Build GRCh38
rs750227183 Chr.20: 56392526-56392526 on Build GRCh38
rs766611348 Chr.20: 56392569-56392569 on Build GRCh38
rs928717520 Chr.20: 56392734-56392734 on Build GRCh38
rs1049661582 Chr.20: 56392483-56392483 on Build GRCh38
rs763618816 Chr.20: 56392454-56392454 on Build GRCh38
rs538384267 Chr.20: 56392317-56392317 on Build GRCh38
rs991915001 Chr.20: 56392746-56392746 on Build GRCh38
rs958535724 Chr.20: 56392649-56392649 on Build GRCh38
rs1025043178 Chr.20: 56392598-56392598 on Build GRCh38
rs989124182 Chr.20: 56392652-56392652 on Build GRCh38
rs537472945 Chr.20: 56392356-56392356 on Build GRCh38
rs940717440 Chr.20: 56392361-56392361 on Build GRCh38
rs970918081 Chr.20: 56392618-56392618 on Build GRCh38
rs975106202 Chr.20: 56392443-56392443 on Build GRCh38
rs913616347 Chr.20: 56392417-56392417 on Build GRCh38
rs746299132 Chr.20: 56392621-56392621 on Build GRCh38
rs952574250 Chr.20: 56392489-56392489 on Build GRCh38
rs938739056 Chr.20: 56392743-56392743 on Build GRCh38
rs939822576 Chr.20: 56392514-56392514 on Build GRCh38
rs576943598 Chr.20: 56392387-56392387 on Build GRCh38
rs532757185 Chr.20: 56392578-56392578 on Build GRCh38
rs534467065 Chr.20: 56392399-56392399 on Build GRCh38
rs999015762 Chr.20: 56392646-56392646 on Build GRCh38
rs3044185 Chr.20: 56392564-56392567 on Build GRCh38
rs1040873120 Chr.20: 56392438-56392438 on Build GRCh38
rs35258222 Chr.20: 56392391-56392391 on Build GRCh38
rs983957622 Chr.20: 56392491-56392491 on Build GRCh38
rs568617623 Chr.20: 56392643-56392643 on Build GRCh38
rs755054652 Chr.20: 56392713-56392713 on Build GRCh38
rs981959013 Chr.20: 56392731-56392731 on Build GRCh38
rs761657926 Chr.20: 56392480-56392480 on Build GRCh38
rs35594166 Chr.20: 56392653-56392653 on Build GRCh38
rs567714294 Chr.20: 56392680-56392680 on Build GRCh38
rs545202900 Chr.20: 56392393-56392393 on Build GRCh38
rs762808037 Chr.20: 56392629-56392629 on Build GRCh38
rs568580175 Chr.20: 56392348-56392348 on Build GRCh38
rs902957089 Chr.20: 56392410-56392410 on Build GRCh38
rs909304391 Chr.20: 56392349-56392349 on Build GRCh38
rs2236207 Chr.20: 56392509-56392509 on Build GRCh38
rs981383110 Chr.20: 56392642-56392642 on Build GRCh38
rs34579616 Chr.20: 56392404-56392404 on Build GRCh38
rs890323143 Chr.20: 56392735-56392747 on Build GRCh38
rs188855328 Chr.20: 56392419-56392419 on Build GRCh38
rs752574426 Chr.20: 56392401-56392401 on Build GRCh38
rs1011707572 Chr.20: 56392403-56392403 on Build GRCh38
rs932491788 Chr.20: 56392691-56392691 on Build GRCh38
rs972581353 Chr.20: 56392525-56392525 on Build GRCh38
rs565393463 Chr.20: 56392397-56392397 on Build GRCh38
rs961237937 Chr.20: 56392728-56392728 on Build GRCh38
rs965442474 Chr.20: 56392436-56392436 on Build GRCh38
rs34554352 Chr.20: 56392703-56392703 on Build GRCh38
rs752408676 Chr.20: 56392538-56392541 on Build GRCh38
rs112169350 Chr.20: 56392587-56392587 on Build GRCh38
rs35400672 Chr.20: 56392567-56392570 on Build GRCh38
rs35340144 Chr.20: 56392488-56392488 on Build GRCh38
rs938714730 Chr.20: 56392557-56392557 on Build GRCh38
rs997110094 Chr.20: 56392469-56392469 on Build GRCh38
rs376132647 Chr.20: 56392482-56392482 on Build GRCh38
rs540510076 Chr.20: 56392382-56392382 on Build GRCh38
rs78654331 Chr.20: 56392367-56392367 on Build GRCh38
rs998534471 Chr.20: 56392413-56392413 on Build GRCh38
rs2104015 Chr.20: 56392556-56392556 on Build GRCh38
rs915881549 Chr.20: 56392755-56392755 on Build GRCh38
rs561225950 Chr.20: 56392446-56392446 on Build GRCh38
rs533869521 Chr.20: 56392699-56392699 on Build GRCh38
rs796213874 Chr.20: 56392563-56392566 on Build GRCh38
rs536830463 Chr.20: 56392543-56392543 on Build GRCh38
rs1028693853 Chr.20: 56392449-56392449 on Build GRCh38
rs554186280 Chr.20: 56392658-56392658 on Build GRCh38
rs938677004 Chr.20: 56392390-56392390 on Build GRCh38
rs879400614 Chr.20: 56392336-56392337 on Build GRCh38
rs1043221541 Chr.20: 56392637-56392637 on Build GRCh38
rs116572350 Chr.20: 56392365-56392365 on Build GRCh38
rs996925705 Chr.20: 56392676-56392676 on Build GRCh38
rs574404216 Chr.20: 56392377-56392377 on Build GRCh38
rs375264602 Chr.20: 56392684-56392684 on Build GRCh38
rs371892655 Chr.20: 56392617-56392617 on Build GRCh38
rs1005442055 Chr.20: 56392726-56392726 on Build GRCh38
rs557707600 Chr.20: 56392724-56392724 on Build GRCh38
rs558604955 Chr.20: 56392344-56392344 on Build GRCh38
rs922381763 Chr.20: 56392689-56392689 on Build GRCh38
rs774207893 Chr.20: 56392478-56392478 on Build GRCh38
rs1025050264 Chr.20: 56392749-56392749 on Build GRCh38

View Full Product Details