Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56397562-56398068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF1
Entrez Gene ID
1477
Gene Name
cleavage stimulation factor subunit 1
Gene Aliases
CstF-50, CstFp50
UniGene
Hs.172865
Gene Chromosome Location
Chr.20: 56392371-56404526 on Build GRCh38
Transcripts
NM_001033521.1 NM_001033522.1 NM_001324.2 XM_011528600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5733329 c.535C>T p.L179F Chr.20: 56397731-56397731 on Build GRCh38
4675227 c.634A>G p.K212E Chr.20: 56397830-56397830 on Build GRCh38
4675226 c.466G>A p.A156T Chr.20: 56397662-56397662 on Build GRCh38
331222 c.493G>A p.V165M Chr.20: 56397689-56397689 on Build GRCh38
4630598 c.630C>A p.A210A Chr.20: 56397826-56397826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763758306 Chr.20: 56397757-56397757 on Build GRCh38
rs770006666 Chr.20: 56397848-56397848 on Build GRCh38
rs112347691 Chr.20: 56397896-56397896 on Build GRCh38
rs544099784 Chr.20: 56397615-56397615 on Build GRCh38
rs763616564 Chr.20: 56397887-56397887 on Build GRCh38
rs772417622 Chr.20: 56397761-56397761 on Build GRCh38
rs774416044 Chr.20: 56397857-56397857 on Build GRCh38
rs913347309 Chr.20: 56398037-56398037 on Build GRCh38
rs759614037 Chr.20: 56397864-56397864 on Build GRCh38
rs1015096617 Chr.20: 56397980-56397980 on Build GRCh38
rs201966295 Chr.20: 56397751-56397751 on Build GRCh38
rs747230088 Chr.20: 56397815-56397815 on Build GRCh38
rs528527599 Chr.20: 56397884-56397884 on Build GRCh38
rs755764132 Chr.20: 56397620-56397620 on Build GRCh38
rs187829047 Chr.20: 56397976-56397976 on Build GRCh38
rs753496471 Chr.20: 56397893-56397893 on Build GRCh38
rs1048733740 Chr.20: 56397811-56397811 on Build GRCh38
rs775200221 Chr.20: 56397876-56397876 on Build GRCh38
rs200022027 Chr.20: 56397748-56397748 on Build GRCh38
rs879091865 Chr.20: 56397592-56397592 on Build GRCh38
rs980078059 Chr.20: 56397967-56397967 on Build GRCh38
rs776942602 Chr.20: 56397835-56397835 on Build GRCh38
rs376736562 Chr.20: 56397604-56397604 on Build GRCh38
rs150765899 Chr.20: 56397746-56397746 on Build GRCh38
rs1023525625 Chr.20: 56397938-56397938 on Build GRCh38
rs540022505 Chr.20: 56397817-56397817 on Build GRCh38
rs759346072 Chr.20: 56397607-56397607 on Build GRCh38
rs773864087 Chr.20: 56397888-56397889 on Build GRCh38
rs1004621013 Chr.20: 56397873-56397873 on Build GRCh38
rs1013869728 Chr.20: 56397901-56397901 on Build GRCh38
rs141380576 Chr.20: 56397727-56397727 on Build GRCh38
rs1040374502 Chr.20: 56397826-56397826 on Build GRCh38
rs34920028 Chr.20: 56397937-56397938 on Build GRCh38
rs971084879 Chr.20: 56398014-56398014 on Build GRCh38
rs767313357 Chr.20: 56397874-56397874 on Build GRCh38
rs879045352 Chr.20: 56397605-56397605 on Build GRCh38
rs887983857 Chr.20: 56397903-56397903 on Build GRCh38
rs1040627786 Chr.20: 56397667-56397667 on Build GRCh38
rs372778719 Chr.20: 56397771-56397771 on Build GRCh38
rs533574021 Chr.20: 56397726-56397726 on Build GRCh38
rs988893858 Chr.20: 56397992-56397992 on Build GRCh38
rs60370355 Chr.20: 56398036-56398036 on Build GRCh38
rs34484018 Chr.20: 56397658-56397659 on Build GRCh38
rs376042143 Chr.20: 56397634-56397634 on Build GRCh38
rs767990043 Chr.20: 56397802-56397802 on Build GRCh38
rs78726609 Chr.20: 56397649-56397649 on Build GRCh38
rs752390009 Chr.20: 56397619-56397619 on Build GRCh38
rs183434691 Chr.20: 56397881-56397881 on Build GRCh38
rs756795384 Chr.20: 56397969-56397969 on Build GRCh38
rs868690310 Chr.20: 56397777-56397777 on Build GRCh38
rs900767715 Chr.20: 56397831-56397831 on Build GRCh38
rs565268427 Chr.20: 56397948-56397948 on Build GRCh38
rs767275109 Chr.20: 56397613-56397613 on Build GRCh38
rs887498756 Chr.20: 56397855-56397855 on Build GRCh38
rs761916174 Chr.20: 56397847-56397847 on Build GRCh38
rs969298711 Chr.20: 56397946-56397946 on Build GRCh38
rs369490590 Chr.20: 56397661-56397661 on Build GRCh38

View Full Product Details