Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56403749-56404258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF1
Entrez Gene ID
1477
Gene Name
cleavage stimulation factor subunit 1
Gene Aliases
CstF-50, CstFp50
UniGene
Hs.172865
Gene Chromosome Location
Chr.20: 56392371-56404526 on Build GRCh38
Transcripts
NM_001033521.1 NM_001033522.1 NM_001324.2 XM_011528600.1

SNP Detail

SNP ID Chromosome Location
rs1014882900 Chr.20: 56403916-56403917 on Build GRCh38
rs761638765 Chr.20: 56403855-56403855 on Build GRCh38
rs768650822 Chr.20: 56403818-56403818 on Build GRCh38
rs538250906 Chr.20: 56403992-56403992 on Build GRCh38
rs958775064 Chr.20: 56403970-56403970 on Build GRCh38
rs944412005 Chr.20: 56403812-56403812 on Build GRCh38
rs545396193 Chr.20: 56403823-56403823 on Build GRCh38
rs780974195 Chr.20: 56404144-56404144 on Build GRCh38
rs995974759 Chr.20: 56403918-56403918 on Build GRCh38
rs1048805628 Chr.20: 56404139-56404139 on Build GRCh38
rs565582750 Chr.20: 56403885-56403885 on Build GRCh38
rs923921154 Chr.20: 56404069-56404069 on Build GRCh38
rs148184670 Chr.20: 56403822-56403822 on Build GRCh38
rs542730432 Chr.20: 56403780-56403780 on Build GRCh38
rs146945258 Chr.20: 56404088-56404088 on Build GRCh38
rs6024853 Chr.20: 56403779-56403779 on Build GRCh38
rs989016548 Chr.20: 56404059-56404059 on Build GRCh38
rs902740376 Chr.20: 56403859-56403859 on Build GRCh38
rs115138879 Chr.20: 56403941-56403941 on Build GRCh38
rs7806 Chr.20: 56404115-56404115 on Build GRCh38
rs546273916 Chr.20: 56403976-56403976 on Build GRCh38
rs1014566326 Chr.20: 56404224-56404224 on Build GRCh38
rs190240313 Chr.20: 56403944-56403944 on Build GRCh38
rs6092312 Chr.20: 56404163-56404163 on Build GRCh38
rs546045358 Chr.20: 56403950-56403950 on Build GRCh38
rs1024714785 Chr.20: 56404184-56404184 on Build GRCh38
rs752969917 Chr.20: 56404101-56404101 on Build GRCh38
rs900254779 Chr.20: 56403828-56403828 on Build GRCh38
rs755831910 Chr.20: 56404203-56404203 on Build GRCh38
rs887597949 Chr.20: 56404103-56404103 on Build GRCh38
rs773481374 Chr.20: 56403936-56403936 on Build GRCh38
rs749112247 Chr.20: 56403806-56403806 on Build GRCh38
rs554775062 Chr.20: 56404113-56404113 on Build GRCh38
rs1027357474 Chr.20: 56403934-56403934 on Build GRCh38
rs996710027 Chr.20: 56404133-56404133 on Build GRCh38
rs887639174 Chr.20: 56403869-56403869 on Build GRCh38
rs6099131 Chr.20: 56404215-56404215 on Build GRCh38
rs1004470910 Chr.20: 56403943-56403943 on Build GRCh38
rs938131571 Chr.20: 56403807-56403807 on Build GRCh38
rs1042390502 Chr.20: 56404027-56404027 on Build GRCh38
rs773966690 Chr.20: 56403829-56403829 on Build GRCh38
rs981746716 Chr.20: 56403955-56403955 on Build GRCh38
rs996002472 Chr.20: 56403847-56403849 on Build GRCh38
rs149041179 Chr.20: 56403775-56403775 on Build GRCh38
rs944339976 Chr.20: 56404084-56404084 on Build GRCh38
rs867533925 Chr.20: 56404020-56404020 on Build GRCh38
rs931703769 Chr.20: 56404102-56404102 on Build GRCh38
rs745582705 Chr.20: 56404159-56404159 on Build GRCh38
rs531073440 Chr.20: 56403937-56403937 on Build GRCh38
rs771299882 Chr.20: 56403772-56403772 on Build GRCh38
rs1014234593 Chr.20: 56404162-56404162 on Build GRCh38
rs760835133 Chr.20: 56403778-56403778 on Build GRCh38
rs1048842161 Chr.20: 56404103-56404104 on Build GRCh38
rs182315222 Chr.20: 56403993-56403993 on Build GRCh38
rs375719269 Chr.20: 56403776-56403776 on Build GRCh38
rs1033577887 Chr.20: 56404013-56404013 on Build GRCh38
rs140219472 Chr.20: 56403969-56403969 on Build GRCh38
rs1002024153 Chr.20: 56404225-56404225 on Build GRCh38
rs905887209 Chr.20: 56404189-56404189 on Build GRCh38
rs900655799 Chr.20: 56403917-56403917 on Build GRCh38
rs1009500268 Chr.20: 56403874-56403874 on Build GRCh38
rs765801662 Chr.20: 56404064-56404064 on Build GRCh38
rs75407746 Chr.20: 56404025-56404025 on Build GRCh38
rs16979884 Chr.20: 56403979-56403979 on Build GRCh38
rs779371347 Chr.20: 56404207-56404207 on Build GRCh38
rs533942892 Chr.20: 56404126-56404126 on Build GRCh38
rs143018521 Chr.20: 56403782-56403782 on Build GRCh38

View Full Product Details