Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58996713-58997224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSZ
Entrez Gene ID
1522
Gene Name
cathepsin Z
Gene Aliases
CTSX
UniGene
Hs.252549
Gene Chromosome Location
Chr.20: 58995187-59007254 on Build GRCh38
Transcripts
NM_001336.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7363288 c.677C>G p.T226S Chr.20: 58996763-58996763 on Build GRCh38
6637848 c.673T>C p.Y225H Chr.20: 58996767-58996767 on Build GRCh38
1172733 c.658G>C p.E220Q Chr.20: 58996782-58996782 on Build GRCh38
1751516 c.687C>T p.I229I Chr.20: 58996753-58996753 on Build GRCh38
6269015 c.639C>A p.S213R Chr.20: 58996801-58996801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535937184 Chr.20: 58997127-58997127 on Build GRCh38
rs750235482 Chr.20: 58996770-58996770 on Build GRCh38
rs564596357 Chr.20: 58996883-58996883 on Build GRCh38
rs574938687 Chr.20: 58996743-58996743 on Build GRCh38
rs754493393 Chr.20: 58996806-58996806 on Build GRCh38
rs534917397 Chr.20: 58997071-58997071 on Build GRCh38
rs914095803 Chr.20: 58997136-58997136 on Build GRCh38
rs56037144 Chr.20: 58997183-58997184 on Build GRCh38
rs564941820 Chr.20: 58996972-58996972 on Build GRCh38
rs749673981 Chr.20: 58996853-58996853 on Build GRCh38
rs769485950 Chr.20: 58997018-58997018 on Build GRCh38
rs765113304 Chr.20: 58996765-58996765 on Build GRCh38
rs970357150 Chr.20: 58997087-58997087 on Build GRCh38
rs762674508 Chr.20: 58996783-58996783 on Build GRCh38
rs867023336 Chr.20: 58997155-58997155 on Build GRCh38
rs374306654 Chr.20: 58996923-58996923 on Build GRCh38
rs775093130 Chr.20: 58997163-58997165 on Build GRCh38
rs989040696 Chr.20: 58997017-58997017 on Build GRCh38
rs912152212 Chr.20: 58997031-58997031 on Build GRCh38
rs777053367 Chr.20: 58997090-58997090 on Build GRCh38
rs772171555 Chr.20: 58996761-58996761 on Build GRCh38
rs1051996768 Chr.20: 58996988-58996988 on Build GRCh38
rs149057628 Chr.20: 58996756-58996756 on Build GRCh38
rs988218883 Chr.20: 58997132-58997132 on Build GRCh38
rs759034997 Chr.20: 58997164-58997165 on Build GRCh38
rs933461971 Chr.20: 58997143-58997143 on Build GRCh38
rs752212937 Chr.20: 58996834-58996834 on Build GRCh38
rs944823117 Chr.20: 58997060-58997060 on Build GRCh38
rs910822826 Chr.20: 58997198-58997198 on Build GRCh38
rs568309034 Chr.20: 58997162-58997164 on Build GRCh38
rs10582096 Chr.20: 58997184-58997184 on Build GRCh38
rs959826393 Chr.20: 58996934-58996934 on Build GRCh38
rs527282200 Chr.20: 58997104-58997104 on Build GRCh38
rs780719726 Chr.20: 58996809-58996809 on Build GRCh38
rs561951554 Chr.20: 58996955-58996955 on Build GRCh38
rs141912408 Chr.20: 58996790-58996790 on Build GRCh38
rs890601881 Chr.20: 58996784-58996784 on Build GRCh38
rs529739774 Chr.20: 58996968-58996968 on Build GRCh38
rs759594176 Chr.20: 58996758-58996758 on Build GRCh38
rs934335118 Chr.20: 58997009-58997009 on Build GRCh38
rs895768688 Chr.20: 58997184-58997184 on Build GRCh38
rs186652190 Chr.20: 58996978-58996978 on Build GRCh38
rs1014156493 Chr.20: 58997080-58997080 on Build GRCh38
rs146697212 Chr.20: 58997091-58997091 on Build GRCh38
rs370964927 Chr.20: 58997048-58997048 on Build GRCh38
rs756710210 Chr.20: 58996843-58996843 on Build GRCh38
rs900582226 Chr.20: 58997072-58997072 on Build GRCh38
rs898924137 Chr.20: 58997039-58997039 on Build GRCh38
rs766010097 Chr.20: 58996789-58996789 on Build GRCh38
rs958180703 Chr.20: 58997109-58997109 on Build GRCh38
rs1052611615 Chr.20: 58996936-58996936 on Build GRCh38
rs969403592 Chr.20: 58996891-58996891 on Build GRCh38
rs760133237 Chr.20: 58997162-58997165 on Build GRCh38
rs1053269661 Chr.20: 58996781-58996781 on Build GRCh38
rs959849490 Chr.20: 58997068-58997068 on Build GRCh38
rs778366073 Chr.20: 58996852-58996852 on Build GRCh38
rs570612720 Chr.20: 58997067-58997067 on Build GRCh38
rs143309663 Chr.20: 58996894-58996894 on Build GRCh38
rs375989357 Chr.20: 58996746-58996746 on Build GRCh38
rs1007238295 Chr.20: 58997030-58997030 on Build GRCh38
rs888806575 Chr.20: 58997020-58997020 on Build GRCh38
rs776475754 Chr.20: 58996762-58996762 on Build GRCh38
rs917940753 Chr.20: 58996885-58996885 on Build GRCh38
rs568302308 Chr.20: 58997114-58997114 on Build GRCh38
rs369765479 Chr.20: 58996747-58996747 on Build GRCh38
rs546455151 Chr.20: 58996881-58996881 on Build GRCh38
rs886418023 Chr.20: 58997103-58997103 on Build GRCh38
rs1049624499 Chr.20: 58997095-58997095 on Build GRCh38
rs1004056693 Chr.20: 58996906-58996906 on Build GRCh38
rs1015482203 Chr.20: 58996933-58996933 on Build GRCh38
rs925407322 Chr.20: 58996895-58996895 on Build GRCh38
rs62639970 Chr.20: 58996763-58996763 on Build GRCh38
rs552164040 Chr.20: 58997049-58997049 on Build GRCh38
rs925672145 Chr.20: 58996987-58996987 on Build GRCh38
rs1037735327 Chr.20: 58997034-58997034 on Build GRCh38
rs1051903567 Chr.20: 58997164-58997164 on Build GRCh38
rs530394973 Chr.20: 58997029-58997029 on Build GRCh38
rs116526567 Chr.20: 58996969-58996969 on Build GRCh38
rs1036873191 Chr.20: 58997156-58997156 on Build GRCh38
rs1022658889 Chr.20: 58996963-58996963 on Build GRCh38
rs11292326 Chr.20: 58997162-58997162 on Build GRCh38
rs1022851820 Chr.20: 58997086-58997086 on Build GRCh38

View Full Product Details