Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 61894830-61895328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH4
Entrez Gene ID
1002
Gene Name
cadherin 4
Gene Aliases
CAD4, R-CAD, RCAD
UniGene
Hs.473231
Gene Chromosome Location
Chr.20: 61252426-61940617 on Build GRCh38
Transcripts
NM_001252338.2 NM_001252339.2 NM_001794.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6055985 c.1093G>A p.E365K Chr.20: 61894952-61894952 on Build GRCh38
1412873 c.1176T>G p.F392L Chr.20: 61895035-61895035 on Build GRCh38
5392492 c.1074C>T p.I358I Chr.20: 61894933-61894933 on Build GRCh38
1412872 c.1141A>G p.T381A Chr.20: 61895000-61895000 on Build GRCh38
4440614 c.1104C>G p.L368L Chr.20: 61894963-61894963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149158235 Chr.20: 61895039-61895039 on Build GRCh38
rs763599303 Chr.20: 61895007-61895007 on Build GRCh38
rs548891820 Chr.20: 61894879-61894879 on Build GRCh38
rs552314715 Chr.20: 61895221-61895221 on Build GRCh38
rs207477702 Chr.20: 61894922-61894922 on Build GRCh38
rs201949663 Chr.20: 61894895-61894895 on Build GRCh38
rs893532489 Chr.20: 61895110-61895110 on Build GRCh38
rs773222468 Chr.20: 61894994-61894994 on Build GRCh38
rs575470738 Chr.20: 61895094-61895094 on Build GRCh38
rs751881 Chr.20: 61895288-61895288 on Build GRCh38
rs751454638 Chr.20: 61895088-61895088 on Build GRCh38
rs760739744 Chr.20: 61894995-61894995 on Build GRCh38
rs937486396 Chr.20: 61895164-61895164 on Build GRCh38
rs757392743 Chr.20: 61895055-61895055 on Build GRCh38
rs368223319 Chr.20: 61894891-61894891 on Build GRCh38
rs750450917 Chr.20: 61895272-61895272 on Build GRCh38
rs1027461647 Chr.20: 61895186-61895186 on Build GRCh38
rs373483932 Chr.20: 61895046-61895046 on Build GRCh38
rs753824315 Chr.20: 61895090-61895090 on Build GRCh38
rs780170651 Chr.20: 61894933-61894933 on Build GRCh38
rs760212150 Chr.20: 61894872-61894872 on Build GRCh38
rs761416568 Chr.20: 61895075-61895075 on Build GRCh38
rs952761070 Chr.20: 61895191-61895191 on Build GRCh38
rs146626878 Chr.20: 61895219-61895219 on Build GRCh38
rs776190389 Chr.20: 61895010-61895010 on Build GRCh38
rs1056624246 Chr.20: 61895212-61895212 on Build GRCh38
rs573461151 Chr.20: 61895265-61895265 on Build GRCh38
rs772441780 Chr.20: 61895065-61895065 on Build GRCh38
rs756029114 Chr.20: 61895014-61895014 on Build GRCh38
rs1052583228 Chr.20: 61895260-61895260 on Build GRCh38
rs888886542 Chr.20: 61895268-61895268 on Build GRCh38
rs376487285 Chr.20: 61895093-61895093 on Build GRCh38
rs953377525 Chr.20: 61894959-61894959 on Build GRCh38
rs368396310 Chr.20: 61895261-61895261 on Build GRCh38
rs778071640 Chr.20: 61894941-61894941 on Build GRCh38
rs540041409 Chr.20: 61894935-61894935 on Build GRCh38
rs918656073 Chr.20: 61895045-61895045 on Build GRCh38
rs138274161 Chr.20: 61895012-61895012 on Build GRCh38
rs758237001 Chr.20: 61894924-61894924 on Build GRCh38
rs148554450 Chr.20: 61895133-61895133 on Build GRCh38
rs752397984 Chr.20: 61895146-61895146 on Build GRCh38
rs1025312521 Chr.20: 61895122-61895123 on Build GRCh38
rs769623176 Chr.20: 61895076-61895078 on Build GRCh38
rs752676634 Chr.20: 61894998-61894998 on Build GRCh38
rs774560707 Chr.20: 61894990-61894990 on Build GRCh38
rs542643194 Chr.20: 61895188-61895188 on Build GRCh38
rs935584487 Chr.20: 61895256-61895256 on Build GRCh38
rs779229235 Chr.20: 61895059-61895059 on Build GRCh38
rs748187890 Chr.20: 61894934-61894934 on Build GRCh38
rs917363344 Chr.20: 61895187-61895187 on Build GRCh38
rs111622539 Chr.20: 61894856-61894856 on Build GRCh38
rs145754297 Chr.20: 61895002-61895002 on Build GRCh38
rs999730771 Chr.20: 61894857-61894857 on Build GRCh38
rs369270154 Chr.20: 61895038-61895038 on Build GRCh38
rs868646026 Chr.20: 61894996-61894996 on Build GRCh38
rs766268812 Chr.20: 61895086-61895086 on Build GRCh38
rs772928641 Chr.20: 61895083-61895083 on Build GRCh38
rs534389589 Chr.20: 61895243-61895243 on Build GRCh38
rs535401599 Chr.20: 61895279-61895279 on Build GRCh38
rs368680932 Chr.20: 61895081-61895081 on Build GRCh38
rs865813105 Chr.20: 61894956-61894956 on Build GRCh38
rs113714586 Chr.20: 61895054-61895054 on Build GRCh38
rs946900613 Chr.20: 61895262-61895262 on Build GRCh38
rs769210187 Chr.20: 61895070-61895070 on Build GRCh38
rs371519806 Chr.20: 61895273-61895273 on Build GRCh38
rs969415174 Chr.20: 61895242-61895242 on Build GRCh38
rs1040455193 Chr.20: 61895278-61895278 on Build GRCh38
rs904509102 Chr.20: 61895226-61895226 on Build GRCh38
rs570754916 Chr.20: 61895228-61895228 on Build GRCh38
rs553325921 Chr.20: 61895116-61895116 on Build GRCh38
rs775910937 Chr.20: 61895066-61895066 on Build GRCh38
rs750281319 Chr.20: 61894921-61894921 on Build GRCh38
rs368574186 Chr.20: 61894980-61894981 on Build GRCh38
rs143299049 Chr.20: 61895047-61895047 on Build GRCh38
rs375994111 Chr.20: 61895017-61895017 on Build GRCh38
rs144440239 Chr.20: 61895144-61895144 on Build GRCh38
rs149375024 Chr.20: 61895001-61895001 on Build GRCh38
rs771237720 Chr.20: 61894983-61894983 on Build GRCh38
rs78938844 Chr.20: 61894982-61894982 on Build GRCh38
rs747510435 Chr.20: 61895068-61895068 on Build GRCh38
rs557184484 Chr.20: 61895089-61895089 on Build GRCh38
rs755356538 Chr.20: 61895111-61895111 on Build GRCh38
rs745362438 Chr.20: 61894882-61894883 on Build GRCh38
rs750144734 Chr.20: 61895020-61895020 on Build GRCh38
rs187801920 Chr.20: 61894939-61894939 on Build GRCh38
rs749463684 Chr.20: 61894954-61894954 on Build GRCh38
rs201033456 Chr.20: 61895026-61895026 on Build GRCh38
rs184489399 Chr.20: 61895155-61895155 on Build GRCh38
rs928225359 Chr.20: 61895250-61895250 on Build GRCh38
rs985215618 Chr.20: 61894854-61894854 on Build GRCh38
rs369263785 Chr.20: 61895061-61895061 on Build GRCh38
rs545974597 Chr.20: 61895106-61895106 on Build GRCh38
rs756273047 Chr.20: 61894938-61894938 on Build GRCh38
rs373333908 Chr.20: 61895074-61895074 on Build GRCh38
rs946131361 Chr.20: 61895211-61895211 on Build GRCh38
rs146877847 Chr.20: 61894925-61894925 on Build GRCh38
rs775253758 Chr.20: 61895080-61895081 on Build GRCh38
rs1040892295 Chr.20: 61895266-61895266 on Build GRCh38
rs192288459 Chr.20: 61895134-61895134 on Build GRCh38
rs993260989 Chr.20: 61895165-61895165 on Build GRCh38
rs189768893 Chr.20: 61895161-61895161 on Build GRCh38
rs541740140 Chr.20: 61895084-61895084 on Build GRCh38

View Full Product Details