Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 61910031-61910549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH4
Entrez Gene ID
1002
Gene Name
cadherin 4
Gene Aliases
CAD4, R-CAD, RCAD
UniGene
Hs.473231
Gene Chromosome Location
Chr.20: 61252426-61940617 on Build GRCh38
Transcripts
NM_001252338.2 NM_001252339.2 NM_001794.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625097 c.1292G>A p.R431H Chr.20: 61910525-61910525 on Build GRCh38
1412875 c.1249G>A p.D417N Chr.20: 61910482-61910482 on Build GRCh38
5894070 c.1243G>A p.V415M Chr.20: 61910476-61910476 on Build GRCh38
5858244 c.1199_1209del11 p.V401fs*131 Chr.20: 61910432-61910442 on Build GRCh38
3548533 c.1228G>A p.V410I Chr.20: 61910461-61910461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753258436 Chr.20: 61910331-61910331 on Build GRCh38
rs372929561 Chr.20: 61910215-61910215 on Build GRCh38
rs759348321 Chr.20: 61910461-61910461 on Build GRCh38
rs377409847 Chr.20: 61910394-61910394 on Build GRCh38
rs921030528 Chr.20: 61910140-61910140 on Build GRCh38
rs777312618 Chr.20: 61910451-61910451 on Build GRCh38
rs757594437 Chr.20: 61910452-61910452 on Build GRCh38
rs775216668 Chr.20: 61910464-61910464 on Build GRCh38
rs3765600 Chr.20: 61910338-61910338 on Build GRCh38
rs1023111489 Chr.20: 61910281-61910281 on Build GRCh38
rs147467462 Chr.20: 61910525-61910525 on Build GRCh38
rs572783423 Chr.20: 61910104-61910104 on Build GRCh38
rs761734131 Chr.20: 61910432-61910432 on Build GRCh38
rs557261809 Chr.20: 61910207-61910207 on Build GRCh38
rs900633629 Chr.20: 61910100-61910100 on Build GRCh38
rs766529605 Chr.20: 61910485-61910485 on Build GRCh38
rs1038666419 Chr.20: 61910236-61910236 on Build GRCh38
rs751872489 Chr.20: 61910447-61910447 on Build GRCh38
rs1034003486 Chr.20: 61910377-61910377 on Build GRCh38
rs368553704 Chr.20: 61910446-61910446 on Build GRCh38
rs568637646 Chr.20: 61910170-61910170 on Build GRCh38
rs763041307 Chr.20: 61910480-61910480 on Build GRCh38
rs377652936 Chr.20: 61910127-61910127 on Build GRCh38
rs923904913 Chr.20: 61910285-61910285 on Build GRCh38
rs374255970 Chr.20: 61910103-61910103 on Build GRCh38
rs375893301 Chr.20: 61910440-61910440 on Build GRCh38
rs1973940 Chr.20: 61910152-61910152 on Build GRCh38
rs986584668 Chr.20: 61910384-61910384 on Build GRCh38
rs564200973 Chr.20: 61910287-61910287 on Build GRCh38
rs775529184 Chr.20: 61910092-61910092 on Build GRCh38
rs151072387 Chr.20: 61910449-61910449 on Build GRCh38
rs769348039 Chr.20: 61910409-61910409 on Build GRCh38
rs570631094 Chr.20: 61910146-61910146 on Build GRCh38
rs747551658 Chr.20: 61910399-61910399 on Build GRCh38
rs865878611 Chr.20: 61910516-61910516 on Build GRCh38
rs759439816 Chr.20: 61910416-61910416 on Build GRCh38
rs780384689 Chr.20: 61910386-61910386 on Build GRCh38
rs971468948 Chr.20: 61910057-61910058 on Build GRCh38
rs376532319 Chr.20: 61910439-61910439 on Build GRCh38
rs557605363 Chr.20: 61910458-61910458 on Build GRCh38
rs924042168 Chr.20: 61910433-61910433 on Build GRCh38
rs758884905 Chr.20: 61910334-61910334 on Build GRCh38
rs750664105 Chr.20: 61910378-61910378 on Build GRCh38
rs756610425 Chr.20: 61910508-61910508 on Build GRCh38
rs529167243 Chr.20: 61910321-61910321 on Build GRCh38
rs187215937 Chr.20: 61910119-61910119 on Build GRCh38
rs149552640 Chr.20: 61910254-61910256 on Build GRCh38
rs1004674502 Chr.20: 61910148-61910148 on Build GRCh38
rs751782763 Chr.20: 61910486-61910486 on Build GRCh38
rs747413747 Chr.20: 61910388-61910388 on Build GRCh38
rs562642027 Chr.20: 61910356-61910356 on Build GRCh38
rs753047655 Chr.20: 61910506-61910506 on Build GRCh38
rs370373224 Chr.20: 61910431-61910431 on Build GRCh38
rs370306312 Chr.20: 61910310-61910310 on Build GRCh38
rs370432072 Chr.20: 61910395-61910395 on Build GRCh38
rs11699608 Chr.20: 61910187-61910187 on Build GRCh38
rs755256769 Chr.20: 61910498-61910498 on Build GRCh38
rs900769232 Chr.20: 61910295-61910295 on Build GRCh38
rs373098048 Chr.20: 61910389-61910389 on Build GRCh38
rs1014995228 Chr.20: 61910151-61910151 on Build GRCh38
rs1034245767 Chr.20: 61910182-61910182 on Build GRCh38
rs1019727285 Chr.20: 61910477-61910477 on Build GRCh38
rs769245029 Chr.20: 61910085-61910085 on Build GRCh38
rs552984117 Chr.20: 61910160-61910160 on Build GRCh38
rs765357196 Chr.20: 61910435-61910435 on Build GRCh38
rs1054017997 Chr.20: 61910292-61910292 on Build GRCh38
rs141404094 Chr.20: 61910503-61910503 on Build GRCh38
rs563164236 Chr.20: 61910128-61910130 on Build GRCh38
rs1054048870 Chr.20: 61910093-61910093 on Build GRCh38
rs562573659 Chr.20: 61910296-61910296 on Build GRCh38
rs575748178 Chr.20: 61910225-61910225 on Build GRCh38
rs766760153 Chr.20: 61910229-61910229 on Build GRCh38
rs114905159 Chr.20: 61910230-61910230 on Build GRCh38
rs780958655 Chr.20: 61910054-61910054 on Build GRCh38
rs770751711 Chr.20: 61910412-61910412 on Build GRCh38
rs540203825 Chr.20: 61910335-61910335 on Build GRCh38
rs778178782 Chr.20: 61910518-61910518 on Build GRCh38
rs141026097 Chr.20: 61910475-61910475 on Build GRCh38
rs767517266 Chr.20: 61910419-61910419 on Build GRCh38
rs750175283 Chr.20: 61910421-61910421 on Build GRCh38
rs1024553500 Chr.20: 61910410-61910410 on Build GRCh38
rs377535852 Chr.20: 61910347-61910347 on Build GRCh38
rs199603882 Chr.20: 61910413-61910413 on Build GRCh38
rs957997062 Chr.20: 61910434-61910434 on Build GRCh38
rs750462295 Chr.20: 61910478-61910478 on Build GRCh38
rs555959239 Chr.20: 61910463-61910463 on Build GRCh38
rs568480999 Chr.20: 61910448-61910448 on Build GRCh38
rs374897454 Chr.20: 61910411-61910411 on Build GRCh38
rs145765240 Chr.20: 61910335-61910337 on Build GRCh38
rs561715311 Chr.20: 61910311-61910311 on Build GRCh38
rs533119956 Chr.20: 61910359-61910359 on Build GRCh38
rs1776237 Chr.20: 61910271-61910271 on Build GRCh38
rs1052361386 Chr.20: 61910303-61910303 on Build GRCh38
rs370611196 Chr.20: 61910382-61910382 on Build GRCh38
rs770671856 Chr.20: 61910457-61910457 on Build GRCh38
rs902557445 Chr.20: 61910083-61910083 on Build GRCh38
rs949498656 Chr.20: 61910254-61910254 on Build GRCh38
rs939662090 Chr.20: 61910233-61910233 on Build GRCh38
rs768626140 Chr.20: 61910128-61910128 on Build GRCh38

View Full Product Details