Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 77806977-77807481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIGA1
Entrez Gene ID
374986
Gene Name
mitoguardin 1
Gene Aliases
FAM73A
UniGene
Hs.437755
Gene Chromosome Location
Chr.1: 77779624-77879540 on Build GRCh38
Transcripts
NM_001270384.1 NM_198549.3 XM_005270846.4 XM_005270847.4 XM_005270848.4 XM_011541422.2 XM_017001227.1 XM_017001228.1 XM_017001229.1 XM_017001230.1 XM_017001231.1 XM_017001232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4009973 c.541G>A p.A181T Chr.1: 77807005-77807005 on Build GRCh38
3492489 c.616C>T p.P206S Chr.1: 77807080-77807080 on Build GRCh38
682037 c.587T>C p.I196T Chr.1: 77807051-77807051 on Build GRCh38
5934291 c.560C>A p.S187Y Chr.1: 77807024-77807024 on Build GRCh38
1158441 c.637+1G>A p.? Chr.1: 77807102-77807102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919512972 Chr.1: 77807321-77807321 on Build GRCh38
rs199799238 Chr.1: 77807079-77807079 on Build GRCh38
rs968066127 Chr.1: 77807248-77807248 on Build GRCh38
rs981288990 Chr.1: 77807278-77807278 on Build GRCh38
rs112864778 Chr.1: 77807126-77807126 on Build GRCh38
rs542587816 Chr.1: 77807444-77807444 on Build GRCh38
rs1043980943 Chr.1: 77807030-77807030 on Build GRCh38
rs866586992 Chr.1: 77807306-77807306 on Build GRCh38
rs758536657 Chr.1: 77807433-77807433 on Build GRCh38
rs929524890 Chr.1: 77807326-77807326 on Build GRCh38
rs577275813 Chr.1: 77807279-77807279 on Build GRCh38
rs1002816669 Chr.1: 77807121-77807122 on Build GRCh38
rs535405244 Chr.1: 77807204-77807204 on Build GRCh38
rs549397592 Chr.1: 77807161-77807161 on Build GRCh38
rs913888290 Chr.1: 77807333-77807334 on Build GRCh38
rs370102859 Chr.1: 77807139-77807139 on Build GRCh38
rs533989510 Chr.1: 77807225-77807225 on Build GRCh38
rs746073640 Chr.1: 77807087-77807092 on Build GRCh38
rs887553094 Chr.1: 77807156-77807156 on Build GRCh38
rs901707447 Chr.1: 77807202-77807202 on Build GRCh38
rs61777142 Chr.1: 77807440-77807440 on Build GRCh38
rs1038745898 Chr.1: 77807192-77807192 on Build GRCh38
rs905346300 Chr.1: 77807112-77807112 on Build GRCh38
rs1052365297 Chr.1: 77807329-77807329 on Build GRCh38
rs537460632 Chr.1: 77807130-77807130 on Build GRCh38
rs200550339 Chr.1: 77807004-77807004 on Build GRCh38
rs376909812 Chr.1: 77807291-77807291 on Build GRCh38
rs773168463 Chr.1: 77807045-77807045 on Build GRCh38
rs752465268 Chr.1: 77807223-77807223 on Build GRCh38
rs759827480 Chr.1: 77807099-77807099 on Build GRCh38
rs771693019 Chr.1: 77807024-77807024 on Build GRCh38
rs560610749 Chr.1: 77807319-77807319 on Build GRCh38
rs760651456 Chr.1: 77807005-77807005 on Build GRCh38
rs562900090 Chr.1: 77807411-77807411 on Build GRCh38
rs779200199 Chr.1: 77807001-77807001 on Build GRCh38
rs759029990 Chr.1: 77807105-77807105 on Build GRCh38
rs753372106 Chr.1: 77807144-77807144 on Build GRCh38
rs567990661 Chr.1: 77807190-77807190 on Build GRCh38
rs993128699 Chr.1: 77807184-77807184 on Build GRCh38
rs1009571074 Chr.1: 77807224-77807224 on Build GRCh38
rs1007692871 Chr.1: 77807178-77807178 on Build GRCh38
rs781658963 Chr.1: 77807290-77807290 on Build GRCh38
rs946750191 Chr.1: 77807334-77807334 on Build GRCh38
rs142939026 Chr.1: 77807236-77807236 on Build GRCh38
rs148559892 Chr.1: 77807222-77807222 on Build GRCh38
rs535174709 Chr.1: 77807317-77807317 on Build GRCh38
rs4949822 Chr.1: 77807217-77807217 on Build GRCh38
rs778773684 Chr.1: 77807009-77807009 on Build GRCh38
rs1043973358 Chr.1: 77807420-77807420 on Build GRCh38
rs770753527 Chr.1: 77807092-77807092 on Build GRCh38
rs984487411 Chr.1: 77807352-77807352 on Build GRCh38
rs112576794 Chr.1: 77807377-77807377 on Build GRCh38
rs373400354 Chr.1: 77807094-77807094 on Build GRCh38
rs757550462 Chr.1: 77807282-77807282 on Build GRCh38
rs12405797 Chr.1: 77807307-77807307 on Build GRCh38
rs910495027 Chr.1: 77807231-77807231 on Build GRCh38
rs763547498 Chr.1: 77807137-77807137 on Build GRCh38
rs375352889 Chr.1: 77807125-77807126 on Build GRCh38
rs865908328 Chr.1: 77807455-77807455 on Build GRCh38
rs144517609 Chr.1: 77807113-77807113 on Build GRCh38
rs748129067 Chr.1: 77807015-77807015 on Build GRCh38

View Full Product Details